Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 121416018-121416598 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KDM2B
Entrez Gene ID
84678
Gene Name
lysine demethylase 2B
Gene Aliases
CXXC2, FBXL10, Fbl10, JHDM1B, PCCX2
UniGene
Hs.524800
Gene Chromosome Location
Chr.12: 121402289-121582268 on Build GRCh38
Transcripts
XM_011538867.2 NM_001256858.1 NM_025126.3 NM_194271.2

Gene Symbol
RNF34
Entrez Gene ID
80196
Gene Name
ring finger protein 34
Gene Aliases
CARP-1, CARP1, RFI, RIF, RIFF, hRFI
UniGene
Hs.292804
Gene Chromosome Location
Chr.12: 121400083-121424353 on Build GRCh38
Transcripts
XM_011538867.2 NM_001256858.1 NM_025126.3 NM_194271.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6767922 c.109G>A p.G37S Chr.12: 121416261-121416261 on Build GRCh38
4039451 c.164C>T p.T55M Chr.12: 121416316-121416316 on Build GRCh38
1605666 c.45G>A p.L15L Chr.12: 121416197-121416197 on Build GRCh38
6767921 c.112G>A p.G38S Chr.12: 121416261-121416261 on Build GRCh38
1586290 c.168A>C p.E56D Chr.12: 121416320-121416320 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782326655 Chr.12: 121416147-121416147 on Build GRCh38
rs747981257 Chr.12: 121416383-121416383 on Build GRCh38
rs782392508 Chr.12: 121416296-121416296 on Build GRCh38
rs555501715 Chr.12: 121416231-121416231 on Build GRCh38
rs188015976 Chr.12: 121416398-121416398 on Build GRCh38
rs782367588 Chr.12: 121416428-121416428 on Build GRCh38
rs782631536 Chr.12: 121416130-121416130 on Build GRCh38
rs781859896 Chr.12: 121416200-121416200 on Build GRCh38
rs782455220 Chr.12: 121416399-121416399 on Build GRCh38
rs782213405 Chr.12: 121416414-121416414 on Build GRCh38
rs782706106 Chr.12: 121416230-121416230 on Build GRCh38
rs1042649056 Chr.12: 121416495-121416495 on Build GRCh38
rs1025055564 Chr.12: 121416549-121416549 on Build GRCh38
rs369500317 Chr.12: 121416386-121416386 on Build GRCh38
rs1032446702 Chr.12: 121416071-121416071 on Build GRCh38
rs782452243 Chr.12: 121416368-121416368 on Build GRCh38
rs782816324 Chr.12: 121416195-121416195 on Build GRCh38
rs531386204 Chr.12: 121416521-121416521 on Build GRCh38
rs145495936 Chr.12: 121416207-121416207 on Build GRCh38
rs782669486 Chr.12: 121416501-121416501 on Build GRCh38
rs112773484 Chr.12: 121416261-121416261 on Build GRCh38
rs537636270 Chr.12: 121416170-121416170 on Build GRCh38
rs782178280 Chr.12: 121416139-121416139 on Build GRCh38
rs1007817101 Chr.12: 121416492-121416492 on Build GRCh38
rs782191729 Chr.12: 121416266-121416266 on Build GRCh38
rs782648816 Chr.12: 121416253-121416253 on Build GRCh38
rs781926683 Chr.12: 121416190-121416190 on Build GRCh38
rs781929620 Chr.12: 121416077-121416077 on Build GRCh38
rs782086917 Chr.12: 121416260-121416260 on Build GRCh38
rs781945749 Chr.12: 121416175-121416175 on Build GRCh38
rs377206024 Chr.12: 121416213-121416213 on Build GRCh38
rs890827560 Chr.12: 121416397-121416397 on Build GRCh38
rs370393586 Chr.12: 121416312-121416312 on Build GRCh38
rs553418450 Chr.12: 121416298-121416298 on Build GRCh38
rs782402058 Chr.12: 121416274-121416274 on Build GRCh38
rs782820111 Chr.12: 121416341-121416341 on Build GRCh38
rs782664752 Chr.12: 121416400-121416400 on Build GRCh38
rs374454541 Chr.12: 121416356-121416356 on Build GRCh38
rs781845352 Chr.12: 121416216-121416216 on Build GRCh38
rs557821046 Chr.12: 121416085-121416085 on Build GRCh38
rs753331968 Chr.12: 121416079-121416079 on Build GRCh38
rs782451830 Chr.12: 121416229-121416229 on Build GRCh38
rs781790379 Chr.12: 121416362-121416362 on Build GRCh38
rs782026372 Chr.12: 121416287-121416287 on Build GRCh38
rs568859609 Chr.12: 121416089-121416089 on Build GRCh38
rs200788702 Chr.12: 121416290-121416290 on Build GRCh38
rs782099773 Chr.12: 121416161-121416161 on Build GRCh38
rs144086612 Chr.12: 121416329-121416329 on Build GRCh38
rs781520353 Chr.12: 121416532-121416532 on Build GRCh38
rs192352613 Chr.12: 121416515-121416515 on Build GRCh38
rs782306563 Chr.12: 121416189-121416189 on Build GRCh38
rs970724861 Chr.12: 121416569-121416569 on Build GRCh38
rs782662202 Chr.12: 121416159-121416159 on Build GRCh38
rs201536085 Chr.12: 121416124-121416124 on Build GRCh38
rs895299304 Chr.12: 121416534-121416534 on Build GRCh38
rs782138638 Chr.12: 121416204-121416204 on Build GRCh38
rs781856945 Chr.12: 121416348-121416348 on Build GRCh38
rs539486553 Chr.12: 121416053-121416053 on Build GRCh38
rs543550025 Chr.12: 121416560-121416560 on Build GRCh38
rs372006434 Chr.12: 121416420-121416420 on Build GRCh38
rs1032953964 Chr.12: 121416533-121416533 on Build GRCh38
rs61760895 Chr.12: 121416316-121416316 on Build GRCh38
rs782210356 Chr.12: 121416255-121416255 on Build GRCh38
rs781851364 Chr.12: 121416140-121416140 on Build GRCh38
rs201929209 Chr.12: 121416185-121416185 on Build GRCh38
rs782139970 Chr.12: 121416194-121416194 on Build GRCh38
rs782546188 Chr.12: 121416244-121416244 on Build GRCh38
rs1012376082 Chr.12: 121416543-121416543 on Build GRCh38
rs781939092 Chr.12: 121416317-121416317 on Build GRCh38

View Full Product Details