Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 76736028-76736567 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MFSD11
Entrez Gene ID
79157
Gene Name
major facilitator superfamily domain containing 11
Gene Aliases
ET
UniGene
Hs.732019
Gene Chromosome Location
Chr.17: 76736565-76803805 on Build GRCh38
Transcripts
NM_001242534.1 NR_030366.1 NM_001195427.1 NM_003016.4 NR_036608.1 XM_017024942.1

Gene Symbol
MIR636
Entrez Gene ID
693221
Gene Name
microRNA 636
Gene Aliases
MIRN636, hsa-mir-636
UniGene
Gene Chromosome Location
Chr.17: 76736450-76736548 on Build GRCh38
Transcripts
NM_001242534.1 NR_030366.1 NM_001195427.1 NM_003016.4 NR_036608.1 XM_017024942.1

Gene Symbol
SRSF2
Entrez Gene ID
6427
Gene Name
serine and arginine rich splicing factor 2
Gene Aliases
PR264, SC-35, SC35, SFRS2, SFRS2A, SRp30b
UniGene
Hs.584801
Gene Chromosome Location
Chr.17: 76734115-76737431 on Build GRCh38
Transcripts
NM_001242534.1 NR_030366.1 NM_001195427.1 NM_003016.4 NR_036608.1 XM_017024942.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3522599 c.445T>C p.S149P Chr.17: 76736382-76736382 on Build GRCh38
5762536 c.582A>C p.P194P Chr.17: 76736245-76736245 on Build GRCh38
6961596 c.445T>A p.S149T Chr.17: 76736322-76736322 on Build GRCh38
6961749 c.518C>T p.S173F Chr.17: 76736309-76736309 on Build GRCh38
1136005 c.408A>G p.R136R Chr.17: 76736359-76736359 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374580410 Chr.17: 76736428-76736428 on Build GRCh38
rs778034650 Chr.17: 76736434-76736434 on Build GRCh38
rs775685820 Chr.17: 76736493-76736493 on Build GRCh38
rs774227432 Chr.17: 76736332-76736332 on Build GRCh38
rs761557673 Chr.17: 76736389-76736389 on Build GRCh38
rs770775763 Chr.17: 76736247-76736247 on Build GRCh38
rs757717237 Chr.17: 76736475-76736475 on Build GRCh38
rs764750794 Chr.17: 76736502-76736502 on Build GRCh38
rs755530979 Chr.17: 76736290-76736290 on Build GRCh38
rs377063669 Chr.17: 76736487-76736487 on Build GRCh38
rs756714282 Chr.17: 76736501-76736501 on Build GRCh38
rs139439392 Chr.17: 76736203-76736203 on Build GRCh38
rs779043792 Chr.17: 76736457-76736457 on Build GRCh38
rs757305177 Chr.17: 76736544-76736544 on Build GRCh38
rs373001591 Chr.17: 76736289-76736289 on Build GRCh38
rs750310954 Chr.17: 76736519-76736532 on Build GRCh38
rs760875189 Chr.17: 76736351-76736374 on Build GRCh38
rs772618222 Chr.17: 76736318-76736347 on Build GRCh38
rs201537332 Chr.17: 76736469-76736469 on Build GRCh38
rs775473667 Chr.17: 76736356-76736356 on Build GRCh38
rs761166094 Chr.17: 76736538-76736538 on Build GRCh38
rs779523734 Chr.17: 76736308-76736313 on Build GRCh38
rs538819582 Chr.17: 76736057-76736057 on Build GRCh38
rs1054526065 Chr.17: 76736452-76736452 on Build GRCh38
rs1026735111 Chr.17: 76736507-76736507 on Build GRCh38
rs780698530 Chr.17: 76736108-76736108 on Build GRCh38
rs377064670 Chr.17: 76736542-76736542 on Build GRCh38
rs750602613 Chr.17: 76736545-76736545 on Build GRCh38
rs535366038 Chr.17: 76736082-76736082 on Build GRCh38
rs753218428 Chr.17: 76736489-76736489 on Build GRCh38
rs758866264 Chr.17: 76736174-76736174 on Build GRCh38
rs1040631368 Chr.17: 76736484-76736484 on Build GRCh38
rs781775847 Chr.17: 76736301-76736301 on Build GRCh38
rs267605064 Chr.17: 76736232-76736232 on Build GRCh38
rs770595099 Chr.17: 76736325-76736325 on Build GRCh38
rs951285534 Chr.17: 76736188-76736188 on Build GRCh38
rs766892786 Chr.17: 76736250-76736250 on Build GRCh38
rs147605645 Chr.17: 76736449-76736449 on Build GRCh38
rs779853165 Chr.17: 76736512-76736512 on Build GRCh38
rs949780859 Chr.17: 76736306-76736306 on Build GRCh38
rs746326579 Chr.17: 76736124-76736124 on Build GRCh38
rs761880835 Chr.17: 76736510-76736511 on Build GRCh38
rs1024057574 Chr.17: 76736348-76736348 on Build GRCh38
rs760218410 Chr.17: 76736264-76736264 on Build GRCh38
rs753364028 Chr.17: 76736372-76736372 on Build GRCh38
rs761911527 Chr.17: 76736318-76736318 on Build GRCh38
rs763609340 Chr.17: 76736369-76736369 on Build GRCh38
rs149695427 Chr.17: 76736253-76736253 on Build GRCh38
rs772436301 Chr.17: 76736125-76736125 on Build GRCh38
rs775978250 Chr.17: 76736536-76736536 on Build GRCh38
rs748814345 Chr.17: 76736548-76736548 on Build GRCh38
rs552779786 Chr.17: 76736062-76736062 on Build GRCh38
rs267605065 Chr.17: 76736248-76736248 on Build GRCh38
rs753547882 Chr.17: 76736266-76736266 on Build GRCh38
rs776106050 Chr.17: 76736130-76736130 on Build GRCh38
rs760165433 Chr.17: 76736361-76736361 on Build GRCh38
rs993549885 Chr.17: 76736506-76736506 on Build GRCh38
rs765186728 Chr.17: 76736517-76736535 on Build GRCh38
rs749426771 Chr.17: 76736437-76736437 on Build GRCh38
rs762727557 Chr.17: 76736525-76736525 on Build GRCh38
rs753312096 Chr.17: 76736419-76736419 on Build GRCh38
rs1041856447 Chr.17: 76736406-76736406 on Build GRCh38
rs775404737 Chr.17: 76736305-76736305 on Build GRCh38
rs769242167 Chr.17: 76736521-76736521 on Build GRCh38
rs554587663 Chr.17: 76736091-76736091 on Build GRCh38
rs369173477 Chr.17: 76736140-76736140 on Build GRCh38
rs748204143 Chr.17: 76736302-76736302 on Build GRCh38
rs199683955 Chr.17: 76736505-76736505 on Build GRCh38
rs971724555 Chr.17: 76736541-76736541 on Build GRCh38
rs1041498003 Chr.17: 76736081-76736081 on Build GRCh38
rs150516175 Chr.17: 76736160-76736160 on Build GRCh38
rs545669570 Chr.17: 76736478-76736478 on Build GRCh38
rs768558412 Chr.17: 76736515-76736515 on Build GRCh38
rs201197454 Chr.17: 76736386-76736386 on Build GRCh38
rs143205325 Chr.17: 76736137-76736137 on Build GRCh38
rs747562858 Chr.17: 76736185-76736185 on Build GRCh38
rs778124920 Chr.17: 76736320-76736320 on Build GRCh38
rs771569253 Chr.17: 76736220-76736225 on Build GRCh38
rs772860153 Chr.17: 76736524-76736524 on Build GRCh38
rs759553108 Chr.17: 76736249-76736249 on Build GRCh38
rs1007596585 Chr.17: 76736517-76736517 on Build GRCh38
rs772803565 Chr.17: 76736482-76736482 on Build GRCh38
rs764657547 Chr.17: 76736539-76736539 on Build GRCh38
rs776469568 Chr.17: 76736519-76736519 on Build GRCh38
rs745672312 Chr.17: 76736464-76736464 on Build GRCh38
rs766603819 Chr.17: 76736392-76736392 on Build GRCh38
rs755334722 Chr.17: 76736402-76736402 on Build GRCh38
rs776583852 Chr.17: 76736486-76736487 on Build GRCh38
rs770113956 Chr.17: 76736213-76736213 on Build GRCh38
rs754391427 Chr.17: 76736157-76736157 on Build GRCh38
rs747685241 Chr.17: 76736520-76736520 on Build GRCh38
rs764896469 Chr.17: 76736138-76736138 on Build GRCh38
rs755789538 Chr.17: 76736162-76736162 on Build GRCh38
rs148850121 Chr.17: 76736411-76736411 on Build GRCh38
rs758640296 Chr.17: 76736547-76736547 on Build GRCh38
rs566375077 Chr.17: 76736064-76736064 on Build GRCh38
rs368528056 Chr.17: 76736116-76736116 on Build GRCh38
rs768004549 Chr.17: 76736488-76736488 on Build GRCh38
rs541687424 Chr.17: 76736535-76736535 on Build GRCh38
rs780414329 Chr.17: 76736181-76736181 on Build GRCh38
rs746200268 Chr.17: 76736172-76736172 on Build GRCh38
rs1000409899 Chr.17: 76736096-76736096 on Build GRCh38
rs775895266 Chr.17: 76736319-76736324 on Build GRCh38
rs369705230 Chr.17: 76736347-76736347 on Build GRCh38
rs145534754 Chr.17: 76736270-76736270 on Build GRCh38
rs1018880469 Chr.17: 76736133-76736133 on Build GRCh38
rs759581207 Chr.17: 76736052-76736052 on Build GRCh38
rs746260518 Chr.17: 76736307-76736308 on Build GRCh38
rs776881024 Chr.17: 76736197-76736197 on Build GRCh38
rs1055432820 Chr.17: 76736049-76736049 on Build GRCh38
rs751926392 Chr.17: 76736396-76736396 on Build GRCh38
rs762648914 Chr.17: 76736236-76736236 on Build GRCh38
rs369604599 Chr.17: 76736510-76736510 on Build GRCh38
rs751578830 Chr.17: 76736148-76736148 on Build GRCh38
rs937212716 Chr.17: 76736269-76736269 on Build GRCh38
rs773606366 Chr.17: 76736216-76736216 on Build GRCh38
rs976353866 Chr.17: 76736494-76736494 on Build GRCh38
rs944552315 Chr.17: 76736143-76736143 on Build GRCh38
rs768551262 Chr.17: 76736191-76736191 on Build GRCh38
rs559151830 Chr.17: 76736513-76736513 on Build GRCh38
rs768906412 Chr.17: 76736445-76736446 on Build GRCh38
rs747814367 Chr.17: 76736111-76736111 on Build GRCh38
rs189841356 Chr.17: 76736167-76736167 on Build GRCh38
rs376398460 Chr.17: 76736323-76736323 on Build GRCh38
rs11550962 Chr.17: 76736388-76736388 on Build GRCh38
rs762854633 Chr.17: 76736542-76736544 on Build GRCh38
rs983359881 Chr.17: 76736480-76736480 on Build GRCh38
rs1033143994 Chr.17: 76736102-76736102 on Build GRCh38
rs768114132 Chr.17: 76736477-76736477 on Build GRCh38
rs374140173 Chr.17: 76736136-76736136 on Build GRCh38
rs757347241 Chr.17: 76736509-76736509 on Build GRCh38
rs780470069 Chr.17: 76736288-76736288 on Build GRCh38
rs114769537 Chr.17: 76736123-76736123 on Build GRCh38
rs372808564 Chr.17: 76736516-76736516 on Build GRCh38
rs237058 Chr.17: 76736425-76736425 on Build GRCh38

View Full Product Details