Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 70175310-70175909 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNJ2
Entrez Gene ID
3759
Gene Name
potassium voltage-gated channel subfamily J member 2
Gene Aliases
ATFB9, HHBIRK1, HHIRK1, IRK1, KIR2.1, LQT7, SQT3
UniGene
Hs.1547
Gene Chromosome Location
Chr.17: 70169535-70180044 on Build GRCh38
Transcripts
NM_000891.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6147431 c.540G>T p.M180I Chr.17: 70175579-70175579 on Build GRCh38
6691157 c.451A>G p.T151A Chr.17: 70175490-70175490 on Build GRCh38
707430 c.718G>A p.G240R Chr.17: 70175757-70175757 on Build GRCh38
5499316 c.276C>T p.F92F Chr.17: 70175315-70175315 on Build GRCh38
983659 c.617G>T p.G206V Chr.17: 70175656-70175656 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs786205815 Chr.17: 70175367-70175367 on Build GRCh38
rs199473375 Chr.17: 70175407-70175407 on Build GRCh38
rs150326473 Chr.17: 70175550-70175550 on Build GRCh38
rs772189737 Chr.17: 70175876-70175876 on Build GRCh38
rs754883107 Chr.17: 70175765-70175765 on Build GRCh38
rs775567368 Chr.17: 70175450-70175450 on Build GRCh38
rs199473657 Chr.17: 70175718-70175718 on Build GRCh38
rs956024087 Chr.17: 70175578-70175578 on Build GRCh38
rs751862699 Chr.17: 70175492-70175492 on Build GRCh38
rs538725136 Chr.17: 70175393-70175393 on Build GRCh38
rs759793133 Chr.17: 70175643-70175643 on Build GRCh38
rs1049263340 Chr.17: 70175849-70175849 on Build GRCh38
rs765303864 Chr.17: 70175339-70175339 on Build GRCh38
rs199473378 Chr.17: 70175469-70175469 on Build GRCh38
rs199473381 Chr.17: 70175605-70175605 on Build GRCh38
rs779856198 Chr.17: 70175733-70175733 on Build GRCh38
rs371331394 Chr.17: 70175361-70175361 on Build GRCh38
rs573888708 Chr.17: 70175552-70175552 on Build GRCh38
rs761179414 Chr.17: 70175660-70175660 on Build GRCh38
rs767844416 Chr.17: 70175654-70175654 on Build GRCh38
rs199473383 Chr.17: 70175683-70175683 on Build GRCh38
rs778699148 Chr.17: 70175556-70175556 on Build GRCh38
rs751948464 Chr.17: 70175352-70175352 on Build GRCh38
rs199473374 Chr.17: 70175340-70175340 on Build GRCh38
rs1037493754 Chr.17: 70175814-70175814 on Build GRCh38
rs141035459 Chr.17: 70175655-70175655 on Build GRCh38
rs151168735 Chr.17: 70175428-70175428 on Build GRCh38
rs147561965 Chr.17: 70175337-70175343 on Build GRCh38
rs1022647325 Chr.17: 70175506-70175506 on Build GRCh38
rs375646186 Chr.17: 70175398-70175398 on Build GRCh38
rs183993685 Chr.17: 70175609-70175609 on Build GRCh38
rs142049682 Chr.17: 70175537-70175537 on Build GRCh38
rs199473656 Chr.17: 70175689-70175689 on Build GRCh38
rs748924113 Chr.17: 70175405-70175405 on Build GRCh38
rs878897599 Chr.17: 70175351-70175351 on Build GRCh38
rs201850260 Chr.17: 70175714-70175714 on Build GRCh38
rs886042211 Chr.17: 70175845-70175845 on Build GRCh38
rs104894583 Chr.17: 70175685-70175685 on Build GRCh38
rs376921497 Chr.17: 70175342-70175342 on Build GRCh38
rs779764704 Chr.17: 70175574-70175574 on Build GRCh38
rs140274795 Chr.17: 70175366-70175366 on Build GRCh38
rs758092571 Chr.17: 70175884-70175884 on Build GRCh38
rs758144475 Chr.17: 70175723-70175723 on Build GRCh38
rs753200997 Chr.17: 70175378-70175378 on Build GRCh38
rs104894584 Chr.17: 70175553-70175553 on Build GRCh38
rs786205821 Chr.17: 70175435-70175436 on Build GRCh38
rs199473384 Chr.17: 70175692-70175692 on Build GRCh38
rs786205816 Chr.17: 70175860-70175860 on Build GRCh38
rs745672560 Chr.17: 70175429-70175429 on Build GRCh38
rs771056622 Chr.17: 70175636-70175636 on Build GRCh38
rs199473382 Chr.17: 70175613-70175613 on Build GRCh38
rs527277177 Chr.17: 70175881-70175881 on Build GRCh38
rs199473385 Chr.17: 70175818-70175818 on Build GRCh38
rs750630010 Chr.17: 70175341-70175341 on Build GRCh38
rs764523123 Chr.17: 70175676-70175676 on Build GRCh38
rs767934529 Chr.17: 70175505-70175505 on Build GRCh38
rs140147979 Chr.17: 70175538-70175538 on Build GRCh38
rs144983213 Chr.17: 70175624-70175624 on Build GRCh38
rs780976921 Chr.17: 70175768-70175768 on Build GRCh38
rs943311933 Chr.17: 70175756-70175756 on Build GRCh38
rs930806279 Chr.17: 70175844-70175844 on Build GRCh38
rs748150144 Chr.17: 70175775-70175775 on Build GRCh38
rs886039094 Chr.17: 70175789-70175789 on Build GRCh38
rs770964805 Chr.17: 70175783-70175783 on Build GRCh38
rs7221086 Chr.17: 70175699-70175699 on Build GRCh38
rs104894578 Chr.17: 70175691-70175691 on Build GRCh38
rs199473379 Chr.17: 70175476-70175476 on Build GRCh38
rs766588803 Chr.17: 70175491-70175491 on Build GRCh38
rs199473655 Chr.17: 70175614-70175614 on Build GRCh38
rs104894581 Chr.17: 70175596-70175596 on Build GRCh38
rs199473380 Chr.17: 70175500-70175500 on Build GRCh38
rs766678770 Chr.17: 70175343-70175343 on Build GRCh38
rs1005537943 Chr.17: 70175432-70175432 on Build GRCh38
rs746781024 Chr.17: 70175749-70175749 on Build GRCh38
rs978533069 Chr.17: 70175569-70175569 on Build GRCh38
rs772055408 Chr.17: 70175449-70175449 on Build GRCh38
rs989113819 Chr.17: 70175641-70175641 on Build GRCh38
rs773272852 Chr.17: 70175618-70175618 on Build GRCh38
rs900684102 Chr.17: 70175363-70175363 on Build GRCh38
rs781674038 Chr.17: 70175368-70175368 on Build GRCh38
rs761209037 Chr.17: 70175562-70175562 on Build GRCh38
rs370571312 Chr.17: 70175656-70175656 on Build GRCh38
rs771965504 Chr.17: 70175571-70175571 on Build GRCh38
rs756826775 Chr.17: 70175703-70175703 on Build GRCh38
rs759879267 Chr.17: 70175504-70175504 on Build GRCh38
rs774551339 Chr.17: 70175639-70175639 on Build GRCh38
rs773185250 Chr.17: 70175474-70175474 on Build GRCh38
rs763190548 Chr.17: 70175483-70175483 on Build GRCh38
rs753215455 Chr.17: 70175516-70175516 on Build GRCh38
rs79650811 Chr.17: 70175331-70175331 on Build GRCh38
rs746913022 Chr.17: 70175597-70175597 on Build GRCh38
rs864622292 Chr.17: 70175446-70175448 on Build GRCh38
rs138877244 Chr.17: 70175411-70175411 on Build GRCh38
rs746052198 Chr.17: 70175838-70175838 on Build GRCh38
rs544384907 Chr.17: 70175570-70175570 on Build GRCh38
rs199473376 Chr.17: 70175446-70175446 on Build GRCh38
rs572394146 Chr.17: 70175456-70175456 on Build GRCh38
rs559935671 Chr.17: 70175717-70175717 on Build GRCh38
rs199473654 Chr.17: 70175475-70175475 on Build GRCh38
rs886053323 Chr.17: 70175383-70175383 on Build GRCh38
rs587781007 Chr.17: 70175812-70175812 on Build GRCh38
rs774461588 Chr.17: 70175820-70175820 on Build GRCh38
rs199473377 Chr.17: 70175470-70175470 on Build GRCh38
rs138778849 Chr.17: 70175804-70175804 on Build GRCh38

View Full Product Details