Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 70324547-70325119 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SOCS6
Entrez Gene ID
9306
Gene Name
suppressor of cytokine signaling 6
Gene Aliases
CIS-4, CIS4, HSPC060, SOCS-4, SOCS-6, SOCS4, SSI4, STAI4, STATI4
UniGene
Hs.44439
Gene Chromosome Location
Chr.18: 70288901-70330199 on Build GRCh38
Transcripts
NM_004232.3 XM_005266783.3 XM_017026086.1 XM_017026087.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
247513 c.51A>T p.K17N Chr.18: 70324719-70324719 on Build GRCh38
5543464 c.296C>T p.A99V Chr.18: 70324964-70324964 on Build GRCh38
6786731 c.343G>T p.D115Y Chr.18: 70325011-70325011 on Build GRCh38
3145519 c.181G>C p.E61Q Chr.18: 70324849-70324849 on Build GRCh38
4140150 c.126A>G p.L42L Chr.18: 70324794-70324794 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372727194 Chr.18: 70324761-70324761 on Build GRCh38
rs771331914 Chr.18: 70324776-70324776 on Build GRCh38
rs766239751 Chr.18: 70324656-70324656 on Build GRCh38
rs143010911 Chr.18: 70324974-70324974 on Build GRCh38
rs760986400 Chr.18: 70324649-70324649 on Build GRCh38
rs776531677 Chr.18: 70324785-70324785 on Build GRCh38
rs1033022600 Chr.18: 70324664-70324664 on Build GRCh38
rs751994123 Chr.18: 70325068-70325068 on Build GRCh38
rs755075112 Chr.18: 70324980-70324982 on Build GRCh38
rs1016461867 Chr.18: 70324570-70324570 on Build GRCh38
rs754219214 Chr.18: 70324828-70324828 on Build GRCh38
rs142610203 Chr.18: 70324720-70324720 on Build GRCh38
rs41328646 Chr.18: 70324857-70324857 on Build GRCh38
rs377177411 Chr.18: 70325076-70325076 on Build GRCh38
rs1047654 Chr.18: 70325031-70325031 on Build GRCh38
rs141810837 Chr.18: 70325013-70325013 on Build GRCh38
rs373053671 Chr.18: 70324641-70324641 on Build GRCh38
rs371416171 Chr.18: 70324891-70324891 on Build GRCh38
rs903336762 Chr.18: 70324597-70324597 on Build GRCh38
rs751091357 Chr.18: 70324820-70324820 on Build GRCh38
rs749147871 Chr.18: 70324951-70324951 on Build GRCh38
rs759264328 Chr.18: 70325064-70325064 on Build GRCh38
rs146906823 Chr.18: 70324831-70324831 on Build GRCh38
rs774460366 Chr.18: 70324631-70324631 on Build GRCh38
rs774218647 Chr.18: 70324965-70324965 on Build GRCh38
rs149490841 Chr.18: 70324945-70324945 on Build GRCh38
rs758946394 Chr.18: 70324914-70324914 on Build GRCh38
rs772570614 Chr.18: 70325047-70325047 on Build GRCh38
rs761371103 Chr.18: 70324884-70324884 on Build GRCh38
rs764564730 Chr.18: 70325003-70325003 on Build GRCh38
rs776285560 Chr.18: 70325059-70325059 on Build GRCh38
rs115351133 Chr.18: 70324989-70324989 on Build GRCh38
rs137973287 Chr.18: 70324879-70324879 on Build GRCh38
rs34330670 Chr.18: 70324868-70324869 on Build GRCh38
rs781656056 Chr.18: 70324598-70324599 on Build GRCh38
rs370251524 Chr.18: 70324862-70324862 on Build GRCh38
rs963862835 Chr.18: 70324591-70324591 on Build GRCh38
rs377209302 Chr.18: 70324817-70324817 on Build GRCh38
rs764067152 Chr.18: 70324881-70324881 on Build GRCh38
rs777982483 Chr.18: 70324844-70324844 on Build GRCh38
rs201422335 Chr.18: 70325070-70325070 on Build GRCh38
rs557673824 Chr.18: 70324977-70324977 on Build GRCh38
rs1042260516 Chr.18: 70324582-70324582 on Build GRCh38
rs769407600 Chr.18: 70324878-70324878 on Build GRCh38
rs147815869 Chr.18: 70324832-70324832 on Build GRCh38
rs753158403 Chr.18: 70324911-70324911 on Build GRCh38
rs754020528 Chr.18: 70325073-70325073 on Build GRCh38
rs757424437 Chr.18: 70324786-70324786 on Build GRCh38
rs751146299 Chr.18: 70324696-70324696 on Build GRCh38
rs751671936 Chr.18: 70324936-70324936 on Build GRCh38
rs749526105 Chr.18: 70325036-70325036 on Build GRCh38
rs750564600 Chr.18: 70325015-70325015 on Build GRCh38
rs371221269 Chr.18: 70324692-70324692 on Build GRCh38
rs762386375 Chr.18: 70325069-70325069 on Build GRCh38
rs749694019 Chr.18: 70324715-70324715 on Build GRCh38
rs543825635 Chr.18: 70324968-70324968 on Build GRCh38
rs535454922 Chr.18: 70324684-70324684 on Build GRCh38
rs780401339 Chr.18: 70324626-70324626 on Build GRCh38
rs763465078 Chr.18: 70324994-70324994 on Build GRCh38
rs537749233 Chr.18: 70324946-70324946 on Build GRCh38
rs748033311 Chr.18: 70325046-70325046 on Build GRCh38
rs2231562 Chr.18: 70325043-70325043 on Build GRCh38
rs760226147 Chr.18: 70324979-70324979 on Build GRCh38
rs756872913 Chr.18: 70324822-70324822 on Build GRCh38
rs756932164 Chr.18: 70324705-70324705 on Build GRCh38
rs150466761 Chr.18: 70324867-70324867 on Build GRCh38
rs971847705 Chr.18: 70324596-70324596 on Build GRCh38
rs781081081 Chr.18: 70325014-70325014 on Build GRCh38
rs917532892 Chr.18: 70324895-70324895 on Build GRCh38
rs766362457 Chr.18: 70325072-70325072 on Build GRCh38
rs758267776 Chr.18: 70325086-70325086 on Build GRCh38
rs764511524 Chr.18: 70325065-70325065 on Build GRCh38
rs767224648 Chr.18: 70324823-70324823 on Build GRCh38
rs756257758 Chr.18: 70325028-70325028 on Build GRCh38
rs765204358 Chr.18: 70324801-70324801 on Build GRCh38
rs769093084 Chr.18: 70324630-70324630 on Build GRCh38
rs756162788 Chr.18: 70325071-70325071 on Build GRCh38
rs370650342 Chr.18: 70324628-70324628 on Build GRCh38
rs374238156 Chr.18: 70324896-70324896 on Build GRCh38
rs773459364 Chr.18: 70324737-70324737 on Build GRCh38
rs573400452 Chr.18: 70324868-70324868 on Build GRCh38
rs550577601 Chr.18: 70324811-70324811 on Build GRCh38
rs752043193 Chr.18: 70325011-70325011 on Build GRCh38
rs371708469 Chr.18: 70325051-70325051 on Build GRCh38
rs987384515 Chr.18: 70324824-70324824 on Build GRCh38
rs778658948 Chr.18: 70325075-70325075 on Build GRCh38
rs752996222 Chr.18: 70324991-70324991 on Build GRCh38
rs773514303 Chr.18: 70324646-70324646 on Build GRCh38
rs201670304 Chr.18: 70324940-70324940 on Build GRCh38
rs768568432 Chr.18: 70324952-70324952 on Build GRCh38
rs758971852 Chr.18: 70324840-70324840 on Build GRCh38
rs770472798 Chr.18: 70325054-70325054 on Build GRCh38
rs1026061801 Chr.18: 70324600-70324600 on Build GRCh38
rs753892759 Chr.18: 70324724-70324726 on Build GRCh38
rs777820754 Chr.18: 70325087-70325087 on Build GRCh38
rs188719904 Chr.18: 70324934-70324934 on Build GRCh38
rs1030056671 Chr.18: 70325029-70325029 on Build GRCh38
rs775229175 Chr.18: 70324804-70324804 on Build GRCh38
rs777278365 Chr.18: 70324695-70324695 on Build GRCh38
rs771774974 Chr.18: 70324635-70324635 on Build GRCh38
rs753711665 Chr.18: 70324680-70324680 on Build GRCh38
rs749636087 Chr.18: 70325032-70325032 on Build GRCh38
rs779691105 Chr.18: 70324947-70324947 on Build GRCh38
rs868241462 Chr.18: 70324843-70324843 on Build GRCh38
rs202207403 Chr.18: 70324683-70324683 on Build GRCh38
rs748565655 Chr.18: 70324839-70324839 on Build GRCh38
rs755240902 Chr.18: 70324830-70324830 on Build GRCh38
rs768375834 Chr.18: 70325041-70325041 on Build GRCh38
rs781334187 Chr.18: 70324941-70324941 on Build GRCh38
rs374960583 Chr.18: 70324870-70324870 on Build GRCh38
rs771981263 Chr.18: 70324734-70324734 on Build GRCh38
rs368532175 Chr.18: 70324883-70324883 on Build GRCh38
rs780346198 Chr.18: 70324709-70324709 on Build GRCh38
rs748703534 Chr.18: 70324726-70324726 on Build GRCh38

View Full Product Details