Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 15321192-15321791 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PIGA
Entrez Gene ID
5277
Gene Name
phosphatidylinositol glycan anchor biosynthesis class A
Gene Aliases
GPI3, MCAHS2, PIG-A, PNH1
UniGene
Hs.137154
Gene Chromosome Location
Chr.X: 15319451-15335554 on Build GRCh38
Transcripts
NM_002641.3 NM_020473.3 NR_033835.1 NR_033836.1 XM_011545539.2 XM_017029581.1

SNP Detail

SNP ID Chromosome Location
rs756816136 Chr.X: 15321494-15321494 on Build GRCh38
rs759697480 Chr.X: 15321760-15321760 on Build GRCh38
rs761613346 Chr.X: 15321558-15321558 on Build GRCh38
rs748161988 Chr.X: 15321742-15321742 on Build GRCh38
rs775805589 Chr.X: 15321770-15321770 on Build GRCh38
rs771058274 Chr.X: 15321700-15321700 on Build GRCh38
rs201783559 Chr.X: 15321595-15321595 on Build GRCh38
rs201361742 Chr.X: 15321747-15321747 on Build GRCh38
rs780815350 Chr.X: 15321734-15321734 on Build GRCh38
rs745560847 Chr.X: 15321514-15321514 on Build GRCh38
rs373138445 Chr.X: 15321463-15321463 on Build GRCh38
rs794727790 Chr.X: 15321759-15321759 on Build GRCh38
rs1024326491 Chr.X: 15321393-15321393 on Build GRCh38
rs745960092 Chr.X: 15321265-15321265 on Build GRCh38
rs202201866 Chr.X: 15321421-15321421 on Build GRCh38
rs778444082 Chr.X: 15321653-15321653 on Build GRCh38
rs139149663 Chr.X: 15321664-15321664 on Build GRCh38
rs786200912 Chr.X: 15321605-15321606 on Build GRCh38
rs958525351 Chr.X: 15321370-15321370 on Build GRCh38
rs774644659 Chr.X: 15321756-15321756 on Build GRCh38
rs184099983 Chr.X: 15321451-15321451 on Build GRCh38
rs746591138 Chr.X: 15321706-15321706 on Build GRCh38
rs192475811 Chr.X: 15321757-15321757 on Build GRCh38
rs773189052 Chr.X: 15321543-15321543 on Build GRCh38
rs914087630 Chr.X: 15321738-15321738 on Build GRCh38
rs1034721128 Chr.X: 15321363-15321363 on Build GRCh38
rs770570366 Chr.X: 15321474-15321474 on Build GRCh38
rs753691551 Chr.X: 15321218-15321220 on Build GRCh38
rs587776728 Chr.X: 15321637-15321638 on Build GRCh38
rs1010005996 Chr.X: 15321230-15321230 on Build GRCh38
rs761116405 Chr.X: 15321593-15321593 on Build GRCh38
rs758351965 Chr.X: 15321448-15321448 on Build GRCh38
rs749702985 Chr.X: 15321491-15321491 on Build GRCh38
rs142772459 Chr.X: 15321540-15321540 on Build GRCh38
rs775330646 Chr.X: 15321534-15321534 on Build GRCh38
rs962072746 Chr.X: 15321435-15321435 on Build GRCh38
rs920567695 Chr.X: 15321449-15321449 on Build GRCh38
rs774546648 Chr.X: 15321574-15321574 on Build GRCh38
rs61760986 Chr.X: 15321541-15321541 on Build GRCh38
rs766602742 Chr.X: 15321570-15321570 on Build GRCh38
rs1014071560 Chr.X: 15321379-15321380 on Build GRCh38
rs754401169 Chr.X: 15321624-15321624 on Build GRCh38
rs759900292 Chr.X: 15321575-15321575 on Build GRCh38
rs192931358 Chr.X: 15321425-15321425 on Build GRCh38
rs749080220 Chr.X: 15321642-15321642 on Build GRCh38
rs142178534 Chr.X: 15321499-15321499 on Build GRCh38
rs148660178 Chr.X: 15321466-15321466 on Build GRCh38
rs772225061 Chr.X: 15321763-15321763 on Build GRCh38
rs369011467 Chr.X: 15321472-15321472 on Build GRCh38
rs769061128 Chr.X: 15321579-15321579 on Build GRCh38
rs387906726 Chr.X: 15321727-15321727 on Build GRCh38
rs773967133 Chr.X: 15321488-15321489 on Build GRCh38
rs754782099 Chr.X: 15321313-15321313 on Build GRCh38
rs768524297 Chr.X: 15321721-15321721 on Build GRCh38
rs904197035 Chr.X: 15321304-15321304 on Build GRCh38
rs752798208 Chr.X: 15321580-15321580 on Build GRCh38
rs771690457 Chr.X: 15321527-15321527 on Build GRCh38
rs757835670 Chr.X: 15321682-15321682 on Build GRCh38
rs149950708 Chr.X: 15321764-15321764 on Build GRCh38
rs776651457 Chr.X: 15321542-15321542 on Build GRCh38
rs35527052 Chr.X: 15321332-15321333 on Build GRCh38

View Full Product Details