Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 154765311-154765830 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DKC1
Entrez Gene ID
1736
Gene Name
dyskerin pseudouridine synthase 1
Gene Aliases
CBF5, DKC, DKCX, NAP57, NOLA4, XAP101
UniGene
Hs.4747
Gene Chromosome Location
Chr.X: 154762742-154777689 on Build GRCh38
Transcripts
NM_001142463.2 NM_001288747.1 NM_001363.4 NR_110021.1 NR_110022.1 NR_110023.1

SNP Detail

SNP ID Chromosome Location
rs782309492 Chr.X: 154765402-154765402 on Build GRCh38
rs782397171 Chr.X: 154765805-154765805 on Build GRCh38
rs781910103 Chr.X: 154765339-154765339 on Build GRCh38
rs782504034 Chr.X: 154765506-154765506 on Build GRCh38
rs961019560 Chr.X: 154765625-154765625 on Build GRCh38
rs28936072 Chr.X: 154765472-154765472 on Build GRCh38
rs121912292 Chr.X: 154765478-154765478 on Build GRCh38
rs199957716 Chr.X: 154765534-154765534 on Build GRCh38
rs782269755 Chr.X: 154765409-154765409 on Build GRCh38
rs973758048 Chr.X: 154765722-154765722 on Build GRCh38
rs137854491 Chr.X: 154765450-154765450 on Build GRCh38
rs782433339 Chr.X: 154765541-154765541 on Build GRCh38
rs121912302 Chr.X: 154765480-154765480 on Build GRCh38
rs1015553644 Chr.X: 154765590-154765590 on Build GRCh38
rs781897289 Chr.X: 154765792-154765792 on Build GRCh38
rs782381308 Chr.X: 154765439-154765439 on Build GRCh38
rs121912293 Chr.X: 154765465-154765465 on Build GRCh38
rs947370657 Chr.X: 154765789-154765789 on Build GRCh38
rs912732713 Chr.X: 154765737-154765737 on Build GRCh38
rs782572592 Chr.X: 154765741-154765741 on Build GRCh38
rs927337545 Chr.X: 154765799-154765799 on Build GRCh38
rs782750352 Chr.X: 154765725-154765725 on Build GRCh38
rs782382476 Chr.X: 154765398-154765398 on Build GRCh38
rs121912296 Chr.X: 154765474-154765474 on Build GRCh38
rs782073684 Chr.X: 154765773-154765773 on Build GRCh38
rs782449722 Chr.X: 154765729-154765729 on Build GRCh38
rs782094431 Chr.X: 154765452-154765452 on Build GRCh38
rs370398142 Chr.X: 154765574-154765574 on Build GRCh38
rs186389886 Chr.X: 154765611-154765611 on Build GRCh38
rs1033528226 Chr.X: 154765546-154765546 on Build GRCh38
rs369777662 Chr.X: 154765497-154765497 on Build GRCh38
rs782537853 Chr.X: 154765780-154765780 on Build GRCh38
rs782755815 Chr.X: 154765576-154765576 on Build GRCh38
rs137854489 Chr.X: 154765468-154765470 on Build GRCh38
rs199422243 Chr.X: 154765486-154765486 on Build GRCh38
rs376044835 Chr.X: 154765573-154765573 on Build GRCh38
rs781888016 Chr.X: 154765547-154765547 on Build GRCh38
rs781941481 Chr.X: 154765487-154765487 on Build GRCh38
rs782074427 Chr.X: 154765730-154765730 on Build GRCh38
rs137854490 Chr.X: 154765468-154765469 on Build GRCh38
rs121912287 Chr.X: 154765525-154765526 on Build GRCh38
rs782251510 Chr.X: 154765648-154765648 on Build GRCh38
rs372511229 Chr.X: 154765456-154765456 on Build GRCh38
rs121912304 Chr.X: 154765505-154765505 on Build GRCh38
rs781839981 Chr.X: 154765686-154765687 on Build GRCh38
rs782028472 Chr.X: 154765403-154765403 on Build GRCh38

View Full Product Details