Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 139749827-139750302 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP11C
Entrez Gene ID
286410
Gene Name
ATPase phospholipid transporting 11C
Gene Aliases
ATPIG, ATPIQ
UniGene
Hs.88252
Gene Chromosome Location
Chr.X: 139726346-139933083 on Build GRCh38
Transcripts
NM_001010986.2 NM_173694.4 XM_017029439.1 XM_017029440.1 XM_017029441.1 XM_017029442.1 XM_017029443.1 XM_017029444.1 XM_017029445.1 XM_017029446.1 XM_017029447.1 XM_017029448.1 XM_017029449.1

SNP Detail

SNP ID Chromosome Location
rs759858539 Chr.X: 139750005-139750005 on Build GRCh38
rs772911480 Chr.X: 139750015-139750015 on Build GRCh38
rs188547981 Chr.X: 139750178-139750178 on Build GRCh38
rs778958285 Chr.X: 139750199-139750199 on Build GRCh38
rs965714834 Chr.X: 139750147-139750147 on Build GRCh38
rs138867304 Chr.X: 139750093-139750093 on Build GRCh38
rs903960240 Chr.X: 139750001-139750001 on Build GRCh38
rs41300305 Chr.X: 139749985-139749985 on Build GRCh38
rs938615516 Chr.X: 139750152-139750152 on Build GRCh38
rs764717586 Chr.X: 139750050-139750050 on Build GRCh38
rs181896961 Chr.X: 139749863-139749863 on Build GRCh38
rs951914054 Chr.X: 139750269-139750269 on Build GRCh38
rs1010102802 Chr.X: 139750262-139750262 on Build GRCh38
rs184574634 Chr.X: 139749933-139749933 on Build GRCh38
rs768467182 Chr.X: 139750191-139750191 on Build GRCh38
rs984018748 Chr.X: 139749925-139749925 on Build GRCh38
rs760188614 Chr.X: 139750065-139750065 on Build GRCh38
rs200565213 Chr.X: 139750096-139750096 on Build GRCh38
rs776882132 Chr.X: 139749997-139749997 on Build GRCh38
rs201400223 Chr.X: 139750034-139750034 on Build GRCh38
rs760557489 Chr.X: 139750174-139750177 on Build GRCh38
rs891629656 Chr.X: 139750182-139750182 on Build GRCh38
rs917926197 Chr.X: 139749926-139749926 on Build GRCh38
rs767272667 Chr.X: 139750008-139750009 on Build GRCh38
rs769446927 Chr.X: 139749978-139749978 on Build GRCh38
rs1008485451 Chr.X: 139750046-139750046 on Build GRCh38
rs755878823 Chr.X: 139750155-139750155 on Build GRCh38
rs765545831 Chr.X: 139750011-139750011 on Build GRCh38
rs751086961 Chr.X: 139750063-139750063 on Build GRCh38
rs779715769 Chr.X: 139750176-139750176 on Build GRCh38
rs1026042900 Chr.X: 139750244-139750244 on Build GRCh38
rs764227309 Chr.X: 139750254-139750254 on Build GRCh38
rs767808205 Chr.X: 139750035-139750035 on Build GRCh38
rs1049956292 Chr.X: 139750007-139750007 on Build GRCh38
rs750121703 Chr.X: 139750151-139750151 on Build GRCh38
rs890040757 Chr.X: 139750045-139750045 on Build GRCh38
rs766351273 Chr.X: 139750192-139750193 on Build GRCh38

View Full Product Details