Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 1211595-1212137 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTBP1
Entrez Gene ID
1487
Gene Name
C-terminal binding protein 1
Gene Aliases
BARS
UniGene
Hs.208597
Gene Chromosome Location
Chr.4: 1211440-1250329 on Build GRCh38
Transcripts
NM_001012614.1 NM_001328.2 XM_005272261.3 XM_005272263.4 XM_011513395.2 XM_011513396.2 XM_011513397.2 XM_017007762.1 XM_017007763.1 XM_017007764.1 XM_017007765.1 XM_017007766.1 XM_017007767.1 NR_104331.1

Gene Symbol
CTBP1-AS
Entrez Gene ID
285463
Gene Name
CTBP1 antisense RNA
Gene Aliases
PCAT10
UniGene
Hs.707614
Gene Chromosome Location
Chr.4: 1210120-1218591 on Build GRCh38
Transcripts
NM_001012614.1 NM_001328.2 XM_005272261.3 XM_005272263.4 XM_011513395.2 XM_011513396.2 XM_011513397.2 XM_017007762.1 XM_017007763.1 XM_017007764.1 XM_017007765.1 XM_017007766.1 XM_017007767.1 NR_104331.1

SNP Detail

SNP ID Chromosome Location
rs563184205 Chr.4: 1211687-1211687 on Build GRCh38
rs1005544211 Chr.4: 1212059-1212059 on Build GRCh38
rs569490067 Chr.4: 1211857-1211857 on Build GRCh38
rs909805634 Chr.4: 1211880-1211880 on Build GRCh38
rs964595256 Chr.4: 1211802-1211802 on Build GRCh38
rs955726735 Chr.4: 1211998-1211998 on Build GRCh38
rs993584251 Chr.4: 1211660-1211660 on Build GRCh38
rs140581613 Chr.4: 1211804-1211804 on Build GRCh38
rs947087201 Chr.4: 1212111-1212111 on Build GRCh38
rs372329092 Chr.4: 1211667-1211667 on Build GRCh38
rs1045480 Chr.4: 1211879-1211879 on Build GRCh38
rs895921251 Chr.4: 1212037-1212037 on Build GRCh38
rs376995762 Chr.4: 1211899-1211899 on Build GRCh38
rs4974572 Chr.4: 1211949-1211949 on Build GRCh38
rs555048849 Chr.4: 1211871-1211871 on Build GRCh38
rs989742813 Chr.4: 1212001-1212001 on Build GRCh38
rs186984898 Chr.4: 1211862-1211862 on Build GRCh38
rs890972667 Chr.4: 1211875-1211875 on Build GRCh38
rs978415667 Chr.4: 1212112-1212112 on Build GRCh38
rs190712719 Chr.4: 1211891-1211891 on Build GRCh38
rs988540959 Chr.4: 1212084-1212084 on Build GRCh38
rs181483701 Chr.4: 1212039-1212039 on Build GRCh38
rs962366565 Chr.4: 1211631-1211631 on Build GRCh38
rs964290027 Chr.4: 1212065-1212065 on Build GRCh38
rs1036946943 Chr.4: 1212077-1212077 on Build GRCh38
rs566302187 Chr.4: 1211670-1211670 on Build GRCh38
rs564855534 Chr.4: 1211684-1211685 on Build GRCh38
rs1027731924 Chr.4: 1211898-1211899 on Build GRCh38
rs905770395 Chr.4: 1211980-1211980 on Build GRCh38
rs912930939 Chr.4: 1211920-1211920 on Build GRCh38
rs1043403735 Chr.4: 1211965-1211965 on Build GRCh38
rs1018951262 Chr.4: 1212064-1212064 on Build GRCh38
rs765714654 Chr.4: 1212073-1212073 on Build GRCh38
rs956858198 Chr.4: 1212083-1212083 on Build GRCh38
rs947033471 Chr.4: 1211929-1211929 on Build GRCh38
rs374555956 Chr.4: 1211881-1211881 on Build GRCh38
rs934348061 Chr.4: 1211725-1211725 on Build GRCh38
rs11540633 Chr.4: 1211795-1211795 on Build GRCh38
rs911475152 Chr.4: 1211801-1211801 on Build GRCh38
rs765360395 Chr.4: 1212016-1212018 on Build GRCh38
rs1045470 Chr.4: 1212104-1212104 on Build GRCh38
rs1032361833 Chr.4: 1211987-1211987 on Build GRCh38
rs984396054 Chr.4: 1211718-1211718 on Build GRCh38
rs572008054 Chr.4: 1211781-1211781 on Build GRCh38
rs1023113942 Chr.4: 1211962-1211962 on Build GRCh38
rs943940976 Chr.4: 1211876-1211876 on Build GRCh38
rs1056926063 Chr.4: 1211993-1211993 on Build GRCh38
rs932413967 Chr.4: 1211846-1211846 on Build GRCh38
rs544119555 Chr.4: 1211995-1211995 on Build GRCh38
rs937690023 Chr.4: 1211865-1211865 on Build GRCh38
rs1025716676 Chr.4: 1212046-1212046 on Build GRCh38
rs76532252 Chr.4: 1211791-1211791 on Build GRCh38
rs1013008678 Chr.4: 1211913-1211913 on Build GRCh38
rs887828831 Chr.4: 1211906-1211906 on Build GRCh38
rs924270159 Chr.4: 1211721-1211721 on Build GRCh38
rs1049051514 Chr.4: 1212098-1212098 on Build GRCh38
rs989857482 Chr.4: 1211754-1211755 on Build GRCh38
rs993944421 Chr.4: 1211897-1211897 on Build GRCh38
rs932379883 Chr.4: 1212018-1212018 on Build GRCh38
rs1042963534 Chr.4: 1211615-1211615 on Build GRCh38
rs576753551 Chr.4: 1211976-1211976 on Build GRCh38
rs905713261 Chr.4: 1211621-1211621 on Build GRCh38
rs1029804010 Chr.4: 1212076-1212076 on Build GRCh38
rs527281344 Chr.4: 1211686-1211686 on Build GRCh38
rs1011421493 Chr.4: 1211614-1211614 on Build GRCh38
rs976914355 Chr.4: 1211807-1211807 on Build GRCh38
rs562305900 Chr.4: 1212089-1212089 on Build GRCh38
rs1025626835 Chr.4: 1211689-1211689 on Build GRCh38
rs1008565041 Chr.4: 1211625-1211625 on Build GRCh38
rs982116635 Chr.4: 1211705-1211705 on Build GRCh38
rs887794181 Chr.4: 1212099-1212099 on Build GRCh38
rs937177832 Chr.4: 1211984-1211984 on Build GRCh38
rs77379446 Chr.4: 1211733-1211733 on Build GRCh38
rs1036216618 Chr.4: 1211668-1211668 on Build GRCh38
rs879622540 Chr.4: 1211623-1211623 on Build GRCh38
rs1037386363 Chr.4: 1211890-1211890 on Build GRCh38
rs923017738 Chr.4: 1211886-1211886 on Build GRCh38
rs952953013 Chr.4: 1211714-1211714 on Build GRCh38
rs147222661 Chr.4: 1211630-1211630 on Build GRCh38
rs888488723 Chr.4: 1212055-1212055 on Build GRCh38
rs914358719 Chr.4: 1211775-1211775 on Build GRCh38
rs751473672 Chr.4: 1211780-1211781 on Build GRCh38

View Full Product Details