Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 38796370-38796900 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TLR1
Entrez Gene ID
7096
Gene Name
toll like receptor 1
Gene Aliases
CD281, TIL, TIL. LPRS5, rsc786
UniGene
Hs.621817
Gene Chromosome Location
Chr.4: 38787555-38805800 on Build GRCh38
Transcripts
NM_003263.3 XM_005262662.4 XM_011513742.2 XM_011513743.2 XM_011513744.2 XM_011513745.2 XM_017008571.1 XM_017008572.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3603514 c.2344G>A p.E782K Chr.4: 38796488-38796488 on Build GRCh38
5011270 c.2148C>A p.H716Q Chr.4: 38796684-38796684 on Build GRCh38
7194113 c.2042A>T p.E681V Chr.4: 38796790-38796790 on Build GRCh38
1429559 c.2136C>A p.Y712* Chr.4: 38796696-38796696 on Build GRCh38
3993655 c.2339T>A p.L780Q Chr.4: 38796493-38796493 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766981900 Chr.4: 38796529-38796529 on Build GRCh38
rs201743225 Chr.4: 38796790-38796790 on Build GRCh38
rs752847915 Chr.4: 38796555-38796555 on Build GRCh38
rs758511588 Chr.4: 38796561-38796561 on Build GRCh38
rs1037452192 Chr.4: 38796593-38796593 on Build GRCh38
rs938524085 Chr.4: 38796723-38796723 on Build GRCh38
rs201203468 Chr.4: 38796468-38796468 on Build GRCh38
rs779826162 Chr.4: 38796574-38796574 on Build GRCh38
rs778712360 Chr.4: 38796777-38796777 on Build GRCh38
rs141789763 Chr.4: 38796696-38796696 on Build GRCh38
rs750106853 Chr.4: 38796870-38796870 on Build GRCh38
rs745646625 Chr.4: 38796567-38796567 on Build GRCh38
rs148014858 Chr.4: 38796801-38796801 on Build GRCh38
rs187624160 Chr.4: 38796778-38796778 on Build GRCh38
rs995657631 Chr.4: 38796617-38796617 on Build GRCh38
rs763872659 Chr.4: 38796526-38796527 on Build GRCh38
rs41305843 Chr.4: 38796614-38796614 on Build GRCh38
rs138709743 Chr.4: 38796824-38796824 on Build GRCh38
rs907828093 Chr.4: 38796775-38796775 on Build GRCh38
rs779775263 Chr.4: 38796669-38796669 on Build GRCh38
rs770733242 Chr.4: 38796712-38796712 on Build GRCh38
rs759987918 Chr.4: 38796720-38796720 on Build GRCh38
rs762515837 Chr.4: 38796462-38796462 on Build GRCh38
rs763452642 Chr.4: 38796750-38796750 on Build GRCh38
rs756776247 Chr.4: 38796726-38796727 on Build GRCh38
rs770680181 Chr.4: 38796797-38796797 on Build GRCh38
rs371529271 Chr.4: 38796500-38796500 on Build GRCh38
rs145802478 Chr.4: 38796621-38796621 on Build GRCh38
rs773692791 Chr.4: 38796599-38796599 on Build GRCh38
rs751660047 Chr.4: 38796756-38796756 on Build GRCh38
rs940359088 Chr.4: 38796585-38796585 on Build GRCh38
rs200356797 Chr.4: 38796526-38796526 on Build GRCh38
rs367864546 Chr.4: 38796465-38796465 on Build GRCh38
rs41311402 Chr.4: 38796742-38796742 on Build GRCh38
rs752691684 Chr.4: 38796633-38796633 on Build GRCh38
rs769181591 Chr.4: 38796808-38796808 on Build GRCh38
rs5743620 Chr.4: 38796864-38796864 on Build GRCh38
rs372163988 Chr.4: 38796807-38796807 on Build GRCh38
rs757430331 Chr.4: 38796760-38796760 on Build GRCh38
rs200811735 Chr.4: 38796603-38796603 on Build GRCh38
rs764294322 Chr.4: 38796650-38796650 on Build GRCh38
rs56205407 Chr.4: 38796796-38796796 on Build GRCh38
rs761172313 Chr.4: 38796524-38796524 on Build GRCh38
rs867428031 Chr.4: 38796525-38796526 on Build GRCh38
rs780918602 Chr.4: 38796422-38796422 on Build GRCh38
rs753553782 Chr.4: 38796772-38796772 on Build GRCh38
rs745697697 Chr.4: 38796426-38796426 on Build GRCh38
rs778804422 Chr.4: 38796548-38796548 on Build GRCh38
rs764099685 Chr.4: 38796754-38796754 on Build GRCh38
rs564294259 Chr.4: 38796702-38796702 on Build GRCh38
rs201148050 Chr.4: 38796682-38796682 on Build GRCh38
rs753809555 Chr.4: 38796654-38796655 on Build GRCh38
rs768660931 Chr.4: 38796597-38796597 on Build GRCh38
rs752564378 Chr.4: 38796879-38796879 on Build GRCh38
rs751709241 Chr.4: 38796651-38796651 on Build GRCh38
rs771828972 Chr.4: 38796604-38796604 on Build GRCh38
rs765642823 Chr.4: 38796730-38796730 on Build GRCh38
rs1028913676 Chr.4: 38796618-38796618 on Build GRCh38
rs774382003 Chr.4: 38796809-38796809 on Build GRCh38
rs545555159 Chr.4: 38796428-38796428 on Build GRCh38
rs1019823380 Chr.4: 38796816-38796816 on Build GRCh38
rs778956415 Chr.4: 38796684-38796684 on Build GRCh38
rs777372219 Chr.4: 38796780-38796780 on Build GRCh38
rs773058398 Chr.4: 38796612-38796612 on Build GRCh38
rs140268578 Chr.4: 38796860-38796860 on Build GRCh38
rs151285692 Chr.4: 38796493-38796493 on Build GRCh38
rs775469175 Chr.4: 38796765-38796765 on Build GRCh38
rs747598922 Chr.4: 38796685-38796685 on Build GRCh38
rs759915354 Chr.4: 38796628-38796628 on Build GRCh38
rs758425255 Chr.4: 38796779-38796779 on Build GRCh38
rs137853175 Chr.4: 38796812-38796812 on Build GRCh38
rs912583990 Chr.4: 38796418-38796418 on Build GRCh38
rs986725974 Chr.4: 38796405-38796405 on Build GRCh38
rs137853174 Chr.4: 38796880-38796880 on Build GRCh38
rs956021377 Chr.4: 38796672-38796672 on Build GRCh38
rs143548463 Chr.4: 38796766-38796766 on Build GRCh38
rs772853375 Chr.4: 38796701-38796701 on Build GRCh38
rs144733677 Chr.4: 38796489-38796489 on Build GRCh38
rs780950630 Chr.4: 38796798-38796798 on Build GRCh38
rs143251194 Chr.4: 38796715-38796715 on Build GRCh38
rs377263744 Chr.4: 38796563-38796563 on Build GRCh38
rs141132868 Chr.4: 38796632-38796632 on Build GRCh38
rs757481200 Chr.4: 38796667-38796667 on Build GRCh38
rs145159325 Chr.4: 38796627-38796627 on Build GRCh38
rs992723971 Chr.4: 38796769-38796769 on Build GRCh38
rs5743621 Chr.4: 38796634-38796634 on Build GRCh38
rs755967784 Chr.4: 38796571-38796571 on Build GRCh38
rs767620465 Chr.4: 38796834-38796834 on Build GRCh38
rs748594696 Chr.4: 38796519-38796519 on Build GRCh38
rs749120670 Chr.4: 38796583-38796583 on Build GRCh38
rs41311400 Chr.4: 38796804-38796804 on Build GRCh38
rs375393776 Chr.4: 38796434-38796434 on Build GRCh38
rs768946074 Chr.4: 38796534-38796534 on Build GRCh38
rs113706342 Chr.4: 38796673-38796673 on Build GRCh38
rs542161166 Chr.4: 38796539-38796539 on Build GRCh38
rs755514578 Chr.4: 38796543-38796543 on Build GRCh38

View Full Product Details