Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 121672317-121672817 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANXA5
Entrez Gene ID
308
Gene Name
annexin A5
Gene Aliases
ANX5, ENX2, HEL-S-7, PP4, RPRGL3
UniGene
Hs.480653
Gene Chromosome Location
Chr.4: 121667997-121696992 on Build GRCh38
Transcripts
NM_001154.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5672857 c.625+3G>T p.? Chr.4: 121672530-121672530 on Build GRCh38
7209103 c.553C>A p.L185I Chr.4: 121672605-121672605 on Build GRCh38
4662159 c.601C>T p.R201* Chr.4: 121672557-121672557 on Build GRCh38
5785635 c.537A>C p.L179F Chr.4: 121672621-121672621 on Build GRCh38
3725684 c.618G>A p.L206L Chr.4: 121672540-121672540 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs542133861 Chr.4: 121672575-121672575 on Build GRCh38
rs146935113 Chr.4: 121672533-121672533 on Build GRCh38
rs765777930 Chr.4: 121672545-121672545 on Build GRCh38
rs187894444 Chr.4: 121672479-121672479 on Build GRCh38
rs147110745 Chr.4: 121672557-121672557 on Build GRCh38
rs750780903 Chr.4: 121672548-121672548 on Build GRCh38
rs1019751257 Chr.4: 121672462-121672465 on Build GRCh38
rs952705883 Chr.4: 121672526-121672526 on Build GRCh38
rs137877895 Chr.4: 121672553-121672553 on Build GRCh38
rs760985496 Chr.4: 121672517-121672517 on Build GRCh38
rs1015168276 Chr.4: 121672365-121672365 on Build GRCh38
rs1052852555 Chr.4: 121672430-121672430 on Build GRCh38
rs1003867692 Chr.4: 121672727-121672727 on Build GRCh38
rs772196450 Chr.4: 121672518-121672519 on Build GRCh38
rs561854322 Chr.4: 121672646-121672646 on Build GRCh38
rs117028412 Chr.4: 121672372-121672372 on Build GRCh38
rs182739597 Chr.4: 121672412-121672412 on Build GRCh38
rs74981340 Chr.4: 121672344-121672344 on Build GRCh38
rs890234090 Chr.4: 121672448-121672448 on Build GRCh38
rs948538805 Chr.4: 121672722-121672722 on Build GRCh38
rs568239590 Chr.4: 121672738-121672738 on Build GRCh38
rs140583301 Chr.4: 121672774-121672774 on Build GRCh38
rs373045443 Chr.4: 121672455-121672455 on Build GRCh38
rs771723289 Chr.4: 121672676-121672676 on Build GRCh38
rs3214942 Chr.4: 121672478-121672479 on Build GRCh38
rs527367919 Chr.4: 121672599-121672599 on Build GRCh38
rs748874524 Chr.4: 121672578-121672578 on Build GRCh38
rs747637188 Chr.4: 121672508-121672508 on Build GRCh38
rs759289771 Chr.4: 121672500-121672500 on Build GRCh38
rs963638984 Chr.4: 121672761-121672761 on Build GRCh38
rs978642161 Chr.4: 121672734-121672734 on Build GRCh38
rs377645865 Chr.4: 121672483-121672483 on Build GRCh38
rs997893081 Chr.4: 121672610-121672610 on Build GRCh38
rs753925200 Chr.4: 121672543-121672543 on Build GRCh38
rs995427330 Chr.4: 121672789-121672789 on Build GRCh38
rs1052338174 Chr.4: 121672506-121672506 on Build GRCh38
rs916669450 Chr.4: 121672434-121672434 on Build GRCh38
rs770960160 Chr.4: 121672597-121672597 on Build GRCh38
rs904150971 Chr.4: 121672407-121672407 on Build GRCh38
rs370660162 Chr.4: 121672513-121672513 on Build GRCh38
rs761699421 Chr.4: 121672701-121672701 on Build GRCh38
rs886806194 Chr.4: 121672725-121672725 on Build GRCh38
rs575361087 Chr.4: 121672647-121672647 on Build GRCh38
rs774283232 Chr.4: 121672490-121672490 on Build GRCh38
rs745838009 Chr.4: 121672672-121672672 on Build GRCh38
rs775422068 Chr.4: 121672654-121672657 on Build GRCh38
rs554845190 Chr.4: 121672342-121672342 on Build GRCh38
rs925898361 Chr.4: 121672737-121672737 on Build GRCh38
rs935332046 Chr.4: 121672504-121672504 on Build GRCh38
rs775523941 Chr.4: 121672514-121672514 on Build GRCh38
rs982086054 Chr.4: 121672475-121672475 on Build GRCh38
rs1015613891 Chr.4: 121672745-121672745 on Build GRCh38
rs771050168 Chr.4: 121672489-121672489 on Build GRCh38
rs57273550 Chr.4: 121672450-121672450 on Build GRCh38
rs141889791 Chr.4: 121672556-121672556 on Build GRCh38
rs762291276 Chr.4: 121672544-121672544 on Build GRCh38
rs759402511 Chr.4: 121672498-121672498 on Build GRCh38
rs556979824 Chr.4: 121672405-121672405 on Build GRCh38

View Full Product Details