Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 176184741-176185301 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPATA4
Entrez Gene ID
132851
Gene Name
spermatogenesis associated 4
Gene Aliases
SPEF1B, TSARG2
UniGene
Hs.481235
Gene Chromosome Location
Chr.4: 176184574-176195671 on Build GRCh38
Transcripts
NM_144644.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
315542 c.838G>T p.E280* Chr.4: 176184860-176184860 on Build GRCh38
4123518 c.870T>G p.S290S Chr.4: 176184828-176184828 on Build GRCh38
7131929 c.878C>A p.S293Y Chr.4: 176184820-176184820 on Build GRCh38
4386206 c.844C>T p.H282Y Chr.4: 176184854-176184854 on Build GRCh38
4943987 c.877T>A p.S293T Chr.4: 176184821-176184821 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs148258175 Chr.4: 176185051-176185051 on Build GRCh38
rs562455301 Chr.4: 176184868-176184868 on Build GRCh38
rs28381989 Chr.4: 176184859-176184862 on Build GRCh38
rs531101418 Chr.4: 176184869-176184869 on Build GRCh38
rs750976030 Chr.4: 176184975-176184975 on Build GRCh38
rs761199318 Chr.4: 176184799-176184799 on Build GRCh38
rs972730897 Chr.4: 176184945-176184945 on Build GRCh38
rs772486353 Chr.4: 176184790-176184790 on Build GRCh38
rs558274925 Chr.4: 176184966-176184966 on Build GRCh38
rs183375789 Chr.4: 176185237-176185237 on Build GRCh38
rs894209148 Chr.4: 176184960-176184960 on Build GRCh38
rs574642828 Chr.4: 176185195-176185195 on Build GRCh38
rs539827715 Chr.4: 176184767-176184767 on Build GRCh38
rs1019386467 Chr.4: 176185209-176185209 on Build GRCh38
rs557700823 Chr.4: 176185108-176185108 on Build GRCh38
rs774566292 Chr.4: 176184784-176184784 on Build GRCh38
rs777748923 Chr.4: 176184771-176184771 on Build GRCh38
rs187094395 Chr.4: 176185048-176185048 on Build GRCh38
rs201475322 Chr.4: 176184860-176184860 on Build GRCh38
rs772816307 Chr.4: 176184936-176184936 on Build GRCh38
rs368179395 Chr.4: 176184860-176184863 on Build GRCh38
rs557254815 Chr.4: 176184901-176184901 on Build GRCh38
rs964154812 Chr.4: 176185058-176185058 on Build GRCh38
rs1057314989 Chr.4: 176185042-176185042 on Build GRCh38
rs750156432 Chr.4: 176184835-176184835 on Build GRCh38
rs1047315565 Chr.4: 176185098-176185098 on Build GRCh38
rs145109730 Chr.4: 176184853-176184853 on Build GRCh38
rs935798768 Chr.4: 176184773-176184773 on Build GRCh38
rs754894435 Chr.4: 176184858-176184858 on Build GRCh38
rs548622141 Chr.4: 176184796-176184796 on Build GRCh38
rs938889999 Chr.4: 176185038-176185038 on Build GRCh38
rs542234219 Chr.4: 176185241-176185241 on Build GRCh38
rs575670151 Chr.4: 176185054-176185054 on Build GRCh38
rs761320451 Chr.4: 176184810-176184810 on Build GRCh38
rs550918092 Chr.4: 176184872-176184872 on Build GRCh38
rs762212328 Chr.4: 176184922-176184922 on Build GRCh38
rs188829575 Chr.4: 176185265-176185265 on Build GRCh38
rs372324492 Chr.4: 176184900-176184900 on Build GRCh38
rs908785995 Chr.4: 176185123-176185123 on Build GRCh38
rs368507758 Chr.4: 176184864-176184864 on Build GRCh38
rs753296995 Chr.4: 176184809-176184809 on Build GRCh38
rs775851890 Chr.4: 176184886-176184886 on Build GRCh38
rs189752082 Chr.4: 176184991-176184991 on Build GRCh38
rs1044883259 Chr.4: 176184961-176184961 on Build GRCh38
rs764850058 Chr.4: 176184923-176184923 on Build GRCh38
rs975869982 Chr.4: 176185064-176185064 on Build GRCh38
rs1038498617 Chr.4: 176185178-176185178 on Build GRCh38
rs190222898 Chr.4: 176185072-176185072 on Build GRCh38
rs146281879 Chr.4: 176185218-176185218 on Build GRCh38
rs771071567 Chr.4: 176184780-176184780 on Build GRCh38
rs766081599 Chr.4: 176184943-176184943 on Build GRCh38
rs866743861 Chr.4: 176184793-176184794 on Build GRCh38
rs764647834 Chr.4: 176184826-176184826 on Build GRCh38
rs1023963848 Chr.4: 176184988-176184988 on Build GRCh38
rs934267882 Chr.4: 176184820-176184820 on Build GRCh38
rs746140676 Chr.4: 176184870-176184870 on Build GRCh38
rs1000495002 Chr.4: 176185055-176185055 on Build GRCh38
rs887294225 Chr.4: 176184881-176184881 on Build GRCh38
rs547497972 Chr.4: 176185274-176185274 on Build GRCh38
rs922630922 Chr.4: 176185065-176185065 on Build GRCh38
rs747313337 Chr.4: 176184896-176184896 on Build GRCh38
rs764610137 Chr.4: 176184806-176184806 on Build GRCh38
rs141922460 Chr.4: 176184852-176184852 on Build GRCh38
rs977386256 Chr.4: 176184995-176184995 on Build GRCh38
rs758099717 Chr.4: 176184851-176184851 on Build GRCh38
rs10004325 Chr.4: 176185138-176185138 on Build GRCh38
rs75438443 Chr.4: 176185050-176185050 on Build GRCh38
rs73005175 Chr.4: 176184789-176184789 on Build GRCh38
rs919821302 Chr.4: 176185259-176185259 on Build GRCh38
rs143076076 Chr.4: 176184775-176184775 on Build GRCh38
rs182417543 Chr.4: 176185043-176185043 on Build GRCh38

View Full Product Details