Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 145653862-145654397 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MMAA
Entrez Gene ID
166785
Gene Name
methylmalonic aciduria (cobalamin deficiency) cblA type
Gene Aliases
cblA
UniGene
Hs.452864
Gene Chromosome Location
Chr.4: 145619388-145660035 on Build GRCh38
Transcripts
NM_172250.2 XM_011531684.2 XM_011531685.2 XM_011531686.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7076498 c.899G>A p.R300Q Chr.4: 145654073-145654073 on Build GRCh38
1051767 c.911C>T p.A304V Chr.4: 145654085-145654085 on Build GRCh38
1427389 c.840C>T p.I280I Chr.4: 145654014-145654014 on Build GRCh38
276084 c.898C>T p.R300* Chr.4: 145654072-145654072 on Build GRCh38
1051768 c.940C>T p.R314C Chr.4: 145654114-145654114 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs989186816 Chr.4: 145654128-145654128 on Build GRCh38
rs903547988 Chr.4: 145654149-145654149 on Build GRCh38
rs146352309 Chr.4: 145654053-145654053 on Build GRCh38
rs867001059 Chr.4: 145654254-145654254 on Build GRCh38
rs1037579070 Chr.4: 145654231-145654231 on Build GRCh38
rs1033150411 Chr.4: 145654344-145654344 on Build GRCh38
rs761964238 Chr.4: 145654007-145654007 on Build GRCh38
rs377228966 Chr.4: 145654121-145654121 on Build GRCh38
rs980392859 Chr.4: 145654082-145654082 on Build GRCh38
rs142063642 Chr.4: 145654032-145654032 on Build GRCh38
rs188844104 Chr.4: 145654227-145654227 on Build GRCh38
rs997282360 Chr.4: 145653976-145653976 on Build GRCh38
rs148142853 Chr.4: 145654115-145654115 on Build GRCh38
rs769242756 Chr.4: 145654234-145654234 on Build GRCh38
rs1014359518 Chr.4: 145654308-145654308 on Build GRCh38
rs774958165 Chr.4: 145653999-145654000 on Build GRCh38
rs777773616 Chr.4: 145654058-145654058 on Build GRCh38
rs141974563 Chr.4: 145654120-145654120 on Build GRCh38
rs1016732691 Chr.4: 145654282-145654282 on Build GRCh38
rs774068339 Chr.4: 145653956-145653956 on Build GRCh38
rs1007611523 Chr.4: 145654152-145654152 on Build GRCh38
rs1026370614 Chr.4: 145654245-145654245 on Build GRCh38
rs769082931 Chr.4: 145654101-145654101 on Build GRCh38
rs17225095 Chr.4: 145653884-145653884 on Build GRCh38
rs889065873 Chr.4: 145654188-145654188 on Build GRCh38
rs200254160 Chr.4: 145654004-145654004 on Build GRCh38
rs758654806 Chr.4: 145654024-145654024 on Build GRCh38
rs751717131 Chr.4: 145654126-145654126 on Build GRCh38
rs752514914 Chr.4: 145654046-145654046 on Build GRCh38
rs749124431 Chr.4: 145654070-145654070 on Build GRCh38
rs746714195 Chr.4: 145654190-145654190 on Build GRCh38
rs778354704 Chr.4: 145654077-145654077 on Build GRCh38
rs1035950442 Chr.4: 145654343-145654343 on Build GRCh38
rs1038948135 Chr.4: 145654161-145654161 on Build GRCh38
rs58912137 Chr.4: 145653893-145653893 on Build GRCh38
rs368766697 Chr.4: 145654092-145654092 on Build GRCh38
rs745565526 Chr.4: 145653958-145653958 on Build GRCh38
rs886059089 Chr.4: 145654036-145654036 on Build GRCh38
rs1023064586 Chr.4: 145654089-145654089 on Build GRCh38
rs770176887 Chr.4: 145653963-145653963 on Build GRCh38
rs757913614 Chr.4: 145654185-145654185 on Build GRCh38
rs993267869 Chr.4: 145654238-145654238 on Build GRCh38
rs371624542 Chr.4: 145654010-145654010 on Build GRCh38
rs759260546 Chr.4: 145653967-145653968 on Build GRCh38
rs765287661 Chr.4: 145654012-145654012 on Build GRCh38
rs183941385 Chr.4: 145653934-145653934 on Build GRCh38
rs371769807 Chr.4: 145654086-145654086 on Build GRCh38
rs368666860 Chr.4: 145654281-145654281 on Build GRCh38
rs374165891 Chr.4: 145654187-145654187 on Build GRCh38
rs868815345 Chr.4: 145653892-145653892 on Build GRCh38
rs978896004 Chr.4: 145654342-145654342 on Build GRCh38
rs749982700 Chr.4: 145654184-145654184 on Build GRCh38
rs374795215 Chr.4: 145654114-145654114 on Build GRCh38
rs369128670 Chr.4: 145654078-145654078 on Build GRCh38
rs771873633 Chr.4: 145654085-145654085 on Build GRCh38
rs757207009 Chr.4: 145654073-145654073 on Build GRCh38
rs538710234 Chr.4: 145654166-145654166 on Build GRCh38
rs760693374 Chr.4: 145654318-145654318 on Build GRCh38
rs190750337 Chr.4: 145653930-145653930 on Build GRCh38
rs150692463 Chr.4: 145654140-145654140 on Build GRCh38
rs1053243530 Chr.4: 145654104-145654104 on Build GRCh38
rs757182784 Chr.4: 145654162-145654162 on Build GRCh38
rs765080486 Chr.4: 145654138-145654138 on Build GRCh38
rs750509072 Chr.4: 145654014-145654014 on Build GRCh38
rs764728312 Chr.4: 145653997-145653997 on Build GRCh38
rs568373078 Chr.4: 145653955-145653955 on Build GRCh38
rs369712158 Chr.4: 145654158-145654158 on Build GRCh38
rs985276517 Chr.4: 145654036-145654036 on Build GRCh38
rs980470081 Chr.4: 145654339-145654339 on Build GRCh38
rs888972294 Chr.4: 145653941-145653941 on Build GRCh38
rs368025764 Chr.4: 145653991-145653991 on Build GRCh38
rs776045663 Chr.4: 145653970-145653970 on Build GRCh38
rs778860275 Chr.4: 145654163-145654163 on Build GRCh38
rs773218154 Chr.4: 145654116-145654116 on Build GRCh38
rs760236568 Chr.4: 145654008-145654009 on Build GRCh38
rs548975889 Chr.4: 145653983-145653983 on Build GRCh38
rs1026142191 Chr.4: 145654214-145654214 on Build GRCh38
rs901730572 Chr.4: 145654175-145654175 on Build GRCh38

View Full Product Details