Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 41194299-41194898 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TREML2
Entrez Gene ID
79865
Gene Name
triggering receptor expressed on myeloid cells like 2
Gene Aliases
C6orf76, TLT-2, TLT2, dJ238O23.1
UniGene
Hs.164797
Gene Chromosome Location
Chr.6: 41189749-41201233 on Build GRCh38
Transcripts
NM_024807.3 XM_011514917.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4735837 c.650A>G p.D217G Chr.6: 41194737-41194737 on Build GRCh38
6106603 c.813C>A p.T271T Chr.6: 41194574-41194574 on Build GRCh38
4620489 c.577C>T p.P193S Chr.6: 41194810-41194810 on Build GRCh38
3762065 c.563C>G p.T188S Chr.6: 41194824-41194824 on Build GRCh38
1643073 c.555T>C p.A185A Chr.6: 41194832-41194832 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs917378040 Chr.6: 41194439-41194439 on Build GRCh38
rs377248699 Chr.6: 41194853-41194853 on Build GRCh38
rs115991880 Chr.6: 41194466-41194466 on Build GRCh38
rs1050864003 Chr.6: 41194801-41194801 on Build GRCh38
rs574402521 Chr.6: 41194867-41194867 on Build GRCh38
rs979978805 Chr.6: 41194462-41194462 on Build GRCh38
rs768840352 Chr.6: 41194507-41194507 on Build GRCh38
rs561006845 Chr.6: 41194819-41194819 on Build GRCh38
rs115024549 Chr.6: 41194828-41194828 on Build GRCh38
rs3747742 Chr.6: 41194780-41194780 on Build GRCh38
rs750874759 Chr.6: 41194694-41194694 on Build GRCh38
rs560704107 Chr.6: 41194577-41194577 on Build GRCh38
rs369023699 Chr.6: 41194463-41194463 on Build GRCh38
rs777687651 Chr.6: 41194857-41194857 on Build GRCh38
rs747119388 Chr.6: 41194429-41194429 on Build GRCh38
rs776381738 Chr.6: 41194873-41194873 on Build GRCh38
rs561280519 Chr.6: 41194325-41194325 on Build GRCh38
rs531665321 Chr.6: 41194551-41194551 on Build GRCh38
rs771412277 Chr.6: 41194498-41194498 on Build GRCh38
rs780996699 Chr.6: 41194385-41194385 on Build GRCh38
rs759003328 Chr.6: 41194856-41194856 on Build GRCh38
rs759731078 Chr.6: 41194440-41194440 on Build GRCh38
rs767531955 Chr.6: 41194545-41194545 on Build GRCh38
rs747454081 Chr.6: 41194495-41194495 on Build GRCh38
rs556690771 Chr.6: 41194644-41194644 on Build GRCh38
rs770927652 Chr.6: 41194435-41194435 on Build GRCh38
rs772160027 Chr.6: 41194619-41194619 on Build GRCh38
rs755765965 Chr.6: 41194467-41194467 on Build GRCh38
rs748517434 Chr.6: 41194417-41194417 on Build GRCh38
rs756566346 Chr.6: 41194701-41194701 on Build GRCh38
rs369223607 Chr.6: 41194855-41194855 on Build GRCh38
rs770766858 Chr.6: 41194767-41194767 on Build GRCh38
rs774329903 Chr.6: 41194611-41194611 on Build GRCh38
rs745927895 Chr.6: 41194510-41194510 on Build GRCh38
rs572065459 Chr.6: 41194763-41194763 on Build GRCh38
rs775186269 Chr.6: 41194450-41194450 on Build GRCh38
rs545085836 Chr.6: 41194732-41194732 on Build GRCh38
rs781646812 Chr.6: 41194586-41194586 on Build GRCh38
rs146102372 Chr.6: 41194762-41194762 on Build GRCh38
rs757567572 Chr.6: 41194661-41194661 on Build GRCh38
rs755604012 Chr.6: 41194556-41194556 on Build GRCh38
rs765854421 Chr.6: 41194640-41194640 on Build GRCh38
rs141822677 Chr.6: 41194574-41194574 on Build GRCh38
rs61734887 Chr.6: 41194824-41194824 on Build GRCh38
rs866093005 Chr.6: 41194566-41194567 on Build GRCh38
rs914510258 Chr.6: 41194425-41194425 on Build GRCh38
rs755071758 Chr.6: 41194852-41194852 on Build GRCh38
rs775731926 Chr.6: 41194513-41194513 on Build GRCh38
rs745460534 Chr.6: 41194399-41194399 on Build GRCh38
rs370336343 Chr.6: 41194826-41194826 on Build GRCh38
rs140877740 Chr.6: 41194494-41194494 on Build GRCh38
rs998309615 Chr.6: 41194550-41194550 on Build GRCh38
rs576566590 Chr.6: 41194670-41194670 on Build GRCh38
rs371732807 Chr.6: 41194382-41194382 on Build GRCh38
rs769876403 Chr.6: 41194588-41194588 on Build GRCh38
rs551450784 Chr.6: 41194871-41194871 on Build GRCh38
rs759022688 Chr.6: 41194792-41194792 on Build GRCh38
rs372822570 Chr.6: 41194493-41194493 on Build GRCh38
rs760720511 Chr.6: 41194765-41194765 on Build GRCh38
rs747297666 Chr.6: 41194818-41194818 on Build GRCh38
rs749299214 Chr.6: 41194721-41194721 on Build GRCh38
rs567985004 Chr.6: 41194616-41194616 on Build GRCh38
rs896774295 Chr.6: 41194783-41194783 on Build GRCh38
rs185375765 Chr.6: 41194459-41194459 on Build GRCh38
rs753995605 Chr.6: 41194848-41194848 on Build GRCh38
rs1021463877 Chr.6: 41194699-41194699 on Build GRCh38
rs970263731 Chr.6: 41194709-41194709 on Build GRCh38
rs759249846 Chr.6: 41194647-41194647 on Build GRCh38
rs769957038 Chr.6: 41194511-41194511 on Build GRCh38
rs761910388 Chr.6: 41194528-41194528 on Build GRCh38
rs374114737 Chr.6: 41194803-41194803 on Build GRCh38
rs762687733 Chr.6: 41194515-41194515 on Build GRCh38
rs1031360865 Chr.6: 41194350-41194350 on Build GRCh38
rs774274973 Chr.6: 41194527-41194527 on Build GRCh38
rs142007199 Chr.6: 41194438-41194438 on Build GRCh38
rs897223564 Chr.6: 41194502-41194502 on Build GRCh38
rs143539514 Chr.6: 41194571-41194571 on Build GRCh38
rs956090772 Chr.6: 41194401-41194401 on Build GRCh38
rs551464347 Chr.6: 41194559-41194559 on Build GRCh38
rs752306363 Chr.6: 41194813-41194813 on Build GRCh38
rs779737509 Chr.6: 41194475-41194475 on Build GRCh38
rs755779850 Chr.6: 41194411-41194411 on Build GRCh38
rs745956797 Chr.6: 41194587-41194587 on Build GRCh38
rs933667343 Chr.6: 41194796-41194796 on Build GRCh38
rs779232431 Chr.6: 41194742-41194742 on Build GRCh38
rs763726067 Chr.6: 41194628-41194630 on Build GRCh38
rs972718952 Chr.6: 41194800-41194800 on Build GRCh38
rs182617084 Chr.6: 41194820-41194820 on Build GRCh38
rs773665047 Chr.6: 41194427-41194427 on Build GRCh38
rs369601224 Chr.6: 41194657-41194657 on Build GRCh38
rs145455750 Chr.6: 41194633-41194633 on Build GRCh38
rs779213711 Chr.6: 41194412-41194412 on Build GRCh38
rs773032359 Chr.6: 41194764-41194764 on Build GRCh38
rs768997331 Chr.6: 41194602-41194602 on Build GRCh38
rs533971302 Chr.6: 41194573-41194573 on Build GRCh38
rs147159415 Chr.6: 41194572-41194572 on Build GRCh38
rs372479731 Chr.6: 41194483-41194483 on Build GRCh38
rs763925087 Chr.6: 41194526-41194526 on Build GRCh38
rs13216201 Chr.6: 41194322-41194322 on Build GRCh38
rs772509209 Chr.6: 41194422-41194422 on Build GRCh38
rs776756608 Chr.6: 41194505-41194505 on Build GRCh38
rs760111540 Chr.6: 41194637-41194637 on Build GRCh38
rs749582081 Chr.6: 41194601-41194601 on Build GRCh38
rs377165160 Chr.6: 41194648-41194648 on Build GRCh38
rs780153970 Chr.6: 41194590-41194590 on Build GRCh38
rs746746495 Chr.6: 41194868-41194868 on Build GRCh38
rs765490544 Chr.6: 41194442-41194442 on Build GRCh38
rs1006718674 Chr.6: 41194565-41194565 on Build GRCh38
rs941580073 Chr.6: 41194726-41194726 on Build GRCh38

View Full Product Details