Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 125993360-125993959 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRMT11
Entrez Gene ID
60487
Gene Name
tRNA methyltransferase 11 homolog
Gene Aliases
C6orf75, MDS024, TRM11, TRMT11-1
UniGene
Hs.404186
Gene Chromosome Location
Chr.6: 125986430-126083131 on Build GRCh38
Transcripts
NM_001031712.2 XM_017011168.1 XM_017011169.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1230465 c.128C>G p.T43S Chr.6: 125993812-125993812 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs138395697 Chr.6: 125993822-125993822 on Build GRCh38
rs74726612 Chr.6: 125993848-125993848 on Build GRCh38
rs779578639 Chr.6: 125993643-125993643 on Build GRCh38
rs556392016 Chr.6: 125993614-125993614 on Build GRCh38
rs778327424 Chr.6: 125993777-125993777 on Build GRCh38
rs1039430119 Chr.6: 125993933-125993933 on Build GRCh38
rs999550042 Chr.6: 125993667-125993667 on Build GRCh38
rs567727079 Chr.6: 125993844-125993844 on Build GRCh38
rs1023687940 Chr.6: 125993766-125993766 on Build GRCh38
rs377072623 Chr.6: 125993671-125993671 on Build GRCh38
rs749948701 Chr.6: 125993796-125993796 on Build GRCh38
rs1022291458 Chr.6: 125993401-125993401 on Build GRCh38
rs753728954 Chr.6: 125993754-125993754 on Build GRCh38
rs762899899 Chr.6: 125993929-125993929 on Build GRCh38
rs757903982 Chr.6: 125993798-125993798 on Build GRCh38
rs190321362 Chr.6: 125993858-125993858 on Build GRCh38
rs1039007754 Chr.6: 125993525-125993525 on Build GRCh38
rs1032664352 Chr.6: 125993689-125993689 on Build GRCh38
rs752447655 Chr.6: 125993713-125993713 on Build GRCh38
rs546855576 Chr.6: 125993650-125993650 on Build GRCh38
rs911488914 Chr.6: 125993469-125993469 on Build GRCh38
rs1041623856 Chr.6: 125993647-125993647 on Build GRCh38
rs916734049 Chr.6: 125993444-125993444 on Build GRCh38
rs7760163 Chr.6: 125993445-125993445 on Build GRCh38
rs974326420 Chr.6: 125993447-125993447 on Build GRCh38
rs772085147 Chr.6: 125993870-125993870 on Build GRCh38
rs752929025 Chr.6: 125993736-125993736 on Build GRCh38
rs1013220248 Chr.6: 125993625-125993625 on Build GRCh38
rs763928016 Chr.6: 125993752-125993752 on Build GRCh38
rs538144449 Chr.6: 125993574-125993574 on Build GRCh38
rs929673203 Chr.6: 125993495-125993495 on Build GRCh38
rs181684577 Chr.6: 125993451-125993451 on Build GRCh38
rs759651862 Chr.6: 125993603-125993603 on Build GRCh38
rs770907827 Chr.6: 125993419-125993419 on Build GRCh38
rs368754731 Chr.6: 125993792-125993792 on Build GRCh38
rs960848873 Chr.6: 125993694-125993694 on Build GRCh38
rs956572434 Chr.6: 125993692-125993692 on Build GRCh38
rs757215984 Chr.6: 125993768-125993768 on Build GRCh38
rs779747932 Chr.6: 125993810-125993810 on Build GRCh38
rs552478217 Chr.6: 125993547-125993547 on Build GRCh38
rs766171763 Chr.6: 125993860-125993865 on Build GRCh38
rs529297630 Chr.6: 125993452-125993452 on Build GRCh38
rs974361039 Chr.6: 125993917-125993917 on Build GRCh38
rs942504430 Chr.6: 125993931-125993931 on Build GRCh38
rs930101991 Chr.6: 125993551-125993551 on Build GRCh38
rs991025225 Chr.6: 125993430-125993430 on Build GRCh38
rs746676930 Chr.6: 125993816-125993816 on Build GRCh38
rs966684882 Chr.6: 125993403-125993403 on Build GRCh38
rs568520870 Chr.6: 125993843-125993843 on Build GRCh38
rs549098027 Chr.6: 125993725-125993725 on Build GRCh38
rs749749263 Chr.6: 125993832-125993832 on Build GRCh38
rs1020384297 Chr.6: 125993808-125993808 on Build GRCh38
rs911249897 Chr.6: 125993896-125993896 on Build GRCh38
rs755907068 Chr.6: 125993751-125993751 on Build GRCh38
rs571161214 Chr.6: 125993548-125993548 on Build GRCh38
rs951700031 Chr.6: 125993838-125993838 on Build GRCh38
rs964037082 Chr.6: 125993905-125993905 on Build GRCh38
rs185855587 Chr.6: 125993521-125993521 on Build GRCh38
rs961050527 Chr.6: 125993429-125993429 on Build GRCh38
rs978463317 Chr.6: 125993404-125993404 on Build GRCh38
rs760650371 Chr.6: 125993828-125993833 on Build GRCh38
rs770170482 Chr.6: 125993819-125993819 on Build GRCh38
rs1035412959 Chr.6: 125993693-125993693 on Build GRCh38

View Full Product Details