Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 44212359-44212905 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
YKT6
Entrez Gene ID
10652
Gene Name
YKT6 v-SNARE homolog (S. cerevisiae)
Gene Aliases
-
UniGene
Hs.520794
Gene Chromosome Location
Chr.7: 44200968-44214294 on Build GRCh38
Transcripts
NM_006555.3 XM_005249582.4 XM_017011682.1

SNP Detail

SNP ID Chromosome Location
rs558826473 Chr.7: 44212521-44212521 on Build GRCh38
rs941759667 Chr.7: 44212764-44212764 on Build GRCh38
rs1005900593 Chr.7: 44212475-44212475 on Build GRCh38
rs1021661617 Chr.7: 44212835-44212835 on Build GRCh38
rs541334282 Chr.7: 44212655-44212655 on Build GRCh38
rs765320888 Chr.7: 44212468-44212468 on Build GRCh38
rs932957397 Chr.7: 44212805-44212805 on Build GRCh38
rs902859786 Chr.7: 44212607-44212607 on Build GRCh38
rs980416133 Chr.7: 44212666-44212666 on Build GRCh38
rs1037752073 Chr.7: 44212490-44212490 on Build GRCh38
rs1017475621 Chr.7: 44212746-44212746 on Build GRCh38
rs1055329918 Chr.7: 44212461-44212461 on Build GRCh38
rs928489520 Chr.7: 44212700-44212700 on Build GRCh38
rs983255701 Chr.7: 44212759-44212759 on Build GRCh38
rs948620685 Chr.7: 44212415-44212415 on Build GRCh38
rs113418097 Chr.7: 44212593-44212593 on Build GRCh38
rs878877586 Chr.7: 44212758-44212758 on Build GRCh38
rs1035614503 Chr.7: 44212886-44212886 on Build GRCh38
rs938496431 Chr.7: 44212755-44212755 on Build GRCh38
rs3840675 Chr.7: 44212589-44212590 on Build GRCh38
rs11540053 Chr.7: 44212846-44212846 on Build GRCh38
rs898774893 Chr.7: 44212494-44212494 on Build GRCh38
rs970460158 Chr.7: 44212838-44212838 on Build GRCh38
rs577203698 Chr.7: 44212634-44212634 on Build GRCh38
rs368059901 Chr.7: 44212566-44212567 on Build GRCh38
rs181872409 Chr.7: 44212420-44212420 on Build GRCh38
rs78352778 Chr.7: 44212409-44212409 on Build GRCh38
rs3840673 Chr.7: 44212710-44212710 on Build GRCh38
rs3840674 Chr.7: 44212592-44212593 on Build GRCh38
rs376447625 Chr.7: 44212834-44212834 on Build GRCh38
rs1008362201 Chr.7: 44212829-44212830 on Build GRCh38
rs976486557 Chr.7: 44212383-44212383 on Build GRCh38
rs866482712 Chr.7: 44212829-44212829 on Build GRCh38
rs1014045774 Chr.7: 44212588-44212588 on Build GRCh38
rs1037403163 Chr.7: 44212767-44212767 on Build GRCh38
rs12670584 Chr.7: 44212419-44212419 on Build GRCh38
rs78455736 Chr.7: 44212404-44212404 on Build GRCh38
rs907645757 Chr.7: 44212762-44212762 on Build GRCh38
rs950735355 Chr.7: 44212885-44212885 on Build GRCh38
rs1012058849 Chr.7: 44212831-44212831 on Build GRCh38
rs79911862 Chr.7: 44212594-44212594 on Build GRCh38
rs768417300 Chr.7: 44212777-44212778 on Build GRCh38
rs976199481 Chr.7: 44212821-44212821 on Build GRCh38
rs998743008 Chr.7: 44212612-44212612 on Build GRCh38
rs879198983 Chr.7: 44212802-44212802 on Build GRCh38
rs560456450 Chr.7: 44212830-44212830 on Build GRCh38
rs201234587 Chr.7: 44212709-44212709 on Build GRCh38
rs1032947796 Chr.7: 44212624-44212624 on Build GRCh38
rs938162137 Chr.7: 44212438-44212438 on Build GRCh38
rs536306411 Chr.7: 44212408-44212408 on Build GRCh38
rs957338535 Chr.7: 44212630-44212630 on Build GRCh38
rs879129239 Chr.7: 44212807-44212807 on Build GRCh38
rs752321674 Chr.7: 44212412-44212412 on Build GRCh38
rs966484333 Chr.7: 44212806-44212806 on Build GRCh38
rs1045386252 Chr.7: 44212603-44212603 on Build GRCh38
rs886149889 Chr.7: 44212465-44212465 on Build GRCh38
rs1047508344 Chr.7: 44212458-44212458 on Build GRCh38

View Full Product Details