Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 213167761-213168137 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPS6KC1
Entrez Gene ID
26750
Gene Name
ribosomal protein S6 kinase C1
Gene Aliases
RPK118, RSKL1, S6K-delta-1, S6PKh1, humS6PKh1
UniGene
Hs.591416
Gene Chromosome Location
Chr.1: 213051233-213378753 on Build GRCh38
Transcripts
NM_001136138.2 NM_001287218.1 NM_001287219.1 NM_001287220.1 NM_001287221.1 NM_012424.4 XM_005273095.1 XM_005273096.1 XM_006711272.1 XM_017001024.1 XM_017001025.1 XM_017001026.1 XM_017001027.1 XM_017001028.1 XM_017001029.1 XM_017001030.1 XM_017001031.1 XM_017001032.1 XM_017001033.1 XM_017001034.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4027964 c.885G>A p.E295E Chr.1: 213167907-213167907 on Build GRCh38
6449208 c.872G>A p.R291K Chr.1: 213167894-213167894 on Build GRCh38
5978225 c.869A>G p.K290R Chr.1: 213167891-213167891 on Build GRCh38
4562808 c.943G>T p.V315L Chr.1: 213167965-213167965 on Build GRCh38
4778416 c.837A>G p.G279G Chr.1: 213167859-213167859 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1032127796 Chr.1: 213168077-213168077 on Build GRCh38
rs116712428 Chr.1: 213168094-213168094 on Build GRCh38
rs955490881 Chr.1: 213168100-213168100 on Build GRCh38
rs752252959 Chr.1: 213167842-213167842 on Build GRCh38
rs759476637 Chr.1: 213167960-213167960 on Build GRCh38
rs760522018 Chr.1: 213167807-213167807 on Build GRCh38
rs766315724 Chr.1: 213167818-213167818 on Build GRCh38
rs762499668 Chr.1: 213167968-213167968 on Build GRCh38
rs957771239 Chr.1: 213168085-213168085 on Build GRCh38
rs753822486 Chr.1: 213167821-213167821 on Build GRCh38
rs761043603 Chr.1: 213167917-213167917 on Build GRCh38
rs754206613 Chr.1: 213167993-213167993 on Build GRCh38
rs373118301 Chr.1: 213167976-213167976 on Build GRCh38
rs1055066798 Chr.1: 213167860-213167860 on Build GRCh38
rs764590329 Chr.1: 213167828-213167828 on Build GRCh38
rs200954000 Chr.1: 213167904-213167904 on Build GRCh38
rs1047585955 Chr.1: 213167936-213167936 on Build GRCh38
rs779498093 Chr.1: 213168008-213168008 on Build GRCh38
rs879055764 Chr.1: 213167826-213167826 on Build GRCh38
rs755024267 Chr.1: 213167827-213167827 on Build GRCh38
rs1013848256 Chr.1: 213168023-213168023 on Build GRCh38
rs200774550 Chr.1: 213167847-213167847 on Build GRCh38
rs772186099 Chr.1: 213168021-213168021 on Build GRCh38
rs900180687 Chr.1: 213168010-213168010 on Build GRCh38
rs769165349 Chr.1: 213167885-213167885 on Build GRCh38
rs997485411 Chr.1: 213168086-213168086 on Build GRCh38
rs969584224 Chr.1: 213168063-213168063 on Build GRCh38
rs374003961 Chr.1: 213167894-213167894 on Build GRCh38
rs773450346 Chr.1: 213167914-213167914 on Build GRCh38
rs1050537762 Chr.1: 213167905-213167905 on Build GRCh38
rs1014878313 Chr.1: 213167969-213167969 on Build GRCh38
rs372751505 Chr.1: 213167837-213167837 on Build GRCh38
rs780338157 Chr.1: 213167981-213167981 on Build GRCh38
rs987828478 Chr.1: 213168093-213168093 on Build GRCh38
rs148984854 Chr.1: 213167875-213167875 on Build GRCh38
rs943674217 Chr.1: 213168106-213168106 on Build GRCh38
rs756442120 Chr.1: 213167862-213167862 on Build GRCh38
rs778792223 Chr.1: 213167891-213167891 on Build GRCh38
rs773204726 Chr.1: 213167917-213167917 on Build GRCh38
rs1025622775 Chr.1: 213168051-213168051 on Build GRCh38
rs748641104 Chr.1: 213168018-213168018 on Build GRCh38
rs556701673 Chr.1: 213167962-213167962 on Build GRCh38
rs757944948 Chr.1: 213167843-213167843 on Build GRCh38
rs763556033 Chr.1: 213167972-213167972 on Build GRCh38
rs866202416 Chr.1: 213168110-213168110 on Build GRCh38
rs370851168 Chr.1: 213167918-213167918 on Build GRCh38
rs755401942 Chr.1: 213167995-213167995 on Build GRCh38
rs769608427 Chr.1: 213167855-213167856 on Build GRCh38
rs748063612 Chr.1: 213167845-213167846 on Build GRCh38
rs886188654 Chr.1: 213167945-213167945 on Build GRCh38
rs567683596 Chr.1: 213167846-213167846 on Build GRCh38
rs981313228 Chr.1: 213168067-213168068 on Build GRCh38
rs887831036 Chr.1: 213167934-213167934 on Build GRCh38
rs368914061 Chr.1: 213167872-213167872 on Build GRCh38
rs1005952247 Chr.1: 213167964-213167964 on Build GRCh38
rs746715230 Chr.1: 213167855-213167855 on Build GRCh38
rs112694881 Chr.1: 213168036-213168036 on Build GRCh38

View Full Product Details