Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 34648866-34649260 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EHF
Entrez Gene ID
26298
Gene Name
ETS homologous factor
Gene Aliases
ESE3, ESE3B, ESEJ
UniGene
Hs.653859
Gene Chromosome Location
Chr.11: 34621041-34663288 on Build GRCh38
Transcripts
NM_001206615.1 NM_001206616.1 NM_012153.5 XM_005252857.1 XM_005252860.1 XM_005252861.1 XM_005252862.1 XM_011519983.1 XM_011519984.1 XM_011519985.1 XM_017017534.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4032540 c.383T>C p.V128A Chr.11: 34649058-34649058 on Build GRCh38
466770 c.363G>C p.L121L Chr.11: 34649038-34649038 on Build GRCh38
1251043 c.398A>G p.E133G Chr.11: 34649073-34649073 on Build GRCh38
3943639 c.395C>T p.T132I Chr.11: 34649070-34649070 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs761346519 Chr.11: 34649088-34649088 on Build GRCh38
rs372714599 Chr.11: 34649012-34649012 on Build GRCh38
rs758213492 Chr.11: 34648997-34648997 on Build GRCh38
rs556378793 Chr.11: 34648948-34648948 on Build GRCh38
rs373692532 Chr.11: 34649065-34649065 on Build GRCh38
rs777969161 Chr.11: 34649011-34649011 on Build GRCh38
rs961241127 Chr.11: 34649105-34649105 on Build GRCh38
rs1023656132 Chr.11: 34649052-34649052 on Build GRCh38
rs761685993 Chr.11: 34649192-34649192 on Build GRCh38
rs375908067 Chr.11: 34649129-34649129 on Build GRCh38
rs769388192 Chr.11: 34649089-34649089 on Build GRCh38
rs1049415930 Chr.11: 34649175-34649175 on Build GRCh38
rs762672767 Chr.11: 34649091-34649091 on Build GRCh38
rs764988391 Chr.11: 34648987-34648987 on Build GRCh38
rs1008123827 Chr.11: 34649195-34649195 on Build GRCh38
rs766251032 Chr.11: 34649004-34649004 on Build GRCh38
rs902697132 Chr.11: 34648947-34648947 on Build GRCh38
rs1027108228 Chr.11: 34649232-34649232 on Build GRCh38
rs766268838 Chr.11: 34649094-34649094 on Build GRCh38
rs186968926 Chr.11: 34649231-34649231 on Build GRCh38
rs12721519 Chr.11: 34649059-34649059 on Build GRCh38
rs527885621 Chr.11: 34648968-34648968 on Build GRCh38
rs578013619 Chr.11: 34649130-34649130 on Build GRCh38
rs368395080 Chr.11: 34649106-34649106 on Build GRCh38
rs370700655 Chr.11: 34649124-34649124 on Build GRCh38
rs866935399 Chr.11: 34649204-34649204 on Build GRCh38
rs753916514 Chr.11: 34649111-34649111 on Build GRCh38
rs922537373 Chr.11: 34649149-34649149 on Build GRCh38
rs189376267 Chr.11: 34649008-34649008 on Build GRCh38
rs540501039 Chr.11: 34649132-34649132 on Build GRCh38
rs774130080 Chr.11: 34649184-34649184 on Build GRCh38
rs549297659 Chr.11: 34649070-34649070 on Build GRCh38
rs553741335 Chr.11: 34649212-34649212 on Build GRCh38
rs1035718617 Chr.11: 34648981-34648981 on Build GRCh38
rs933903370 Chr.11: 34649156-34649156 on Build GRCh38
rs750180047 Chr.11: 34648992-34648992 on Build GRCh38
rs767383602 Chr.11: 34649110-34649110 on Build GRCh38
rs991253204 Chr.11: 34649013-34649013 on Build GRCh38
rs536753826 Chr.11: 34648937-34648937 on Build GRCh38
rs775856876 Chr.11: 34649038-34649038 on Build GRCh38
rs961777886 Chr.11: 34648945-34648945 on Build GRCh38
rs376867946 Chr.11: 34649006-34649006 on Build GRCh38
rs757422615 Chr.11: 34649115-34649115 on Build GRCh38
rs1050082206 Chr.11: 34649164-34649164 on Build GRCh38
rs757512472 Chr.11: 34649023-34649023 on Build GRCh38
rs147842712 Chr.11: 34649054-34649054 on Build GRCh38
rs767451287 Chr.11: 34649219-34649219 on Build GRCh38
rs779296141 Chr.11: 34649027-34649027 on Build GRCh38
rs772569143 Chr.11: 34649031-34649031 on Build GRCh38
rs946350871 Chr.11: 34649185-34649185 on Build GRCh38
rs772866372 Chr.11: 34649090-34649090 on Build GRCh38
rs181535835 Chr.11: 34649128-34649128 on Build GRCh38
rs749029382 Chr.11: 34648954-34648954 on Build GRCh38
rs746173974 Chr.11: 34649030-34649030 on Build GRCh38
rs897013323 Chr.11: 34649229-34649229 on Build GRCh38
rs1021969461 Chr.11: 34648931-34648931 on Build GRCh38
rs567511305 Chr.11: 34648892-34648892 on Build GRCh38
rs373947441 Chr.11: 34648980-34648980 on Build GRCh38
rs1027469585 Chr.11: 34648979-34648979 on Build GRCh38

View Full Product Details