Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 107961748-107962338 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAB39A
Entrez Gene ID
54734
Gene Name
RAB39A, member RAS oncogene family
Gene Aliases
RAB39
UniGene
Hs.354906
Gene Chromosome Location
Chr.11: 107928475-107963482 on Build GRCh38
Transcripts
NM_017516.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7103291 c.316G>C p.D106H Chr.11: 107962034-107962034 on Build GRCh38
4902935 c.267A>G p.G89G Chr.11: 107961985-107961985 on Build GRCh38
7386635 c.331G>T p.A111S Chr.11: 107962049-107962049 on Build GRCh38
4957454 c.518G>A p.R173K Chr.11: 107962236-107962236 on Build GRCh38
7128291 c.288T>A p.T96T Chr.11: 107962006-107962006 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs954645867 Chr.11: 107961908-107961908 on Build GRCh38
rs181805673 Chr.11: 107962245-107962245 on Build GRCh38
rs913294528 Chr.11: 107961785-107961785 on Build GRCh38
rs932249971 Chr.11: 107961874-107961874 on Build GRCh38
rs752948052 Chr.11: 107962294-107962294 on Build GRCh38
rs748278240 Chr.11: 107962067-107962067 on Build GRCh38
rs745689266 Chr.11: 107961946-107961946 on Build GRCh38
rs755105920 Chr.11: 107962175-107962175 on Build GRCh38
rs759971573 Chr.11: 107962234-107962234 on Build GRCh38
rs537914931 Chr.11: 107962056-107962056 on Build GRCh38
rs756805872 Chr.11: 107962131-107962131 on Build GRCh38
rs923028810 Chr.11: 107961890-107961890 on Build GRCh38
rs756378799 Chr.11: 107962302-107962302 on Build GRCh38
rs766719712 Chr.11: 107962111-107962111 on Build GRCh38
rs566759898 Chr.11: 107961773-107961773 on Build GRCh38
rs760217812 Chr.11: 107962121-107962121 on Build GRCh38
rs754805450 Chr.11: 107962275-107962275 on Build GRCh38
rs146648885 Chr.11: 107962241-107962241 on Build GRCh38
rs947369774 Chr.11: 107961841-107961841 on Build GRCh38
rs374766991 Chr.11: 107961911-107961911 on Build GRCh38
rs764816095 Chr.11: 107962257-107962257 on Build GRCh38
rs189062613 Chr.11: 107962060-107962060 on Build GRCh38
rs779959964 Chr.11: 107962158-107962158 on Build GRCh38
rs142969707 Chr.11: 107962153-107962153 on Build GRCh38
rs780700252 Chr.11: 107962051-107962051 on Build GRCh38
rs773710320 Chr.11: 107962081-107962081 on Build GRCh38
rs77231534 Chr.11: 107961994-107961994 on Build GRCh38
rs774749712 Chr.11: 107962120-107962120 on Build GRCh38
rs763317978 Chr.11: 107961982-107961982 on Build GRCh38
rs747510652 Chr.11: 107962277-107962278 on Build GRCh38
rs755623771 Chr.11: 107961914-107961914 on Build GRCh38
rs72992527 Chr.11: 107961872-107961872 on Build GRCh38
rs777889656 Chr.11: 107962202-107962202 on Build GRCh38
rs1031935100 Chr.11: 107961855-107961855 on Build GRCh38
rs572739559 Chr.11: 107962263-107962263 on Build GRCh38
rs745603013 Chr.11: 107962037-107962037 on Build GRCh38
rs763515951 Chr.11: 107962100-107962100 on Build GRCh38
rs748109452 Chr.11: 107962181-107962181 on Build GRCh38
rs776819828 Chr.11: 107961959-107961959 on Build GRCh38
rs921909273 Chr.11: 107962247-107962247 on Build GRCh38
rs749990580 Chr.11: 107962261-107962261 on Build GRCh38
rs774659501 Chr.11: 107962226-107962226 on Build GRCh38
rs751427939 Chr.11: 107962160-107962160 on Build GRCh38
rs1052387116 Chr.11: 107961885-107961885 on Build GRCh38
rs747210795 Chr.11: 107962057-107962057 on Build GRCh38
rs765799648 Chr.11: 107961970-107961970 on Build GRCh38
rs753432336 Chr.11: 107962025-107962025 on Build GRCh38
rs780915113 Chr.11: 107962292-107962292 on Build GRCh38
rs1057448709 Chr.11: 107962163-107962163 on Build GRCh38
rs772372540 Chr.11: 107961956-107961956 on Build GRCh38
rs753697995 Chr.11: 107962014-107962014 on Build GRCh38
rs757272364 Chr.11: 107961925-107961925 on Build GRCh38
rs201799500 Chr.11: 107962124-107962124 on Build GRCh38
rs749775277 Chr.11: 107962205-107962205 on Build GRCh38
rs900789017 Chr.11: 107961873-107961873 on Build GRCh38
rs1052041488 Chr.11: 107962295-107962295 on Build GRCh38
rs758889349 Chr.11: 107961925-107961927 on Build GRCh38
rs750370765 Chr.11: 107962156-107962156 on Build GRCh38
rs979321200 Chr.11: 107961850-107961850 on Build GRCh38
rs201681603 Chr.11: 107962010-107962010 on Build GRCh38
rs780840979 Chr.11: 107961906-107961906 on Build GRCh38
rs754679607 Chr.11: 107962218-107962218 on Build GRCh38
rs549480565 Chr.11: 107962099-107962099 on Build GRCh38
rs757103206 Chr.11: 107962026-107962026 on Build GRCh38
rs778933509 Chr.11: 107962030-107962030 on Build GRCh38
rs756124351 Chr.11: 107961957-107961957 on Build GRCh38
rs770139140 Chr.11: 107962074-107962074 on Build GRCh38
rs771469391 Chr.11: 107962214-107962214 on Build GRCh38
rs781489700 Chr.11: 107962178-107962178 on Build GRCh38
rs753239418 Chr.11: 107962130-107962130 on Build GRCh38
rs1035989007 Chr.11: 107962058-107962058 on Build GRCh38
rs755856456 Chr.11: 107962013-107962013 on Build GRCh38
rs1010294702 Chr.11: 107962233-107962233 on Build GRCh38
rs779291941 Chr.11: 107962284-107962284 on Build GRCh38
rs749172460 Chr.11: 107961919-107961919 on Build GRCh38
rs200392395 Chr.11: 107961928-107961928 on Build GRCh38
rs777801190 Chr.11: 107962306-107962306 on Build GRCh38
rs549198709 Chr.11: 107961968-107961968 on Build GRCh38
rs766438012 Chr.11: 107962265-107962265 on Build GRCh38
rs758455976 Chr.11: 107962157-107962157 on Build GRCh38
rs751254039 Chr.11: 107962268-107962268 on Build GRCh38
rs369131963 Chr.11: 107961916-107961916 on Build GRCh38
rs111615525 Chr.11: 107962061-107962061 on Build GRCh38
rs778540979 Chr.11: 107961933-107961933 on Build GRCh38
rs761300643 Chr.11: 107961958-107961958 on Build GRCh38
rs942224223 Chr.11: 107962046-107962046 on Build GRCh38
rs769911013 Chr.11: 107962193-107962193 on Build GRCh38

View Full Product Details