Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 109850620-109851168 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLTP
Entrez Gene ID
51228
Gene Name
glycolipid transfer protein
Gene Aliases
-
UniGene
Hs.381256
Gene Chromosome Location
Chr.12: 109850943-109880488 on Build GRCh38
Transcripts
NM_016433.3 XM_011538434.2

SNP Detail

SNP ID Chromosome Location
rs1049509107 Chr.12: 109850898-109850898 on Build GRCh38
rs1040932428 Chr.12: 109850917-109850917 on Build GRCh38
rs1027896817 Chr.12: 109851122-109851122 on Build GRCh38
rs932588343 Chr.12: 109850934-109850934 on Build GRCh38
rs34299889 Chr.12: 109850949-109850950 on Build GRCh38
rs949924205 Chr.12: 109850741-109850741 on Build GRCh38
rs116403501 Chr.12: 109850932-109850932 on Build GRCh38
rs191833312 Chr.12: 109850681-109850681 on Build GRCh38
rs890876954 Chr.12: 109850902-109850902 on Build GRCh38
rs898103015 Chr.12: 109850926-109850926 on Build GRCh38
rs950866905 Chr.12: 109851136-109851136 on Build GRCh38
rs905846829 Chr.12: 109851103-109851103 on Build GRCh38
rs751499219 Chr.12: 109851137-109851137 on Build GRCh38
rs1009107163 Chr.12: 109850915-109850915 on Build GRCh38
rs934554346 Chr.12: 109850840-109850840 on Build GRCh38
rs976391735 Chr.12: 109850734-109850734 on Build GRCh38
rs1044823124 Chr.12: 109851092-109851092 on Build GRCh38
rs908254947 Chr.12: 109850697-109850697 on Build GRCh38
rs58370620 Chr.12: 109850746-109850746 on Build GRCh38
rs921204192 Chr.12: 109850910-109850910 on Build GRCh38
rs932272313 Chr.12: 109850857-109850857 on Build GRCh38
rs915955023 Chr.12: 109850738-109850738 on Build GRCh38
rs55975160 Chr.12: 109850998-109850998 on Build GRCh38
rs925813762 Chr.12: 109851000-109851000 on Build GRCh38
rs775786124 Chr.12: 109850816-109850816 on Build GRCh38
rs544997129 Chr.12: 109851010-109851010 on Build GRCh38
rs1028263563 Chr.12: 109850674-109850674 on Build GRCh38
rs761774566 Chr.12: 109851144-109851144 on Build GRCh38
rs764256146 Chr.12: 109851005-109851005 on Build GRCh38
rs914255331 Chr.12: 109850845-109850845 on Build GRCh38
rs995538726 Chr.12: 109851108-109851108 on Build GRCh38
rs995088138 Chr.12: 109850646-109850646 on Build GRCh38
rs1054354567 Chr.12: 109850844-109850844 on Build GRCh38
rs1042960518 Chr.12: 109850938-109850938 on Build GRCh38
rs562421649 Chr.12: 109850764-109850764 on Build GRCh38
rs184808023 Chr.12: 109850797-109850797 on Build GRCh38
rs560358757 Chr.12: 109851090-109851090 on Build GRCh38
rs187778234 Chr.12: 109851141-109851141 on Build GRCh38
rs991539962 Chr.12: 109850711-109850711 on Build GRCh38
rs895881502 Chr.12: 109851027-109851027 on Build GRCh38
rs981674724 Chr.12: 109850766-109850766 on Build GRCh38
rs965043264 Chr.12: 109850698-109850698 on Build GRCh38
rs1053135348 Chr.12: 109851016-109851016 on Build GRCh38
rs763191071 Chr.12: 109850831-109850831 on Build GRCh38
rs34346536 Chr.12: 109850768-109850769 on Build GRCh38
rs544041193 Chr.12: 109850860-109850860 on Build GRCh38
rs982561837 Chr.12: 109850689-109850689 on Build GRCh38
rs542973450 Chr.12: 109850660-109850660 on Build GRCh38
rs562326783 Chr.12: 109850930-109850930 on Build GRCh38

View Full Product Details