Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 41182559-41183135 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZHX3
Entrez Gene ID
23051
Gene Name
zinc fingers and homeoboxes 3
Gene Aliases
TIX1
UniGene
Hs.380133
Gene Chromosome Location
Chr.20: 41178449-41317606 on Build GRCh38
Transcripts
NM_015035.3 XM_005260341.3 XM_011528720.1 XM_017027737.1 XM_017027738.1

SNP Detail

SNP ID Chromosome Location
rs922546648 Chr.20: 41182954-41182954 on Build GRCh38
rs375450218 Chr.20: 41182671-41182671 on Build GRCh38
rs1012385959 Chr.20: 41182810-41182810 on Build GRCh38
rs1020946176 Chr.20: 41182798-41182798 on Build GRCh38
rs891706242 Chr.20: 41182768-41182768 on Build GRCh38
rs891737491 Chr.20: 41183016-41183016 on Build GRCh38
rs975215926 Chr.20: 41182934-41182934 on Build GRCh38
rs1012459420 Chr.20: 41183041-41183041 on Build GRCh38
rs894220236 Chr.20: 41183040-41183040 on Build GRCh38
rs375435098 Chr.20: 41182599-41182601 on Build GRCh38
rs575160315 Chr.20: 41182813-41182813 on Build GRCh38
rs933897631 Chr.20: 41182972-41182972 on Build GRCh38
rs915280244 Chr.20: 41182925-41182925 on Build GRCh38
rs537449620 Chr.20: 41182921-41182921 on Build GRCh38
rs116666507 Chr.20: 41182809-41182809 on Build GRCh38
rs577458611 Chr.20: 41182962-41182962 on Build GRCh38
rs963882493 Chr.20: 41182926-41182926 on Build GRCh38
rs1009597712 Chr.20: 41182773-41182773 on Build GRCh38
rs369697183 Chr.20: 41183105-41183105 on Build GRCh38
rs886888227 Chr.20: 41182622-41182622 on Build GRCh38
rs750898202 Chr.20: 41182758-41182758 on Build GRCh38
rs970965072 Chr.20: 41182850-41182850 on Build GRCh38
rs553724862 Chr.20: 41182580-41182580 on Build GRCh38
rs940983680 Chr.20: 41183008-41183008 on Build GRCh38
rs941788708 Chr.20: 41182958-41182958 on Build GRCh38
rs535364928 Chr.20: 41182674-41182674 on Build GRCh38
rs781358256 Chr.20: 41182997-41182997 on Build GRCh38
rs906811472 Chr.20: 41183099-41183099 on Build GRCh38
rs983092287 Chr.20: 41182952-41182952 on Build GRCh38
rs755198773 Chr.20: 41182902-41182902 on Build GRCh38
rs1038505033 Chr.20: 41182640-41182640 on Build GRCh38
rs749722791 Chr.20: 41182898-41182898 on Build GRCh38
rs1045236808 Chr.20: 41183057-41183057 on Build GRCh38
rs922516963 Chr.20: 41182583-41182583 on Build GRCh38
rs538347472 Chr.20: 41182602-41182602 on Build GRCh38
rs1038131597 Chr.20: 41183019-41183019 on Build GRCh38
rs1003975221 Chr.20: 41182857-41182857 on Build GRCh38
rs748814593 Chr.20: 41183102-41183103 on Build GRCh38
rs757803984 Chr.20: 41182613-41182613 on Build GRCh38
rs1018101630 Chr.20: 41182827-41182827 on Build GRCh38
rs929537022 Chr.20: 41182903-41182903 on Build GRCh38
rs142024567 Chr.20: 41182594-41182596 on Build GRCh38
rs1003118576 Chr.20: 41182627-41182627 on Build GRCh38
rs1030986497 Chr.20: 41182872-41182872 on Build GRCh38
rs1047393788 Chr.20: 41182986-41182986 on Build GRCh38
rs908220921 Chr.20: 41182993-41182993 on Build GRCh38
rs557380003 Chr.20: 41182946-41182946 on Build GRCh38
rs4297946 Chr.20: 41182635-41182635 on Build GRCh38
rs545957932 Chr.20: 41183018-41183018 on Build GRCh38
rs566121209 Chr.20: 41182664-41182664 on Build GRCh38
rs923570572 Chr.20: 41182896-41182896 on Build GRCh38

View Full Product Details