Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 27748751-27749334 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MN1
Entrez Gene ID
4330
Gene Name
MN1 proto-oncogene, transcriptional regulator
Gene Aliases
MGCR, MGCR1, MGCR1-PEN, dJ353E16.2
UniGene
Hs.268515
Gene Chromosome Location
Chr.22: 27748277-27801498 on Build GRCh38
Transcripts
NM_002430.2

SNP Detail

SNP ID Chromosome Location
rs895227617 Chr.22: 27749251-27749251 on Build GRCh38
rs915792461 Chr.22: 27749269-27749269 on Build GRCh38
rs990275535 Chr.22: 27749082-27749082 on Build GRCh38
rs373520564 Chr.22: 27749220-27749220 on Build GRCh38
rs904390566 Chr.22: 27749098-27749098 on Build GRCh38
rs998791186 Chr.22: 27748828-27748828 on Build GRCh38
rs993448938 Chr.22: 27748912-27748912 on Build GRCh38
rs186824463 Chr.22: 27748969-27748969 on Build GRCh38
rs5752632 Chr.22: 27749160-27749160 on Build GRCh38
rs776317787 Chr.22: 27748923-27748923 on Build GRCh38
rs915831171 Chr.22: 27749051-27749051 on Build GRCh38
rs998199456 Chr.22: 27749132-27749132 on Build GRCh38
rs1040200504 Chr.22: 27749105-27749105 on Build GRCh38
rs530431242 Chr.22: 27749156-27749156 on Build GRCh38
rs1054213351 Chr.22: 27748800-27748800 on Build GRCh38
rs181801322 Chr.22: 27749124-27749124 on Build GRCh38
rs1041361373 Chr.22: 27748789-27748789 on Build GRCh38
rs1040637035 Chr.22: 27748852-27748852 on Build GRCh38
rs954314846 Chr.22: 27748904-27748904 on Build GRCh38
rs1005699326 Chr.22: 27749173-27749173 on Build GRCh38
rs1017094464 Chr.22: 27749200-27749200 on Build GRCh38
rs570241659 Chr.22: 27749253-27749253 on Build GRCh38
rs535261762 Chr.22: 27748893-27748893 on Build GRCh38
rs925782940 Chr.22: 27749304-27749304 on Build GRCh38
rs908933235 Chr.22: 27748926-27748926 on Build GRCh38
rs948624232 Chr.22: 27749285-27749285 on Build GRCh38
rs938393334 Chr.22: 27749023-27749023 on Build GRCh38
rs978320401 Chr.22: 27749028-27749028 on Build GRCh38
rs1043081019 Chr.22: 27748776-27748776 on Build GRCh38
rs145466523 Chr.22: 27749000-27749000 on Build GRCh38
rs577291927 Chr.22: 27749094-27749094 on Build GRCh38
rs888921006 Chr.22: 27748834-27748834 on Build GRCh38
rs963140639 Chr.22: 27748937-27748937 on Build GRCh38
rs748546950 Chr.22: 27749014-27749014 on Build GRCh38
rs998836708 Chr.22: 27748863-27748863 on Build GRCh38
rs1026286637 Chr.22: 27748914-27748914 on Build GRCh38
rs190562335 Chr.22: 27749093-27749093 on Build GRCh38
rs929170025 Chr.22: 27749125-27749125 on Build GRCh38
rs901167534 Chr.22: 27749131-27749131 on Build GRCh38
rs148836234 Chr.22: 27749108-27749108 on Build GRCh38
rs1025106980 Chr.22: 27749257-27749257 on Build GRCh38
rs371015212 Chr.22: 27749237-27749237 on Build GRCh38
rs992791850 Chr.22: 27748772-27748772 on Build GRCh38
rs975965072 Chr.22: 27749155-27749155 on Build GRCh38
rs758005567 Chr.22: 27749021-27749021 on Build GRCh38
rs935685165 Chr.22: 27749035-27749035 on Build GRCh38
rs959750164 Chr.22: 27748956-27748956 on Build GRCh38
rs965438235 Chr.22: 27748857-27748857 on Build GRCh38
rs978287679 Chr.22: 27749268-27749268 on Build GRCh38
rs948529925 Chr.22: 27749070-27749070 on Build GRCh38
rs1028959410 Chr.22: 27749140-27749140 on Build GRCh38
rs531057439 Chr.22: 27749221-27749221 on Build GRCh38
rs117679774 Chr.22: 27748879-27748879 on Build GRCh38
rs549450628 Chr.22: 27748775-27748775 on Build GRCh38
rs41277847 Chr.22: 27748897-27748897 on Build GRCh38
rs552828118 Chr.22: 27748783-27748783 on Build GRCh38
rs532206175 Chr.22: 27749034-27749034 on Build GRCh38
rs1033783361 Chr.22: 27748948-27748948 on Build GRCh38
rs1032073198 Chr.22: 27748841-27748841 on Build GRCh38
rs534356667 Chr.22: 27748777-27748777 on Build GRCh38
rs569454948 Chr.22: 27749024-27749024 on Build GRCh38
rs904631623 Chr.22: 27748798-27748798 on Build GRCh38
rs45619034 Chr.22: 27749226-27749226 on Build GRCh38
rs779483397 Chr.22: 27748910-27748910 on Build GRCh38
rs978678703 Chr.22: 27749299-27749299 on Build GRCh38
rs868709649 Chr.22: 27749005-27749005 on Build GRCh38
rs45451602 Chr.22: 27749265-27749265 on Build GRCh38
rs1014121295 Chr.22: 27749252-27749252 on Build GRCh38

View Full Product Details