Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 73762069-73762634 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUSP11
Entrez Gene ID
8446
Gene Name
dual specificity phosphatase 11
Gene Aliases
PIR1
UniGene
Hs.14611
Gene Chromosome Location
Chr.2: 73762184-73780157 on Build GRCh38
Transcripts
NM_003584.2

SNP Detail

SNP ID Chromosome Location
rs565111873 Chr.2: 73762198-73762198 on Build GRCh38
rs11538238 Chr.2: 73762367-73762367 on Build GRCh38
rs977499815 Chr.2: 73762106-73762106 on Build GRCh38
rs928446881 Chr.2: 73762213-73762213 on Build GRCh38
rs938821464 Chr.2: 73762139-73762139 on Build GRCh38
rs565676184 Chr.2: 73762585-73762585 on Build GRCh38
rs543712606 Chr.2: 73762173-73762173 on Build GRCh38
rs559620942 Chr.2: 73762281-73762281 on Build GRCh38
rs578021368 Chr.2: 73762303-73762303 on Build GRCh38
rs139620490 Chr.2: 73762216-73762216 on Build GRCh38
rs550115739 Chr.2: 73762547-73762547 on Build GRCh38
rs1802053 Chr.2: 73762462-73762462 on Build GRCh38
rs149754457 Chr.2: 73762313-73762313 on Build GRCh38
rs1051237775 Chr.2: 73762461-73762461 on Build GRCh38
rs576290322 Chr.2: 73762156-73762156 on Build GRCh38
rs893680044 Chr.2: 73762516-73762516 on Build GRCh38
rs544930304 Chr.2: 73762175-73762175 on Build GRCh38
rs983330103 Chr.2: 73762221-73762221 on Build GRCh38
rs553291652 Chr.2: 73762288-73762292 on Build GRCh38
rs563703790 Chr.2: 73762343-73762343 on Build GRCh38
rs931592203 Chr.2: 73762217-73762217 on Build GRCh38
rs548248080 Chr.2: 73762329-73762329 on Build GRCh38
rs547464771 Chr.2: 73762579-73762579 on Build GRCh38
rs749242144 Chr.2: 73762328-73762328 on Build GRCh38
rs768638383 Chr.2: 73762550-73762550 on Build GRCh38
rs765366940 Chr.2: 73762143-73762143 on Build GRCh38
rs571449114 Chr.2: 73762553-73762553 on Build GRCh38
rs555236215 Chr.2: 73762121-73762121 on Build GRCh38
rs145553625 Chr.2: 73762358-73762358 on Build GRCh38
rs908879681 Chr.2: 73762237-73762237 on Build GRCh38
rs181058487 Chr.2: 73762219-73762219 on Build GRCh38
rs189857806 Chr.2: 73762119-73762119 on Build GRCh38
rs35245567 Chr.2: 73762400-73762401 on Build GRCh38
rs933057517 Chr.2: 73762432-73762432 on Build GRCh38
rs900934519 Chr.2: 73762322-73762322 on Build GRCh38
rs372817267 Chr.2: 73762355-73762355 on Build GRCh38
rs538861797 Chr.2: 73762563-73762563 on Build GRCh38
rs939731139 Chr.2: 73762296-73762296 on Build GRCh38
rs1031671795 Chr.2: 73762091-73762091 on Build GRCh38
rs1028158704 Chr.2: 73762383-73762384 on Build GRCh38

View Full Product Details