Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 79086693-79087069 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
REG1B
Entrez Gene ID
5968
Gene Name
regenerating family member 1 beta
Gene Aliases
PSPS2, REGH, REGI-BETA, REGL
UniGene
Hs.4158
Gene Chromosome Location
Chr.2: 79085023-79088024 on Build GRCh38
Transcripts
NM_006507.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
722369 c.85C>A p.L29M Chr.2: 79086910-79086910 on Build GRCh38
3583532 c.154G>A p.E52K Chr.2: 79086841-79086841 on Build GRCh38
196869 c.88C>T p.P30S Chr.2: 79086907-79086907 on Build GRCh38
2150735 c.71A>G p.E24G Chr.2: 79086924-79086924 on Build GRCh38
3583533 c.94C>T p.P32S Chr.2: 79086901-79086901 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374789460 Chr.2: 79086798-79086798 on Build GRCh38
rs772764057 Chr.2: 79086762-79086763 on Build GRCh38
rs1054688102 Chr.2: 79086945-79086945 on Build GRCh38
rs1006196217 Chr.2: 79087027-79087027 on Build GRCh38
rs144244545 Chr.2: 79086728-79086728 on Build GRCh38
rs375323595 Chr.2: 79086904-79086904 on Build GRCh38
rs771263591 Chr.2: 79086723-79086723 on Build GRCh38
rs767791154 Chr.2: 79086915-79086915 on Build GRCh38
rs995402757 Chr.2: 79087030-79087030 on Build GRCh38
rs376912875 Chr.2: 79086954-79086954 on Build GRCh38
rs747713468 Chr.2: 79086884-79086884 on Build GRCh38
rs983946747 Chr.2: 79086849-79086849 on Build GRCh38
rs773113096 Chr.2: 79086756-79086756 on Build GRCh38
rs758583034 Chr.2: 79086719-79086719 on Build GRCh38
rs771872974 Chr.2: 79086818-79086818 on Build GRCh38
rs769395955 Chr.2: 79086726-79086726 on Build GRCh38
rs1018048474 Chr.2: 79086987-79086987 on Build GRCh38
rs370382444 Chr.2: 79086786-79086786 on Build GRCh38
rs962492377 Chr.2: 79087021-79087021 on Build GRCh38
rs62640882 Chr.2: 79086864-79086864 on Build GRCh38
rs779502058 Chr.2: 79086944-79086944 on Build GRCh38
rs746797132 Chr.2: 79086896-79086896 on Build GRCh38
rs1055189936 Chr.2: 79086736-79086736 on Build GRCh38
rs751772817 Chr.2: 79086861-79086861 on Build GRCh38
rs1051321140 Chr.2: 79086963-79086963 on Build GRCh38
rs185604160 Chr.2: 79086898-79086898 on Build GRCh38
rs755341588 Chr.2: 79086932-79086933 on Build GRCh38
rs766889732 Chr.2: 79086937-79086937 on Build GRCh38
rs1013969312 Chr.2: 79086766-79086766 on Build GRCh38
rs374698692 Chr.2: 79086973-79086973 on Build GRCh38
rs755387990 Chr.2: 79086942-79086942 on Build GRCh38
rs34303807 Chr.2: 79086946-79086946 on Build GRCh38
rs757336197 Chr.2: 79086981-79086988 on Build GRCh38
rs561732090 Chr.2: 79086715-79086715 on Build GRCh38
rs773880840 Chr.2: 79086744-79086744 on Build GRCh38
rs774256969 Chr.2: 79086908-79086908 on Build GRCh38
rs772955313 Chr.2: 79086832-79086832 on Build GRCh38
rs766338935 Chr.2: 79086770-79086770 on Build GRCh38
rs751783770 Chr.2: 79086778-79086778 on Build GRCh38
rs777401881 Chr.2: 79086952-79086952 on Build GRCh38
rs193284335 Chr.2: 79086717-79086717 on Build GRCh38
rs757913102 Chr.2: 79086951-79086951 on Build GRCh38
rs372283770 Chr.2: 79086897-79086897 on Build GRCh38
rs761785197 Chr.2: 79086981-79086981 on Build GRCh38
rs763072147 Chr.2: 79086848-79086848 on Build GRCh38
rs771580503 Chr.2: 79086888-79086888 on Build GRCh38
rs780702420 Chr.2: 79086870-79086870 on Build GRCh38
rs766251204 Chr.2: 79086834-79086834 on Build GRCh38
rs533079813 Chr.2: 79086751-79086751 on Build GRCh38
rs528952213 Chr.2: 79086850-79086850 on Build GRCh38
rs757616510 Chr.2: 79086785-79086785 on Build GRCh38
rs531039910 Chr.2: 79086937-79086938 on Build GRCh38
rs752141270 Chr.2: 79086894-79086895 on Build GRCh38
rs11548892 Chr.2: 79086893-79086893 on Build GRCh38
rs6723688 Chr.2: 79086968-79086968 on Build GRCh38
rs760649403 Chr.2: 79086765-79086765 on Build GRCh38
rs367866883 Chr.2: 79086773-79086773 on Build GRCh38
rs111936697 Chr.2: 79086937-79086937 on Build GRCh38
rs760488377 Chr.2: 79086833-79086833 on Build GRCh38
rs753897494 Chr.2: 79086953-79086954 on Build GRCh38
rs6738657 Chr.2: 79086955-79086955 on Build GRCh38
rs749826554 Chr.2: 79086874-79086874 on Build GRCh38
rs778334069 Chr.2: 79086878-79086878 on Build GRCh38
rs546147932 Chr.2: 79086802-79086802 on Build GRCh38
rs777918837 Chr.2: 79086952-79086955 on Build GRCh38
rs761115284 Chr.2: 79086936-79086936 on Build GRCh38
rs886474475 Chr.2: 79086835-79086835 on Build GRCh38
rs368421116 Chr.2: 79086946-79086947 on Build GRCh38
rs781713506 Chr.2: 79086725-79086725 on Build GRCh38
rs955155684 Chr.2: 79086899-79086899 on Build GRCh38
rs72913219 Chr.2: 79086730-79086730 on Build GRCh38
rs747328231 Chr.2: 79086722-79086722 on Build GRCh38
rs777885449 Chr.2: 79086720-79086720 on Build GRCh38
rs139007123 Chr.2: 79086914-79086914 on Build GRCh38
rs768118109 Chr.2: 79086805-79086805 on Build GRCh38
rs776613553 Chr.2: 79086842-79086842 on Build GRCh38
rs755039763 Chr.2: 79086714-79086714 on Build GRCh38
rs768297299 Chr.2: 79086741-79086741 on Build GRCh38
rs750732443 Chr.2: 79086916-79086916 on Build GRCh38
rs746264577 Chr.2: 79086790-79086790 on Build GRCh38
rs754478614 Chr.2: 79086876-79086876 on Build GRCh38
rs775991773 Chr.2: 79086772-79086772 on Build GRCh38
rs796255251 Chr.2: 79086937-79086938 on Build GRCh38
rs377645272 Chr.2: 79086865-79086865 on Build GRCh38
rs754345931 Chr.2: 79086939-79086939 on Build GRCh38
rs372727754 Chr.2: 79086836-79086836 on Build GRCh38
rs747806649 Chr.2: 79086814-79086814 on Build GRCh38
rs762379799 Chr.2: 79086803-79086806 on Build GRCh38
rs779016386 Chr.2: 79086716-79086716 on Build GRCh38
rs756359139 Chr.2: 79086936-79086937 on Build GRCh38
rs766922489 Chr.2: 79086853-79086855 on Build GRCh38
rs764369175 Chr.2: 79086776-79086776 on Build GRCh38
rs761665860 Chr.2: 79086746-79086746 on Build GRCh38
rs146166989 Chr.2: 79086873-79086873 on Build GRCh38
rs112754434 Chr.2: 79086803-79086803 on Build GRCh38
rs201217603 Chr.2: 79086895-79086895 on Build GRCh38
rs775511086 Chr.2: 79086974-79086974 on Build GRCh38
rs774041730 Chr.2: 79086811-79086811 on Build GRCh38

View Full Product Details