Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 226735690-226736285 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IRS1
Entrez Gene ID
3667
Gene Name
insulin receptor substrate 1
Gene Aliases
HIRS-1
UniGene
Hs.471508
Gene Chromosome Location
Chr.2: 226731317-226798790 on Build GRCh38
Transcripts
NM_005544.2

SNP Detail

SNP ID Chromosome Location
rs15625 Chr.2: 226736165-226736165 on Build GRCh38
rs1029330323 Chr.2: 226735824-226735824 on Build GRCh38
rs772013181 Chr.2: 226736039-226736039 on Build GRCh38
rs1025677868 Chr.2: 226735909-226735909 on Build GRCh38
rs1005977909 Chr.2: 226736232-226736232 on Build GRCh38
rs935131551 Chr.2: 226736009-226736009 on Build GRCh38
rs893435426 Chr.2: 226736088-226736088 on Build GRCh38
rs572194788 Chr.2: 226735924-226735924 on Build GRCh38
rs537809110 Chr.2: 226735743-226735743 on Build GRCh38
rs903941708 Chr.2: 226736222-226736222 on Build GRCh38
rs1049343329 Chr.2: 226735757-226735757 on Build GRCh38
rs994303056 Chr.2: 226736183-226736183 on Build GRCh38
rs1042486900 Chr.2: 226736216-226736216 on Build GRCh38
rs113636756 Chr.2: 226736042-226736043 on Build GRCh38
rs796645133 Chr.2: 226736043-226736043 on Build GRCh38
rs896584471 Chr.2: 226735837-226735837 on Build GRCh38
rs553684007 Chr.2: 226735876-226735876 on Build GRCh38
rs774270348 Chr.2: 226735878-226735878 on Build GRCh38
rs966070999 Chr.2: 226735992-226735992 on Build GRCh38
rs893709585 Chr.2: 226735753-226735753 on Build GRCh38
rs1050334239 Chr.2: 226736136-226736136 on Build GRCh38
rs952198055 Chr.2: 226736074-226736074 on Build GRCh38
rs999954579 Chr.2: 226735884-226735884 on Build GRCh38
rs144208373 Chr.2: 226735802-226735802 on Build GRCh38
rs1023867865 Chr.2: 226736258-226736258 on Build GRCh38
rs75038209 Chr.2: 226735945-226735945 on Build GRCh38
rs1000026810 Chr.2: 226736262-226736262 on Build GRCh38
rs953294084 Chr.2: 226735942-226735942 on Build GRCh38
rs901257699 Chr.2: 226736172-226736172 on Build GRCh38
rs948171811 Chr.2: 226735991-226735991 on Build GRCh38
rs1018762369 Chr.2: 226736160-226736160 on Build GRCh38
rs1009639811 Chr.2: 226735872-226735872 on Build GRCh38
rs550454148 Chr.2: 226735967-226735967 on Build GRCh38
rs982292544 Chr.2: 226735993-226735993 on Build GRCh38
rs756998827 Chr.2: 226736021-226736023 on Build GRCh38
rs1019634127 Chr.2: 226735857-226735857 on Build GRCh38
rs114279438 Chr.2: 226735929-226735929 on Build GRCh38
rs889050881 Chr.2: 226736152-226736152 on Build GRCh38
rs953073099 Chr.2: 226735920-226735920 on Build GRCh38
rs927677878 Chr.2: 226736007-226736007 on Build GRCh38
rs140076732 Chr.2: 226736211-226736211 on Build GRCh38
rs79783536 Chr.2: 226736155-226736155 on Build GRCh38
rs930773793 Chr.2: 226735747-226735747 on Build GRCh38
rs568116373 Chr.2: 226735879-226735879 on Build GRCh38
rs938223414 Chr.2: 226736200-226736200 on Build GRCh38
rs1056895547 Chr.2: 226736201-226736201 on Build GRCh38
rs1052128155 Chr.2: 226735728-226735728 on Build GRCh38
rs192725964 Chr.2: 226735836-226735836 on Build GRCh38
rs916840619 Chr.2: 226735981-226735981 on Build GRCh38
rs917936269 Chr.2: 226736209-226736209 on Build GRCh38
rs778740347 Chr.2: 226736117-226736117 on Build GRCh38
rs918961316 Chr.2: 226736073-226736073 on Build GRCh38
rs146074484 Chr.2: 226735831-226735831 on Build GRCh38
rs1018600866 Chr.2: 226735953-226735953 on Build GRCh38
rs971856913 Chr.2: 226736066-226736066 on Build GRCh38

View Full Product Details