Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 60286241-60286811 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPM1A
Entrez Gene ID
5494
Gene Name
protein phosphatase, Mg2+/Mn2+ dependent 1A
Gene Aliases
PP2C-ALPHA, PP2CA, PP2Calpha
UniGene
Hs.130036
Gene Chromosome Location
Chr.14: 60245752-60299087 on Build GRCh38
Transcripts
NM_021003.4 NM_177951.2 NM_177952.2 XM_005267779.1 XM_005267781.1 XM_011536872.1 XM_011536876.1 XM_011536877.1 XM_011536878.1 XM_011536879.2 XM_011536880.2 XM_011536881.2 XM_011536882.2 XM_011536883.1 XM_011536884.1 XM_017021381.1 XM_017021382.1 XM_017021383.1 XM_017021384.1 XM_017021385.1 XM_017021386.1 XM_017021387.1

SNP Detail

SNP ID Chromosome Location
rs898592078 Chr.14: 60286293-60286293 on Build GRCh38
rs1008166401 Chr.14: 60286362-60286362 on Build GRCh38
rs955810151 Chr.14: 60286323-60286323 on Build GRCh38
rs935120139 Chr.14: 60286482-60286482 on Build GRCh38
rs964290934 Chr.14: 60286736-60286736 on Build GRCh38
rs1054419249 Chr.14: 60286567-60286567 on Build GRCh38
rs937590580 Chr.14: 60286565-60286565 on Build GRCh38
rs946130510 Chr.14: 60286580-60286580 on Build GRCh38
rs908052321 Chr.14: 60286537-60286537 on Build GRCh38
rs964140901 Chr.14: 60286428-60286428 on Build GRCh38
rs530378077 Chr.14: 60286272-60286272 on Build GRCh38
rs368804082 Chr.14: 60286531-60286533 on Build GRCh38
rs1018176233 Chr.14: 60286368-60286368 on Build GRCh38
rs138571709 Chr.14: 60286592-60286592 on Build GRCh38
rs1008086219 Chr.14: 60286613-60286613 on Build GRCh38
rs1026056387 Chr.14: 60286649-60286649 on Build GRCh38
rs559073517 Chr.14: 60286730-60286730 on Build GRCh38
rs988210385 Chr.14: 60286520-60286520 on Build GRCh38
rs987046593 Chr.14: 60286329-60286329 on Build GRCh38
rs576498353 Chr.14: 60286625-60286625 on Build GRCh38
rs1018488683 Chr.14: 60286393-60286393 on Build GRCh38
rs1010312388 Chr.14: 60286288-60286288 on Build GRCh38
rs951242002 Chr.14: 60286527-60286527 on Build GRCh38
rs187936312 Chr.14: 60286548-60286548 on Build GRCh38
rs140966037 Chr.14: 60286442-60286442 on Build GRCh38
rs917662468 Chr.14: 60286781-60286781 on Build GRCh38
rs182864295 Chr.14: 60286303-60286303 on Build GRCh38
rs951054204 Chr.14: 60286706-60286706 on Build GRCh38
rs1008185435 Chr.14: 60286687-60286687 on Build GRCh38
rs927464771 Chr.14: 60286789-60286789 on Build GRCh38
rs994301487 Chr.14: 60286610-60286610 on Build GRCh38
rs1017008950 Chr.14: 60286280-60286280 on Build GRCh38
rs925020364 Chr.14: 60286450-60286450 on Build GRCh38
rs771086827 Chr.14: 60286401-60286401 on Build GRCh38
rs193142710 Chr.14: 60286584-60286584 on Build GRCh38
rs973759448 Chr.14: 60286435-60286435 on Build GRCh38
rs536503764 Chr.14: 60286586-60286586 on Build GRCh38
rs1032551340 Chr.14: 60286758-60286758 on Build GRCh38
rs759481205 Chr.14: 60286552-60286552 on Build GRCh38
rs891093910 Chr.14: 60286650-60286650 on Build GRCh38
rs763626647 Chr.14: 60286641-60286641 on Build GRCh38
rs1027067645 Chr.14: 60286681-60286681 on Build GRCh38
rs551043682 Chr.14: 60286531-60286531 on Build GRCh38
rs141362294 Chr.14: 60286776-60286776 on Build GRCh38
rs553343837 Chr.14: 60286589-60286589 on Build GRCh38
rs775136954 Chr.14: 60286581-60286581 on Build GRCh38
rs1026650896 Chr.14: 60286471-60286471 on Build GRCh38
rs917627085 Chr.14: 60286522-60286522 on Build GRCh38
rs760535053 Chr.14: 60286311-60286311 on Build GRCh38
rs893249720 Chr.14: 60286279-60286279 on Build GRCh38
rs979551555 Chr.14: 60286742-60286742 on Build GRCh38
rs905131378 Chr.14: 60286543-60286543 on Build GRCh38
rs1056585005 Chr.14: 60286444-60286444 on Build GRCh38
rs145848692 Chr.14: 60286529-60286531 on Build GRCh38
rs571035526 Chr.14: 60286539-60286539 on Build GRCh38
rs537488440 Chr.14: 60286478-60286478 on Build GRCh38
rs150223114 Chr.14: 60286570-60286570 on Build GRCh38
rs1024751324 Chr.14: 60286746-60286746 on Build GRCh38
rs956652352 Chr.14: 60286770-60286770 on Build GRCh38
rs982958918 Chr.14: 60286714-60286714 on Build GRCh38
rs545454166 Chr.14: 60286720-60286720 on Build GRCh38

View Full Product Details