Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 77143332-77143925 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SEC14L1
Entrez Gene ID
6397
Gene Name
SEC14 like lipid binding 1
Gene Aliases
PRELID4A, SEC14L
UniGene
Hs.464184
Gene Chromosome Location
Chr.17: 77088643-77217101 on Build GRCh38
Transcripts
NM_001039573.2 NM_001143998.1 NM_001143999.1 NM_001204408.1 NM_001204410.1 NM_003003.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
707083 c.4G>T p.V2L Chr.17: 77143600-77143600 on Build GRCh38
3522648 c.18G>A p.Q6Q Chr.17: 77143614-77143614 on Build GRCh38
3522647 c.18G>A p.Q6Q Chr.17: 77143614-77143614 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs371430429 Chr.17: 77143600-77143600 on Build GRCh38
rs1022614311 Chr.17: 77143874-77143874 on Build GRCh38
rs766198434 Chr.17: 77143699-77143699 on Build GRCh38
rs766187120 Chr.17: 77143477-77143477 on Build GRCh38
rs780686653 Chr.17: 77143577-77143577 on Build GRCh38
rs758317180 Chr.17: 77143695-77143695 on Build GRCh38
rs974691456 Chr.17: 77143445-77143445 on Build GRCh38
rs772476558 Chr.17: 77143608-77143608 on Build GRCh38
rs1041900520 Chr.17: 77143704-77143704 on Build GRCh38
rs370628215 Chr.17: 77143690-77143690 on Build GRCh38
rs765182013 Chr.17: 77143551-77143551 on Build GRCh38
rs554312863 Chr.17: 77143535-77143535 on Build GRCh38
rs185972476 Chr.17: 77143531-77143531 on Build GRCh38
rs921205289 Chr.17: 77143747-77143747 on Build GRCh38
rs576354503 Chr.17: 77143705-77143705 on Build GRCh38
rs955106040 Chr.17: 77143408-77143408 on Build GRCh38
rs113520636 Chr.17: 77143572-77143572 on Build GRCh38
rs750379199 Chr.17: 77143552-77143552 on Build GRCh38
rs139865801 Chr.17: 77143474-77143474 on Build GRCh38
rs74814291 Chr.17: 77143661-77143661 on Build GRCh38
rs538775458 Chr.17: 77143493-77143493 on Build GRCh38
rs988818938 Chr.17: 77143726-77143726 on Build GRCh38
rs932555722 Chr.17: 77143753-77143753 on Build GRCh38
rs758271913 Chr.17: 77143553-77143553 on Build GRCh38
rs200599922 Chr.17: 77143679-77143679 on Build GRCh38
rs144668416 Chr.17: 77143603-77143603 on Build GRCh38
rs914210957 Chr.17: 77143897-77143897 on Build GRCh38
rs916086620 Chr.17: 77143450-77143450 on Build GRCh38
rs752931158 Chr.17: 77143507-77143507 on Build GRCh38
rs1049412 Chr.17: 77143617-77143617 on Build GRCh38
rs576273995 Chr.17: 77143738-77143738 on Build GRCh38
rs898239055 Chr.17: 77143798-77143798 on Build GRCh38
rs530382873 Chr.17: 77143398-77143398 on Build GRCh38
rs551979076 Chr.17: 77143500-77143500 on Build GRCh38
rs765514952 Chr.17: 77143692-77143694 on Build GRCh38
rs1044003057 Chr.17: 77143517-77143517 on Build GRCh38
rs962973406 Chr.17: 77143443-77143443 on Build GRCh38
rs762046920 Chr.17: 77143571-77143572 on Build GRCh38
rs773175515 Chr.17: 77143357-77143357 on Build GRCh38
rs963938248 Chr.17: 77143861-77143861 on Build GRCh38
rs995602742 Chr.17: 77143801-77143801 on Build GRCh38
rs62078293 Chr.17: 77143423-77143423 on Build GRCh38
rs928745017 Chr.17: 77143489-77143489 on Build GRCh38
rs899741977 Chr.17: 77143522-77143522 on Build GRCh38
rs199763176 Chr.17: 77143589-77143589 on Build GRCh38
rs757095903 Chr.17: 77143548-77143548 on Build GRCh38
rs558967833 Chr.17: 77143794-77143794 on Build GRCh38
rs911179788 Chr.17: 77143857-77143857 on Build GRCh38
rs376841241 Chr.17: 77143594-77143594 on Build GRCh38
rs945832928 Chr.17: 77143900-77143900 on Build GRCh38
rs377526158 Chr.17: 77143684-77143684 on Build GRCh38
rs199574629 Chr.17: 77143588-77143588 on Build GRCh38
rs750266660 Chr.17: 77143692-77143692 on Build GRCh38
rs746533863 Chr.17: 77143574-77143574 on Build GRCh38
rs536484603 Chr.17: 77143421-77143421 on Build GRCh38
rs757311034 Chr.17: 77143760-77143760 on Build GRCh38
rs970662851 Chr.17: 77143456-77143456 on Build GRCh38
rs377304866 Chr.17: 77143708-77143708 on Build GRCh38
rs894729349 Chr.17: 77143736-77143736 on Build GRCh38
rs538770012 Chr.17: 77143757-77143757 on Build GRCh38
rs890924678 Chr.17: 77143780-77143780 on Build GRCh38
rs1049411 Chr.17: 77143605-77143605 on Build GRCh38
rs765057500 Chr.17: 77143691-77143691 on Build GRCh38
rs1053063996 Chr.17: 77143498-77143498 on Build GRCh38
rs565398753 Chr.17: 77143518-77143518 on Build GRCh38
rs376718665 Chr.17: 77143576-77143576 on Build GRCh38
rs986642926 Chr.17: 77143856-77143857 on Build GRCh38
rs764133202 Chr.17: 77143732-77143732 on Build GRCh38
rs947409644 Chr.17: 77143743-77143743 on Build GRCh38
rs201648349 Chr.17: 77143620-77143620 on Build GRCh38
rs958657245 Chr.17: 77143403-77143403 on Build GRCh38
rs572545895 Chr.17: 77143788-77143788 on Build GRCh38
rs370610460 Chr.17: 77143717-77143717 on Build GRCh38
rs1003941204 Chr.17: 77143567-77143567 on Build GRCh38
rs572350196 Chr.17: 77143881-77143881 on Build GRCh38
rs779569334 Chr.17: 77143556-77143556 on Build GRCh38
rs189443849 Chr.17: 77143795-77143795 on Build GRCh38
rs981630168 Chr.17: 77143470-77143470 on Build GRCh38
rs754150469 Chr.17: 77143547-77143547 on Build GRCh38
rs545524121 Chr.17: 77143746-77143746 on Build GRCh38
rs760636719 Chr.17: 77143676-77143676 on Build GRCh38
rs754436385 Chr.17: 77143709-77143709 on Build GRCh38

View Full Product Details