Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 32042034-32042506 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RNF125
Entrez Gene ID
54941
Gene Name
ring finger protein 125
Gene Aliases
TNORS, TRAC-1, TRAC1
UniGene
Hs.633703
Gene Chromosome Location
Chr.18: 32018203-32090751 on Build GRCh38
Transcripts
NM_017831.3 XM_011526045.2 XM_011526046.2 XM_011526047.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
987711 c.413G>T p.R138M Chr.18: 32042273-32042273 on Build GRCh38
6378804 c.404C>A p.T135K Chr.18: 32042264-32042264 on Build GRCh38
987709 c.369A>G p.I123M Chr.18: 32042229-32042229 on Build GRCh38
987710 c.391G>T p.E131* Chr.18: 32042251-32042251 on Build GRCh38
4853589 c.336G>T p.M112I Chr.18: 32042196-32042196 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1035945215 Chr.18: 32042483-32042483 on Build GRCh38
rs547922916 Chr.18: 32042116-32042116 on Build GRCh38
rs930652565 Chr.18: 32042133-32042133 on Build GRCh38
rs916584376 Chr.18: 32042120-32042120 on Build GRCh38
rs368852977 Chr.18: 32042152-32042152 on Build GRCh38
rs371525573 Chr.18: 32042194-32042194 on Build GRCh38
rs1014406174 Chr.18: 32042437-32042437 on Build GRCh38
rs757375525 Chr.18: 32042242-32042242 on Build GRCh38
rs779881029 Chr.18: 32042244-32042244 on Build GRCh38
rs768503380 Chr.18: 32042317-32042317 on Build GRCh38
rs895989790 Chr.18: 32042434-32042434 on Build GRCh38
rs147178607 Chr.18: 32042184-32042184 on Build GRCh38
rs35880474 Chr.18: 32042426-32042426 on Build GRCh38
rs752863451 Chr.18: 32042227-32042227 on Build GRCh38
rs777017254 Chr.18: 32042320-32042320 on Build GRCh38
rs144268137 Chr.18: 32042247-32042247 on Build GRCh38
rs140351103 Chr.18: 32042205-32042205 on Build GRCh38
rs146291499 Chr.18: 32042228-32042228 on Build GRCh38
rs760207927 Chr.18: 32042311-32042311 on Build GRCh38
rs987354556 Chr.18: 32042351-32042351 on Build GRCh38
rs772751882 Chr.18: 32042302-32042302 on Build GRCh38
rs376483101 Chr.18: 32042299-32042299 on Build GRCh38
rs779716970 Chr.18: 32042147-32042150 on Build GRCh38
rs1055853724 Chr.18: 32042431-32042431 on Build GRCh38
rs1050723648 Chr.18: 32042348-32042348 on Build GRCh38
rs529992794 Chr.18: 32042089-32042089 on Build GRCh38
rs569488414 Chr.18: 32042358-32042358 on Build GRCh38
rs958879868 Chr.18: 32042109-32042109 on Build GRCh38
rs139642160 Chr.18: 32042246-32042246 on Build GRCh38
rs141623788 Chr.18: 32042058-32042058 on Build GRCh38
rs200871836 Chr.18: 32042173-32042173 on Build GRCh38
rs1011232107 Chr.18: 32042362-32042362 on Build GRCh38
rs946051965 Chr.18: 32042360-32042360 on Build GRCh38
rs557113849 Chr.18: 32042338-32042338 on Build GRCh38
rs747857110 Chr.18: 32042291-32042291 on Build GRCh38
rs376221651 Chr.18: 32042249-32042249 on Build GRCh38
rs774191338 Chr.18: 32042256-32042256 on Build GRCh38
rs765863296 Chr.18: 32042313-32042313 on Build GRCh38
rs35541487 Chr.18: 32042441-32042441 on Build GRCh38
rs1002138986 Chr.18: 32042081-32042081 on Build GRCh38
rs926054968 Chr.18: 32042417-32042417 on Build GRCh38
rs747059757 Chr.18: 32042235-32042235 on Build GRCh38
rs562805259 Chr.18: 32042061-32042061 on Build GRCh38
rs748966687 Chr.18: 32042165-32042167 on Build GRCh38
rs1002674785 Chr.18: 32042463-32042463 on Build GRCh38
rs778055153 Chr.18: 32042231-32042231 on Build GRCh38
rs786201014 Chr.18: 32042196-32042196 on Build GRCh38
rs761221706 Chr.18: 32042129-32042129 on Build GRCh38
rs191704906 Chr.18: 32042440-32042440 on Build GRCh38
rs536906631 Chr.18: 32042401-32042401 on Build GRCh38

View Full Product Details