Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 7619574-7620137 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
XAB2
Entrez Gene ID
56949
Gene Name
XPA binding protein 2
Gene Aliases
HCNP, HCRN, NTC90, SYF1
UniGene
Hs.9822
Gene Chromosome Location
Chr.19: 7619525-7629553 on Build GRCh38
Transcripts
NM_020196.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1751262 c.2485G>C p.E829Q Chr.19: 7619768-7619768 on Build GRCh38
6228709 c.2370C>T p.R790R Chr.19: 7619972-7619972 on Build GRCh38
1751261 c.2497G>A p.E833K Chr.19: 7619756-7619756 on Build GRCh38
6823652 c.2403C>T p.D801D Chr.19: 7619850-7619850 on Build GRCh38
6087131 c.2512C>A p.R838R Chr.19: 7619642-7619642 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763586732 Chr.19: 7619640-7619640 on Build GRCh38
rs745389101 Chr.19: 7619995-7619995 on Build GRCh38
rs758108479 Chr.19: 7619660-7619660 on Build GRCh38
rs747283014 Chr.19: 7619827-7619827 on Build GRCh38
rs144798612 Chr.19: 7619641-7619641 on Build GRCh38
rs750034998 Chr.19: 7619702-7619702 on Build GRCh38
rs770549378 Chr.19: 7619927-7619927 on Build GRCh38
rs368601786 Chr.19: 7619698-7619698 on Build GRCh38
rs769108296 Chr.19: 7619740-7619740 on Build GRCh38
rs767288318 Chr.19: 7619940-7619940 on Build GRCh38
rs763833507 Chr.19: 7619951-7619951 on Build GRCh38
rs568491893 Chr.19: 7619943-7619943 on Build GRCh38
rs12610460 Chr.19: 7619900-7619900 on Build GRCh38
rs901446128 Chr.19: 7619973-7619973 on Build GRCh38
rs778056102 Chr.19: 7619984-7619984 on Build GRCh38
rs772010020 Chr.19: 7619604-7619604 on Build GRCh38
rs768983446 Chr.19: 7619765-7619766 on Build GRCh38
rs764653754 Chr.19: 7619771-7619771 on Build GRCh38
rs868307678 Chr.19: 7620024-7620024 on Build GRCh38
rs756670916 Chr.19: 7619635-7619635 on Build GRCh38
rs574958927 Chr.19: 7619642-7619642 on Build GRCh38
rs78619637 Chr.19: 7619638-7619638 on Build GRCh38
rs779591205 Chr.19: 7620102-7620102 on Build GRCh38
rs552087975 Chr.19: 7619938-7619938 on Build GRCh38
rs771145632 Chr.19: 7619669-7619669 on Build GRCh38
rs758954156 Chr.19: 7620103-7620117 on Build GRCh38
rs949675411 Chr.19: 7620098-7620098 on Build GRCh38
rs199603958 Chr.19: 7619677-7619677 on Build GRCh38
rs753179460 Chr.19: 7619634-7619634 on Build GRCh38
rs372963419 Chr.19: 7620106-7620106 on Build GRCh38
rs80188016 Chr.19: 7619884-7619884 on Build GRCh38
rs373646562 Chr.19: 7619683-7619683 on Build GRCh38
rs966373526 Chr.19: 7619914-7619914 on Build GRCh38
rs780282787 Chr.19: 7619826-7619826 on Build GRCh38
rs764111910 Chr.19: 7619892-7619892 on Build GRCh38
rs765810383 Chr.19: 7619878-7619879 on Build GRCh38
rs375485938 Chr.19: 7619721-7619721 on Build GRCh38
rs371353910 Chr.19: 7619728-7619728 on Build GRCh38
rs751413778 Chr.19: 7619712-7619712 on Build GRCh38
rs374852049 Chr.19: 7619736-7619736 on Build GRCh38
rs759582521 Chr.19: 7620058-7620058 on Build GRCh38
rs753803302 Chr.19: 7619965-7619965 on Build GRCh38
rs201891244 Chr.19: 7619717-7619717 on Build GRCh38
rs773740151 Chr.19: 7619921-7619921 on Build GRCh38
rs745577983 Chr.19: 7619655-7619655 on Build GRCh38
rs756355799 Chr.19: 7620092-7620092 on Build GRCh38
rs752952115 Chr.19: 7620083-7620083 on Build GRCh38
rs781181034 Chr.19: 7619789-7619789 on Build GRCh38
rs1054242784 Chr.19: 7620109-7620109 on Build GRCh38
rs756142550 Chr.19: 7619802-7619802 on Build GRCh38
rs772962561 Chr.19: 7619956-7619956 on Build GRCh38
rs766525949 Chr.19: 7619618-7619618 on Build GRCh38
rs751362505 Chr.19: 7619786-7619786 on Build GRCh38
rs566731525 Chr.19: 7619905-7619905 on Build GRCh38
rs1036958441 Chr.19: 7620062-7620062 on Build GRCh38
rs769984282 Chr.19: 7619612-7619612 on Build GRCh38
rs766382249 Chr.19: 7619968-7619968 on Build GRCh38
rs369838336 Chr.19: 7619867-7619867 on Build GRCh38
rs373115604 Chr.19: 7619846-7619846 on Build GRCh38
rs539962673 Chr.19: 7619737-7619737 on Build GRCh38
rs754267310 Chr.19: 7620094-7620094 on Build GRCh38
rs561263665 Chr.19: 7619808-7619808 on Build GRCh38
rs762455723 Chr.19: 7619776-7619776 on Build GRCh38
rs573463512 Chr.19: 7619722-7619722 on Build GRCh38
rs114216182 Chr.19: 7620110-7620110 on Build GRCh38
rs147137635 Chr.19: 7619966-7619966 on Build GRCh38
rs148416638 Chr.19: 7619748-7619748 on Build GRCh38
rs138952301 Chr.19: 7619598-7619598 on Build GRCh38
rs771586890 Chr.19: 7620107-7620107 on Build GRCh38
rs972726121 Chr.19: 7620073-7620073 on Build GRCh38
rs770161674 Chr.19: 7619920-7619920 on Build GRCh38
rs370950233 Chr.19: 7620115-7620115 on Build GRCh38
rs368839409 Chr.19: 7619727-7619727 on Build GRCh38
rs762556810 Chr.19: 7619862-7619862 on Build GRCh38
rs767415248 Chr.19: 7619872-7619872 on Build GRCh38
rs776418121 Chr.19: 7620028-7620028 on Build GRCh38
rs896756281 Chr.19: 7619847-7619847 on Build GRCh38
rs755201256 Chr.19: 7619909-7619909 on Build GRCh38
rs774857932 Chr.19: 7619766-7619777 on Build GRCh38
rs758139693 Chr.19: 7619704-7619704 on Build GRCh38
rs201993769 Chr.19: 7619805-7619805 on Build GRCh38
rs761048086 Chr.19: 7619739-7619739 on Build GRCh38
rs747229954 Chr.19: 7619908-7619908 on Build GRCh38
rs775210977 Chr.19: 7619989-7619989 on Build GRCh38
rs921264732 Chr.19: 7619650-7619650 on Build GRCh38
rs202171193 Chr.19: 7619893-7619893 on Build GRCh38
rs775896628 Chr.19: 7619684-7619684 on Build GRCh38
rs777941990 Chr.19: 7619667-7619667 on Build GRCh38
rs761683993 Chr.19: 7620040-7620040 on Build GRCh38
rs774729857 Chr.19: 7619624-7619624 on Build GRCh38
rs752563124 Chr.19: 7619793-7619793 on Build GRCh38
rs190323188 Chr.19: 7619661-7619661 on Build GRCh38
rs762263051 Chr.19: 7619783-7619788 on Build GRCh38
rs762306239 Chr.19: 7619994-7619994 on Build GRCh38
rs887945394 Chr.19: 7619783-7619783 on Build GRCh38
rs749861730 Chr.19: 7619926-7619926 on Build GRCh38
rs768997779 Chr.19: 7619829-7619829 on Build GRCh38
rs538580383 Chr.19: 7619615-7619615 on Build GRCh38
rs932077127 Chr.19: 7619780-7619780 on Build GRCh38
rs368694532 Chr.19: 7620003-7620003 on Build GRCh38
rs940853669 Chr.19: 7620059-7620059 on Build GRCh38
rs370115615 Chr.19: 7619910-7619910 on Build GRCh38
rs779929039 Chr.19: 7619708-7619708 on Build GRCh38
rs570282562 Chr.19: 7619766-7619771 on Build GRCh38
rs760291429 Chr.19: 7620007-7620007 on Build GRCh38
rs775262544 Chr.19: 7619607-7619607 on Build GRCh38
rs746996282 Chr.19: 7619610-7619610 on Build GRCh38
rs781509959 Chr.19: 7619976-7619976 on Build GRCh38
rs762444031 Chr.19: 7619769-7619769 on Build GRCh38
rs909331517 Chr.19: 7620056-7620056 on Build GRCh38
rs759847926 Chr.19: 7619625-7619625 on Build GRCh38
rs181514316 Chr.19: 7620002-7620002 on Build GRCh38
rs1009811231 Chr.19: 7619945-7619945 on Build GRCh38
rs758439039 Chr.19: 7619901-7619901 on Build GRCh38
rs757785157 Chr.19: 7620101-7620101 on Build GRCh38
rs373447776 Chr.19: 7619911-7619911 on Build GRCh38
rs779500183 Chr.19: 7620108-7620108 on Build GRCh38
rs369175265 Chr.19: 7619682-7619682 on Build GRCh38
rs778508844 Chr.19: 7619993-7619993 on Build GRCh38
rs61735949 Chr.19: 7619850-7619850 on Build GRCh38
rs867882181 Chr.19: 7619843-7619843 on Build GRCh38
rs957233785 Chr.19: 7619720-7619720 on Build GRCh38
rs768495624 Chr.19: 7620026-7620026 on Build GRCh38
rs772548305 Chr.19: 7619860-7619860 on Build GRCh38
rs140344688 Chr.19: 7619974-7619974 on Build GRCh38
rs756481711 Chr.19: 7619981-7619981 on Build GRCh38
rs4134868 Chr.19: 7620082-7620082 on Build GRCh38
rs377515209 Chr.19: 7620065-7620065 on Build GRCh38
rs4134869 Chr.19: 7619616-7619616 on Build GRCh38
rs766028536 Chr.19: 7619864-7619864 on Build GRCh38
rs774123099 Chr.19: 7619865-7619865 on Build GRCh38
rs768719502 Chr.19: 7619663-7619663 on Build GRCh38
rs754790580 Chr.19: 7619787-7619787 on Build GRCh38
rs867673212 Chr.19: 7620018-7620018 on Build GRCh38
rs749403153 Chr.19: 7619668-7619668 on Build GRCh38
rs774070049 Chr.19: 7619935-7619935 on Build GRCh38
rs752904390 Chr.19: 7619979-7619979 on Build GRCh38
rs777104578 Chr.19: 7619768-7619768 on Build GRCh38
rs776148604 Chr.19: 7620091-7620092 on Build GRCh38
rs745659658 Chr.19: 7619602-7619602 on Build GRCh38
rs760547666 Chr.19: 7619950-7619950 on Build GRCh38
rs753293635 Chr.19: 7619692-7619692 on Build GRCh38
rs748540819 Chr.19: 7619849-7619849 on Build GRCh38
rs115862729 Chr.19: 7619691-7619691 on Build GRCh38
rs755942438 Chr.19: 7619890-7619890 on Build GRCh38
rs568074860 Chr.19: 7620093-7620093 on Build GRCh38
rs751598908 Chr.19: 7620046-7620046 on Build GRCh38
rs767962259 Chr.19: 7619631-7619631 on Build GRCh38
rs761099289 Chr.19: 7619688-7619688 on Build GRCh38
rs373327376 Chr.19: 7620041-7620041 on Build GRCh38
rs376467012 Chr.19: 7619971-7619971 on Build GRCh38
rs200577717 Chr.19: 7619747-7619747 on Build GRCh38
rs761200519 Chr.19: 7619697-7619697 on Build GRCh38

View Full Product Details