Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 16395233-16395573 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPS15L1
Entrez Gene ID
58513
Gene Name
epidermal growth factor receptor pathway substrate 15 like 1
Gene Aliases
EPS15R
UniGene
Hs.654639
Gene Chromosome Location
Chr.19: 16355244-16472012 on Build GRCh38
Transcripts
NM_001258374.1 NM_001258375.1 NM_021235.2 NR_047665.1 NR_047666.1 XM_017027086.1 XM_017027087.1 XM_017027088.1 XM_017027089.1 XM_017027090.1 XM_017027091.1 XM_017027092.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5940848 c.1907C>T p.P636L Chr.19: 16395352-16395352 on Build GRCh38
7316503 c.1863G>C p.Q621H Chr.19: 16395396-16395396 on Build GRCh38
6655539 c.1836G>A p.T612T Chr.19: 16395423-16395423 on Build GRCh38
7316507 c.1840G>C p.E614Q Chr.19: 16395419-16395419 on Build GRCh38
6441017 c.1802T>A p.F601Y Chr.19: 16395457-16395457 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs540809329 Chr.19: 16395269-16395269 on Build GRCh38
rs761222396 Chr.19: 16395386-16395386 on Build GRCh38
rs754223152 Chr.19: 16395409-16395409 on Build GRCh38
rs761916250 Chr.19: 16395332-16395332 on Build GRCh38
rs920955338 Chr.19: 16395527-16395527 on Build GRCh38
rs772327375 Chr.19: 16395495-16395495 on Build GRCh38
rs151197144 Chr.19: 16395501-16395501 on Build GRCh38
rs749541309 Chr.19: 16395309-16395309 on Build GRCh38
rs768763664 Chr.19: 16395484-16395484 on Build GRCh38
rs1053049489 Chr.19: 16395433-16395433 on Build GRCh38
rs762568075 Chr.19: 16395413-16395413 on Build GRCh38
rs749055249 Chr.19: 16395470-16395470 on Build GRCh38
rs761149334 Chr.19: 16395506-16395506 on Build GRCh38
rs199882870 Chr.19: 16395403-16395403 on Build GRCh38
rs148227808 Chr.19: 16395399-16395399 on Build GRCh38
rs375957764 Chr.19: 16395292-16395292 on Build GRCh38
rs768910666 Chr.19: 16395325-16395325 on Build GRCh38
rs995048100 Chr.19: 16395257-16395257 on Build GRCh38
rs376008025 Chr.19: 16395512-16395512 on Build GRCh38
rs772736647 Chr.19: 16395352-16395352 on Build GRCh38
rs773806626 Chr.19: 16395379-16395379 on Build GRCh38
rs766689784 Chr.19: 16395407-16395407 on Build GRCh38
rs767464218 Chr.19: 16395301-16395302 on Build GRCh38
rs746436841 Chr.19: 16395419-16395419 on Build GRCh38
rs756074772 Chr.19: 16395452-16395452 on Build GRCh38
rs775324986 Chr.19: 16395306-16395306 on Build GRCh38
rs572312407 Chr.19: 16395273-16395273 on Build GRCh38
rs142506606 Chr.19: 16395423-16395423 on Build GRCh38
rs185088248 Chr.19: 16395282-16395282 on Build GRCh38
rs761519990 Chr.19: 16395296-16395297 on Build GRCh38
rs145871172 Chr.19: 16395457-16395457 on Build GRCh38
rs369675832 Chr.19: 16395424-16395424 on Build GRCh38
rs748259491 Chr.19: 16395494-16395494 on Build GRCh38
rs1009365708 Chr.19: 16395416-16395416 on Build GRCh38
rs969850613 Chr.19: 16395542-16395542 on Build GRCh38
rs750367780 Chr.19: 16395446-16395446 on Build GRCh38
rs774587859 Chr.19: 16395330-16395330 on Build GRCh38
rs769530395 Chr.19: 16395293-16395293 on Build GRCh38
rs28365816 Chr.19: 16395323-16395323 on Build GRCh38
rs750225620 Chr.19: 16395300-16395301 on Build GRCh38
rs549788157 Chr.19: 16395528-16395528 on Build GRCh38
rs779236467 Chr.19: 16395490-16395490 on Build GRCh38
rs750191435 Chr.19: 16395353-16395353 on Build GRCh38

View Full Product Details