Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 101435557-101435905 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALDOB
Entrez Gene ID
229
Gene Name
aldolase, fructose-bisphosphate B
Gene Aliases
ALDB, ALDO2
UniGene
Hs.530274
Gene Chromosome Location
Chr.9: 101420560-101435780 on Build GRCh38
Transcripts
NM_000035.3

SNP Detail

SNP ID Chromosome Location
rs557676320 Chr.9: 101435828-101435828 on Build GRCh38
rs181639417 Chr.9: 101435708-101435708 on Build GRCh38
rs534912208 Chr.9: 101435841-101435841 on Build GRCh38
rs539960672 Chr.9: 101435854-101435854 on Build GRCh38
rs536633217 Chr.9: 101435692-101435692 on Build GRCh38
rs530656849 Chr.9: 101435678-101435678 on Build GRCh38
rs1015617264 Chr.9: 101435693-101435693 on Build GRCh38
rs146081970 Chr.9: 101435828-101435828 on Build GRCh38
rs954389853 Chr.9: 101435767-101435767 on Build GRCh38
rs944466059 Chr.9: 101435818-101435819 on Build GRCh38
rs986984236 Chr.9: 101435776-101435776 on Build GRCh38
rs369832832 Chr.9: 101435866-101435866 on Build GRCh38
rs927810478 Chr.9: 101435804-101435804 on Build GRCh38
rs1028124807 Chr.9: 101435724-101435724 on Build GRCh38
rs566990272 Chr.9: 101435737-101435737 on Build GRCh38
rs1009097912 Chr.9: 101435600-101435600 on Build GRCh38
rs987924201 Chr.9: 101435832-101435832 on Build GRCh38
rs1040561463 Chr.9: 101435647-101435647 on Build GRCh38
rs375538366 Chr.9: 101435714-101435714 on Build GRCh38
rs1004266799 Chr.9: 101435688-101435688 on Build GRCh38
rs960825029 Chr.9: 101435684-101435684 on Build GRCh38
rs1052103133 Chr.9: 101435819-101435819 on Build GRCh38
rs372250890 Chr.9: 101435878-101435878 on Build GRCh38
rs756565943 Chr.9: 101435628-101435628 on Build GRCh38
rs550635631 Chr.9: 101435879-101435879 on Build GRCh38
rs577731807 Chr.9: 101435838-101435838 on Build GRCh38
rs974344427 Chr.9: 101435718-101435718 on Build GRCh38
rs1002038217 Chr.9: 101435666-101435666 on Build GRCh38
rs969332801 Chr.9: 101435787-101435787 on Build GRCh38
rs548654883 Chr.9: 101435687-101435687 on Build GRCh38
rs375472535 Chr.9: 101435796-101435796 on Build GRCh38
rs777967761 Chr.9: 101435630-101435630 on Build GRCh38
rs913587412 Chr.9: 101435819-101435819 on Build GRCh38
rs145578385 Chr.9: 101435756-101435756 on Build GRCh38
rs484567 Chr.9: 101435716-101435716 on Build GRCh38
rs112276330 Chr.9: 101435848-101435848 on Build GRCh38
rs982006299 Chr.9: 101435788-101435788 on Build GRCh38
rs1022963332 Chr.9: 101435768-101435768 on Build GRCh38
rs993626496 Chr.9: 101435702-101435702 on Build GRCh38
rs538095918 Chr.9: 101435806-101435806 on Build GRCh38
rs905056320 Chr.9: 101435635-101435635 on Build GRCh38

View Full Product Details