Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 196812031-196812424 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PAK2
Entrez Gene ID
5062
Gene Name
p21 (RAC1) activated kinase 2
Gene Aliases
PAK65, PAKgamma
UniGene
Hs.518530
Gene Chromosome Location
Chr.3: 196739857-196832647 on Build GRCh38
Transcripts
NM_002577.4 XM_011512870.2 XM_017006501.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4116437 c.774-1G>A p.? Chr.3: 196812218-196812218 on Build GRCh38
4116439 c.793A>G p.T265A Chr.3: 196812238-196812238 on Build GRCh38
5422613 c.774-10delC p.? Chr.3: 196812209-196812209 on Build GRCh38
4166902 c.805G>A p.V269I Chr.3: 196812250-196812250 on Build GRCh38
5059621 c.822G>T p.E274D Chr.3: 196812267-196812267 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs767122683 Chr.3: 196812239-196812239 on Build GRCh38
rs765524408 Chr.3: 196812251-196812251 on Build GRCh38
rs747686520 Chr.3: 196812252-196812252 on Build GRCh38
rs77835098 Chr.3: 196812134-196812134 on Build GRCh38
rs1010844597 Chr.3: 196812197-196812201 on Build GRCh38
rs943001660 Chr.3: 196812298-196812298 on Build GRCh38
rs368926366 Chr.3: 196812127-196812127 on Build GRCh38
rs372719397 Chr.3: 196812184-196812184 on Build GRCh38
rs376064226 Chr.3: 196812064-196812064 on Build GRCh38
rs749182376 Chr.3: 196812200-196812200 on Build GRCh38
rs371672012 Chr.3: 196812280-196812280 on Build GRCh38
rs759017300 Chr.3: 196812229-196812229 on Build GRCh38
rs142522775 Chr.3: 196812385-196812385 on Build GRCh38
rs1024141742 Chr.3: 196812156-196812156 on Build GRCh38
rs752033810 Chr.3: 196812081-196812081 on Build GRCh38
rs772096525 Chr.3: 196812175-196812188 on Build GRCh38
rs751630016 Chr.3: 196812287-196812287 on Build GRCh38
rs200379218 Chr.3: 196812295-196812295 on Build GRCh38
rs75726292 Chr.3: 196812369-196812369 on Build GRCh38
rs1017924322 Chr.3: 196812057-196812057 on Build GRCh38
rs534355759 Chr.3: 196812389-196812389 on Build GRCh38
rs570601062 Chr.3: 196812392-196812392 on Build GRCh38
rs1044390492 Chr.3: 196812153-196812153 on Build GRCh38
rs904044406 Chr.3: 196812351-196812351 on Build GRCh38
rs745848227 Chr.3: 196812319-196812319 on Build GRCh38
rs1018486456 Chr.3: 196812113-196812113 on Build GRCh38
rs758937089 Chr.3: 196812272-196812272 on Build GRCh38
rs375918733 Chr.3: 196812202-196812202 on Build GRCh38
rs769666321 Chr.3: 196812188-196812188 on Build GRCh38
rs1031402457 Chr.3: 196812129-196812129 on Build GRCh38
rs754503513 Chr.3: 196812123-196812123 on Build GRCh38
rs193250457 Chr.3: 196812063-196812063 on Build GRCh38
rs1007026357 Chr.3: 196812109-196812109 on Build GRCh38
rs35267829 Chr.3: 196812075-196812075 on Build GRCh38
rs775082676 Chr.3: 196812249-196812249 on Build GRCh38
rs903929161 Chr.3: 196812119-196812119 on Build GRCh38
rs980340581 Chr.3: 196812174-196812174 on Build GRCh38
rs781609813 Chr.3: 196812175-196812175 on Build GRCh38
rs974358364 Chr.3: 196812098-196812098 on Build GRCh38
rs1051320412 Chr.3: 196812075-196812075 on Build GRCh38
rs992131508 Chr.3: 196812138-196812138 on Build GRCh38
rs35150956 Chr.3: 196812220-196812220 on Build GRCh38
rs758230476 Chr.3: 196812060-196812061 on Build GRCh38
rs773156274 Chr.3: 196812196-196812196 on Build GRCh38
rs367592484 Chr.3: 196812275-196812275 on Build GRCh38
rs112704024 Chr.3: 196812376-196812376 on Build GRCh38
rs867798971 Chr.3: 196812246-196812246 on Build GRCh38
rs113159880 Chr.3: 196812170-196812170 on Build GRCh38
rs199761020 Chr.3: 196812250-196812250 on Build GRCh38
rs992433107 Chr.3: 196812388-196812388 on Build GRCh38
rs1040263357 Chr.3: 196812350-196812350 on Build GRCh38
rs770691065 Chr.3: 196812373-196812373 on Build GRCh38
rs151104446 Chr.3: 196812391-196812391 on Build GRCh38
rs376715953 Chr.3: 196812291-196812291 on Build GRCh38
rs148394531 Chr.3: 196812102-196812102 on Build GRCh38
rs777708684 Chr.3: 196812301-196812301 on Build GRCh38
rs780110686 Chr.3: 196812171-196812171 on Build GRCh38
rs777612846 Chr.3: 196812313-196812313 on Build GRCh38
rs747122238 Chr.3: 196812172-196812172 on Build GRCh38
rs749096090 Chr.3: 196812311-196812311 on Build GRCh38
rs774193642 Chr.3: 196812317-196812317 on Build GRCh38
rs755209114 Chr.3: 196812173-196812173 on Build GRCh38

View Full Product Details