Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 152325908-152326469 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXW7
Entrez Gene ID
55294
Gene Name
F-box and WD repeat domain containing 7
Gene Aliases
AGO, CDC4, FBW6, FBW7, FBX30, FBXO30, FBXW6, SEL-10, SEL10, hAgo, hCdc4
UniGene
Hs.561245
Gene Chromosome Location
Chr.4: 152321258-152536095 on Build GRCh38
Transcripts
NM_001013415.1 NM_018315.4 NM_033632.3 XM_011532083.1 XM_011532084.1 XM_011532085.1 XM_011532086.1 XM_011532087.1 XM_011532088.1 XM_017008362.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6662544 c.1279T>C p.Y427H Chr.4: 152326017-152326017 on Build GRCh38
5490558 c.1598G>T p.C533F Chr.4: 152326052-152326052 on Build GRCh38
4448896 c.1179-2A>C p.? Chr.4: 152326233-152326233 on Build GRCh38
25812 c.1514G>A p.R505H Chr.4: 152326136-152326136 on Build GRCh38
108571 c.796C>T p.R266C Chr.4: 152326137-152326137 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs869059999 Chr.4: 152326079-152326079 on Build GRCh38
rs373440639 Chr.4: 152326409-152326409 on Build GRCh38
rs919458273 Chr.4: 152325965-152325965 on Build GRCh38
rs760308394 Chr.4: 152326274-152326274 on Build GRCh38
rs947038921 Chr.4: 152326374-152326374 on Build GRCh38
rs906636691 Chr.4: 152326290-152326290 on Build GRCh38
rs878863411 Chr.4: 152326333-152326333 on Build GRCh38
rs763590985 Chr.4: 152326024-152326024 on Build GRCh38
rs781115895 Chr.4: 152326238-152326238 on Build GRCh38
rs768872786 Chr.4: 152325967-152325967 on Build GRCh38
rs189542579 Chr.4: 152326286-152326286 on Build GRCh38
rs138251041 Chr.4: 152326448-152326448 on Build GRCh38
rs760764451 Chr.4: 152325943-152325943 on Build GRCh38
rs575517491 Chr.4: 152326280-152326280 on Build GRCh38
rs534491967 Chr.4: 152325957-152325957 on Build GRCh38
rs563418763 Chr.4: 152326438-152326438 on Build GRCh38
rs771865997 Chr.4: 152326259-152326259 on Build GRCh38
rs745947866 Chr.4: 152326250-152326251 on Build GRCh38
rs773219815 Chr.4: 152325972-152325972 on Build GRCh38
rs754980041 Chr.4: 152326260-152326260 on Build GRCh38
rs573615137 Chr.4: 152326308-152326308 on Build GRCh38
rs1002658878 Chr.4: 152326313-152326313 on Build GRCh38
rs532283658 Chr.4: 152326241-152326241 on Build GRCh38
rs141482952 Chr.4: 152325986-152325986 on Build GRCh38
rs370413277 Chr.4: 152326277-152326277 on Build GRCh38
rs757683191 Chr.4: 152326107-152326107 on Build GRCh38
rs1046234488 Chr.4: 152326285-152326285 on Build GRCh38
rs532749003 Chr.4: 152326331-152326331 on Build GRCh38
rs759277957 Chr.4: 152326252-152326252 on Build GRCh38
rs759412025 Chr.4: 152325980-152325980 on Build GRCh38
rs150453955 Chr.4: 152325993-152325993 on Build GRCh38
rs780805225 Chr.4: 152326278-152326278 on Build GRCh38
rs778982804 Chr.4: 152326238-152326238 on Build GRCh38
rs963662952 Chr.4: 152325933-152325933 on Build GRCh38
rs561543795 Chr.4: 152326300-152326300 on Build GRCh38
rs915795065 Chr.4: 152326239-152326239 on Build GRCh38
rs573298111 Chr.4: 152326424-152326424 on Build GRCh38
rs774376522 Chr.4: 152326249-152326249 on Build GRCh38
rs565526456 Chr.4: 152326325-152326325 on Build GRCh38
rs897303017 Chr.4: 152326446-152326446 on Build GRCh38
rs149680468 Chr.4: 152326137-152326137 on Build GRCh38
rs138346992 Chr.4: 152326222-152326222 on Build GRCh38
rs988537768 Chr.4: 152326352-152326352 on Build GRCh38
rs926609770 Chr.4: 152326377-152326377 on Build GRCh38
rs890085541 Chr.4: 152326395-152326395 on Build GRCh38
rs780458954 Chr.4: 152325955-152325955 on Build GRCh38
rs936611105 Chr.4: 152326147-152326147 on Build GRCh38
rs974028171 Chr.4: 152325962-152325962 on Build GRCh38
rs762792259 Chr.4: 152325975-152325975 on Build GRCh38
rs147462419 Chr.4: 152326126-152326126 on Build GRCh38
rs773991203 Chr.4: 152325976-152325976 on Build GRCh38
rs780367283 Chr.4: 152326210-152326210 on Build GRCh38
rs540820638 Chr.4: 152326319-152326319 on Build GRCh38
rs181926996 Chr.4: 152326441-152326441 on Build GRCh38
rs750717620 Chr.4: 152326166-152326166 on Build GRCh38
rs764317243 Chr.4: 152325991-152325991 on Build GRCh38
rs370070523 Chr.4: 152326000-152326000 on Build GRCh38
rs995137230 Chr.4: 152326411-152326411 on Build GRCh38
rs1049854441 Chr.4: 152326386-152326386 on Build GRCh38
rs914226987 Chr.4: 152326372-152326372 on Build GRCh38
rs866987936 Chr.4: 152326214-152326214 on Build GRCh38
rs117291353 Chr.4: 152325942-152325942 on Build GRCh38
rs770738515 Chr.4: 152326247-152326247 on Build GRCh38
rs367926135 Chr.4: 152326335-152326335 on Build GRCh38
rs1039390062 Chr.4: 152326042-152326042 on Build GRCh38
rs753448369 Chr.4: 152326069-152326069 on Build GRCh38
rs375905474 Chr.4: 152326292-152326292 on Build GRCh38
rs773696556 Chr.4: 152326367-152326367 on Build GRCh38
rs920889413 Chr.4: 152326099-152326099 on Build GRCh38
rs374570399 Chr.4: 152326204-152326204 on Build GRCh38
rs567211838 Chr.4: 152326449-152326449 on Build GRCh38

View Full Product Details