Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 119497805-119498395 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC645513
Entrez Gene ID
645513
Gene Name
septin 7 pseudogene
Gene Aliases
-
UniGene
Hs.677546
Gene Chromosome Location
Chr.4: 119454783-119499592 on Build GRCh38
Transcripts
NR_037630.1 NM_001083.3 NM_033430.2 NM_033437.3

Gene Symbol
PDE5A
Entrez Gene ID
8654
Gene Name
phosphodiesterase 5A
Gene Aliases
CGB-PDE, CN5A, PDE5
UniGene
Hs.647971
Gene Chromosome Location
Chr.4: 119494395-119629080 on Build GRCh38
Transcripts
NR_037630.1 NM_001083.3 NM_033430.2 NM_033437.3

SNP Detail

SNP ID Chromosome Location
rs570706806 Chr.4: 119498254-119498254 on Build GRCh38
rs941825363 Chr.4: 119498289-119498289 on Build GRCh38
rs201137671 Chr.4: 119498322-119498322 on Build GRCh38
rs958442909 Chr.4: 119498153-119498153 on Build GRCh38
rs1041754913 Chr.4: 119498147-119498147 on Build GRCh38
rs893158334 Chr.4: 119497947-119497947 on Build GRCh38
rs1041702611 Chr.4: 119497966-119497966 on Build GRCh38
rs201506441 Chr.4: 119498085-119498085 on Build GRCh38
rs1047158062 Chr.4: 119497957-119497957 on Build GRCh38
rs547116380 Chr.4: 119498073-119498073 on Build GRCh38
rs199901673 Chr.4: 119498178-119498178 on Build GRCh38
rs192864860 Chr.4: 119498349-119498349 on Build GRCh38
rs201859069 Chr.4: 119497842-119497842 on Build GRCh38
rs976696696 Chr.4: 119498192-119498192 on Build GRCh38
rs1007725949 Chr.4: 119498195-119498195 on Build GRCh38
rs116765299 Chr.4: 119498177-119498177 on Build GRCh38
rs542418563 Chr.4: 119498170-119498170 on Build GRCh38
rs200342258 Chr.4: 119497929-119497929 on Build GRCh38
rs201626303 Chr.4: 119497928-119497928 on Build GRCh38
rs200355871 Chr.4: 119498350-119498350 on Build GRCh38
rs1030792430 Chr.4: 119498150-119498150 on Build GRCh38
rs201284966 Chr.4: 119497969-119497969 on Build GRCh38
rs981196247 Chr.4: 119498139-119498139 on Build GRCh38
rs542955909 Chr.4: 119497948-119497948 on Build GRCh38
rs907012246 Chr.4: 119497917-119497917 on Build GRCh38
rs963610374 Chr.4: 119498231-119498231 on Build GRCh38
rs188114538 Chr.4: 119498274-119498274 on Build GRCh38
rs539941909 Chr.4: 119498260-119498260 on Build GRCh38
rs774109543 Chr.4: 119498339-119498342 on Build GRCh38
rs573272730 Chr.4: 119498309-119498309 on Build GRCh38
rs770634844 Chr.4: 119498263-119498263 on Build GRCh38
rs1025288212 Chr.4: 119498297-119498297 on Build GRCh38
rs895071749 Chr.4: 119498034-119498034 on Build GRCh38
rs199957318 Chr.4: 119498171-119498171 on Build GRCh38
rs200323703 Chr.4: 119498078-119498078 on Build GRCh38
rs201961968 Chr.4: 119498331-119498331 on Build GRCh38
rs1006087143 Chr.4: 119497974-119497974 on Build GRCh38
rs1030936905 Chr.4: 119498009-119498012 on Build GRCh38
rs937279125 Chr.4: 119497933-119497933 on Build GRCh38
rs973717620 Chr.4: 119498128-119498128 on Build GRCh38
rs200434730 Chr.4: 119498310-119498310 on Build GRCh38
rs78501811 Chr.4: 119498145-119498145 on Build GRCh38
rs930080230 Chr.4: 119497954-119497954 on Build GRCh38
rs200506777 Chr.4: 119498151-119498151 on Build GRCh38
rs1008253933 Chr.4: 119498067-119498071 on Build GRCh38
rs771715167 Chr.4: 119498066-119498066 on Build GRCh38
rs201368158 Chr.4: 119498046-119498046 on Build GRCh38
rs201321553 Chr.4: 119498155-119498155 on Build GRCh38
rs200689208 Chr.4: 119498116-119498116 on Build GRCh38
rs199597306 Chr.4: 119498060-119498060 on Build GRCh38
rs1012297859 Chr.4: 119498062-119498062 on Build GRCh38
rs368940206 Chr.4: 119498169-119498169 on Build GRCh38
rs531331321 Chr.4: 119498162-119498162 on Build GRCh38
rs886258947 Chr.4: 119497967-119497967 on Build GRCh38
rs1037984662 Chr.4: 119498029-119498029 on Build GRCh38
rs928484897 Chr.4: 119498140-119498140 on Build GRCh38
rs928400013 Chr.4: 119498330-119498330 on Build GRCh38
rs953554496 Chr.4: 119498043-119498043 on Build GRCh38
rs772731723 Chr.4: 119498025-119498029 on Build GRCh38
rs202221042 Chr.4: 119498288-119498288 on Build GRCh38
rs1037554019 Chr.4: 119498308-119498308 on Build GRCh38
rs202014536 Chr.4: 119498144-119498144 on Build GRCh38
rs17006236 Chr.4: 119497911-119497911 on Build GRCh38
rs918222651 Chr.4: 119498136-119498136 on Build GRCh38
rs922586967 Chr.4: 119498197-119498197 on Build GRCh38
rs1041029473 Chr.4: 119497838-119497838 on Build GRCh38
rs201443514 Chr.4: 119498346-119498346 on Build GRCh38
rs201437363 Chr.4: 119498357-119498357 on Build GRCh38
rs139582792 Chr.4: 119498174-119498174 on Build GRCh38
rs200352650 Chr.4: 119498016-119498016 on Build GRCh38
rs534535892 Chr.4: 119498329-119498329 on Build GRCh38
rs202153456 Chr.4: 119498001-119498001 on Build GRCh38
rs200163320 Chr.4: 119497858-119497858 on Build GRCh38
rs199520486 Chr.4: 119498113-119498113 on Build GRCh38
rs200267462 Chr.4: 119498337-119498337 on Build GRCh38
rs951152181 Chr.4: 119498138-119498138 on Build GRCh38
rs1056153971 Chr.4: 119498143-119498143 on Build GRCh38
rs937180517 Chr.4: 119498141-119498141 on Build GRCh38
rs951038625 Chr.4: 119498304-119498304 on Build GRCh38
rs562005772 Chr.4: 119498273-119498273 on Build GRCh38

View Full Product Details