Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 34757655-34758123 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SNRPC
Entrez Gene ID
6631
Gene Name
small nuclear ribonucleoprotein polypeptide C
Gene Aliases
U1C, Yhc1
UniGene
Hs.1063
Gene Chromosome Location
Chr.6: 34757094-34773857 on Build GRCh38
Transcripts
NM_003093.2 NR_029472.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7245451 c.51C>G p.L17L Chr.6: 34757891-34757891 on Build GRCh38
4602888 c.38_39insTCA p.C13_R14insH Chr.6: 34757878-34757879 on Build GRCh38
6260195 c.51+1G>T p.? Chr.6: 34757955-34757955 on Build GRCh38
4006481 c.41G>T p.R14L Chr.6: 34757881-34757881 on Build GRCh38
5602345 c.38_39insNCA p.C13>XH Chr.6: 34757878-34757879 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1046513181 Chr.6: 34757963-34757963 on Build GRCh38
rs748117940 Chr.6: 34758019-34758022 on Build GRCh38
rs995189877 Chr.6: 34757965-34757965 on Build GRCh38
rs932757893 Chr.6: 34758050-34758050 on Build GRCh38
rs779958575 Chr.6: 34757968-34757968 on Build GRCh38
rs569156785 Chr.6: 34757731-34757731 on Build GRCh38
rs746133661 Chr.6: 34757966-34757966 on Build GRCh38
rs995220847 Chr.6: 34757683-34757683 on Build GRCh38
rs1004052542 Chr.6: 34758019-34758019 on Build GRCh38
rs982731604 Chr.6: 34757743-34757744 on Build GRCh38
rs965303570 Chr.6: 34758019-34758019 on Build GRCh38
rs921236624 Chr.6: 34757789-34757789 on Build GRCh38
rs756189414 Chr.6: 34757967-34757967 on Build GRCh38
rs974201055 Chr.6: 34757709-34757709 on Build GRCh38
rs745893204 Chr.6: 34757824-34757824 on Build GRCh38
rs778855974 Chr.6: 34757805-34757805 on Build GRCh38
rs936801417 Chr.6: 34757802-34757802 on Build GRCh38
rs774847493 Chr.6: 34757989-34757989 on Build GRCh38
rs767093870 Chr.6: 34757856-34757856 on Build GRCh38
rs200596510 Chr.6: 34757933-34757933 on Build GRCh38
rs748509943 Chr.6: 34757993-34757993 on Build GRCh38
rs776050653 Chr.6: 34757834-34757834 on Build GRCh38
rs1002386868 Chr.6: 34757985-34757985 on Build GRCh38
rs921300715 Chr.6: 34758088-34758088 on Build GRCh38
rs1036661979 Chr.6: 34757906-34757906 on Build GRCh38
rs377232073 Chr.6: 34757868-34757868 on Build GRCh38
rs760222714 Chr.6: 34757872-34757872 on Build GRCh38
rs781440312 Chr.6: 34757781-34757781 on Build GRCh38
rs1027142981 Chr.6: 34758018-34758018 on Build GRCh38
rs961078176 Chr.6: 34757681-34757681 on Build GRCh38
rs28398813 Chr.6: 34757881-34757881 on Build GRCh38
rs990173290 Chr.6: 34758033-34758033 on Build GRCh38
rs1008903792 Chr.6: 34757883-34757883 on Build GRCh38
rs752482561 Chr.6: 34757899-34757899 on Build GRCh38
rs945539647 Chr.6: 34757924-34757924 on Build GRCh38
rs59247079 Chr.6: 34757885-34757886 on Build GRCh38
rs368412079 Chr.6: 34758011-34758011 on Build GRCh38
rs3835274 Chr.6: 34757882-34757883 on Build GRCh38
rs573933076 Chr.6: 34757836-34757836 on Build GRCh38
rs951130358 Chr.6: 34758018-34758019 on Build GRCh38
rs771225751 Chr.6: 34757801-34757801 on Build GRCh38
rs746856986 Chr.6: 34757811-34757812 on Build GRCh38
rs573157845 Chr.6: 34757771-34757771 on Build GRCh38
rs760281284 Chr.6: 34758000-34758000 on Build GRCh38
rs781577264 Chr.6: 34757804-34757804 on Build GRCh38
rs1026671949 Chr.6: 34757975-34757975 on Build GRCh38
rs140215092 Chr.6: 34757881-34757882 on Build GRCh38
rs777402221 Chr.6: 34757812-34757819 on Build GRCh38
rs551083384 Chr.6: 34757727-34757727 on Build GRCh38
rs771270586 Chr.6: 34757878-34757880 on Build GRCh38
rs758271628 Chr.6: 34757912-34757912 on Build GRCh38
rs373509304 Chr.6: 34757976-34757976 on Build GRCh38
rs7743724 Chr.6: 34757701-34757701 on Build GRCh38
rs67671088 Chr.6: 34757881-34757881 on Build GRCh38
rs201765428 Chr.6: 34757880-34757881 on Build GRCh38
rs766082590 Chr.6: 34758002-34758002 on Build GRCh38
rs376508953 Chr.6: 34757988-34757988 on Build GRCh38
rs369712906 Chr.6: 34757895-34757895 on Build GRCh38
rs34594913 Chr.6: 34757878-34757879 on Build GRCh38
rs557974962 Chr.6: 34757812-34757815 on Build GRCh38
rs573131948 Chr.6: 34757746-34757746 on Build GRCh38
rs555521054 Chr.6: 34757832-34757832 on Build GRCh38
rs778948563 Chr.6: 34757740-34757740 on Build GRCh38
rs778889271 Chr.6: 34757797-34757797 on Build GRCh38
rs914657555 Chr.6: 34758036-34758036 on Build GRCh38
rs55756037 Chr.6: 34757936-34757936 on Build GRCh38
rs956010552 Chr.6: 34758031-34758031 on Build GRCh38
rs975317067 Chr.6: 34758063-34758063 on Build GRCh38
rs779409913 Chr.6: 34758090-34758090 on Build GRCh38
rs183675137 Chr.6: 34758008-34758008 on Build GRCh38
rs753679137 Chr.6: 34757700-34757700 on Build GRCh38
rs772902253 Chr.6: 34757999-34757999 on Build GRCh38
rs914690154 Chr.6: 34757810-34757810 on Build GRCh38
rs762182916 Chr.6: 34757850-34757850 on Build GRCh38
rs924576997 Chr.6: 34758014-34758014 on Build GRCh38
rs774649630 Chr.6: 34757846-34757846 on Build GRCh38
rs917838147 Chr.6: 34757735-34757735 on Build GRCh38
rs748062559 Chr.6: 34757806-34757810 on Build GRCh38
rs187933010 Chr.6: 34757798-34757798 on Build GRCh38
rs951288448 Chr.6: 34757732-34757732 on Build GRCh38
rs138468260 Chr.6: 34757752-34757752 on Build GRCh38
rs772488243 Chr.6: 34757995-34757995 on Build GRCh38

View Full Product Details