Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 88145043-88145541 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNR1
Entrez Gene ID
1268
Gene Name
cannabinoid receptor 1
Gene Aliases
CANN6, CB-R, CB1, CB1A, CB1K5, CB1R, CNR
UniGene
Hs.75110
Gene Chromosome Location
Chr.6: 88139864-88167429 on Build GRCh38
Transcripts
NM_001160226.1 NM_001160258.1 NM_001160259.1 NM_016083.4 NM_033181.3 XM_006715330.3 XM_011535424.1 XM_011535425.1 XM_011535426.1 XM_011535428.1 XM_017010238.1 XM_017010239.1 XM_017010240.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5694338 c.75A>T p.S25S Chr.6: 88145200-88145200 on Build GRCh38
6631604 c.63G>A p.L21L Chr.6: 88145212-88145212 on Build GRCh38
4917048 c.125G>A p.G42E Chr.6: 88145150-88145150 on Build GRCh38
6352955 c.202C>T p.P68S Chr.6: 88145073-88145073 on Build GRCh38
4799731 c.94G>T p.D32Y Chr.6: 88145181-88145181 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs148575019 Chr.6: 88145208-88145208 on Build GRCh38
rs564750216 Chr.6: 88145445-88145445 on Build GRCh38
rs758511965 Chr.6: 88145324-88145324 on Build GRCh38
rs777302533 Chr.6: 88145458-88145458 on Build GRCh38
rs62417860 Chr.6: 88145296-88145296 on Build GRCh38
rs977582824 Chr.6: 88145215-88145215 on Build GRCh38
rs985815275 Chr.6: 88145364-88145364 on Build GRCh38
rs941629903 Chr.6: 88145203-88145203 on Build GRCh38
rs778904319 Chr.6: 88145209-88145209 on Build GRCh38
rs368465914 Chr.6: 88145257-88145257 on Build GRCh38
rs1049931908 Chr.6: 88145473-88145473 on Build GRCh38
rs918061237 Chr.6: 88145459-88145459 on Build GRCh38
rs752937809 Chr.6: 88145086-88145086 on Build GRCh38
rs967569058 Chr.6: 88145499-88145499 on Build GRCh38
rs974330731 Chr.6: 88145407-88145407 on Build GRCh38
rs142010122 Chr.6: 88145234-88145234 on Build GRCh38
rs751492689 Chr.6: 88145231-88145231 on Build GRCh38
rs774890934 Chr.6: 88145185-88145185 on Build GRCh38
rs144681727 Chr.6: 88145122-88145122 on Build GRCh38
rs370946660 Chr.6: 88145112-88145112 on Build GRCh38
rs768086951 Chr.6: 88145195-88145195 on Build GRCh38
rs143181991 Chr.6: 88145187-88145187 on Build GRCh38
rs112220724 Chr.6: 88145301-88145301 on Build GRCh38
rs752064133 Chr.6: 88145075-88145075 on Build GRCh38
rs541149512 Chr.6: 88145398-88145398 on Build GRCh38
rs762584169 Chr.6: 88145069-88145069 on Build GRCh38
rs925301113 Chr.6: 88145507-88145507 on Build GRCh38
rs200220585 Chr.6: 88145245-88145245 on Build GRCh38
rs780029930 Chr.6: 88145111-88145111 on Build GRCh38
rs766934700 Chr.6: 88145071-88145071 on Build GRCh38
rs537736163 Chr.6: 88145329-88145329 on Build GRCh38
rs370158252 Chr.6: 88145269-88145269 on Build GRCh38
rs191458319 Chr.6: 88145090-88145090 on Build GRCh38
rs896564896 Chr.6: 88145242-88145242 on Build GRCh38
rs142910458 Chr.6: 88145417-88145417 on Build GRCh38
rs748988990 Chr.6: 88145264-88145264 on Build GRCh38
rs908453397 Chr.6: 88145332-88145332 on Build GRCh38
rs577468690 Chr.6: 88145391-88145391 on Build GRCh38
rs780977671 Chr.6: 88145163-88145163 on Build GRCh38
rs759938792 Chr.6: 88145083-88145083 on Build GRCh38
rs781222994 Chr.6: 88145235-88145235 on Build GRCh38
rs757346364 Chr.6: 88145100-88145100 on Build GRCh38
rs779451294 Chr.6: 88145311-88145311 on Build GRCh38
rs753845620 Chr.6: 88145099-88145099 on Build GRCh38
rs183543261 Chr.6: 88145419-88145419 on Build GRCh38
rs1015901797 Chr.6: 88145386-88145386 on Build GRCh38
rs371432780 Chr.6: 88145266-88145266 on Build GRCh38
rs541841219 Chr.6: 88145464-88145464 on Build GRCh38
rs369660344 Chr.6: 88145312-88145312 on Build GRCh38
rs539677123 Chr.6: 88145136-88145136 on Build GRCh38
rs201496665 Chr.6: 88145174-88145174 on Build GRCh38
rs776807704 Chr.6: 88145294-88145294 on Build GRCh38
rs1056591518 Chr.6: 88145233-88145233 on Build GRCh38
rs893508135 Chr.6: 88145477-88145477 on Build GRCh38
rs747886813 Chr.6: 88145238-88145238 on Build GRCh38
rs138753523 Chr.6: 88145164-88145164 on Build GRCh38
rs767810773 Chr.6: 88145084-88145084 on Build GRCh38
rs748879032 Chr.6: 88145326-88145326 on Build GRCh38
rs747783434 Chr.6: 88145213-88145213 on Build GRCh38
rs747979286 Chr.6: 88145147-88145147 on Build GRCh38
rs376127620 Chr.6: 88145279-88145279 on Build GRCh38
rs773682661 Chr.6: 88145267-88145267 on Build GRCh38
rs985764394 Chr.6: 88145143-88145143 on Build GRCh38
rs75770301 Chr.6: 88145247-88145247 on Build GRCh38
rs775986845 Chr.6: 88145282-88145282 on Build GRCh38
rs563137056 Chr.6: 88145493-88145493 on Build GRCh38
rs777561684 Chr.6: 88145255-88145255 on Build GRCh38
rs756276407 Chr.6: 88145088-88145088 on Build GRCh38
rs1040858031 Chr.6: 88145373-88145373 on Build GRCh38
rs761029757 Chr.6: 88145201-88145201 on Build GRCh38
rs750430909 Chr.6: 88145321-88145321 on Build GRCh38
rs373139581 Chr.6: 88145251-88145251 on Build GRCh38
rs939845623 Chr.6: 88145367-88145367 on Build GRCh38
rs769532946 Chr.6: 88145149-88145149 on Build GRCh38
rs1031320460 Chr.6: 88145277-88145277 on Build GRCh38
rs1021828541 Chr.6: 88145176-88145176 on Build GRCh38
rs992743281 Chr.6: 88145441-88145441 on Build GRCh38
rs752959680 Chr.6: 88145198-88145198 on Build GRCh38
rs975888285 Chr.6: 88145066-88145066 on Build GRCh38
rs16880261 Chr.6: 88145494-88145494 on Build GRCh38
rs140053080 Chr.6: 88145331-88145331 on Build GRCh38
rs1011805931 Chr.6: 88145151-88145151 on Build GRCh38
rs764239536 Chr.6: 88145206-88145206 on Build GRCh38
rs761017586 Chr.6: 88145284-88145284 on Build GRCh38
rs936650395 Chr.6: 88145521-88145521 on Build GRCh38
rs770413069 Chr.6: 88145184-88145184 on Build GRCh38

View Full Product Details