Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 139174105-139174622 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HECA
Entrez Gene ID
51696
Gene Name
hdc homolog, cell cycle regulator
Gene Aliases
HDC, HDCL, HHDC, dJ225E12.1
UniGene
Hs.197644
Gene Chromosome Location
Chr.6: 139135112-139180809 on Build GRCh38
Transcripts
NM_016217.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4609192 c.1431C>A p.Y477* Chr.6: 139174503-139174503 on Build GRCh38
739896 c.1313-1G>T p.? Chr.6: 139174384-139174384 on Build GRCh38
1440834 c.1376G>A p.C459Y Chr.6: 139174448-139174448 on Build GRCh38
5484808 c.1382A>T p.K461M Chr.6: 139174454-139174454 on Build GRCh38
6678662 c.1435T>C p.Y479H Chr.6: 139174507-139174507 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376932158 Chr.6: 139174466-139174466 on Build GRCh38
rs569976017 Chr.6: 139174214-139174214 on Build GRCh38
rs756267872 Chr.6: 139174408-139174408 on Build GRCh38
rs1003586404 Chr.6: 139174217-139174217 on Build GRCh38
rs760108919 Chr.6: 139174577-139174577 on Build GRCh38
rs200895705 Chr.6: 139174467-139174467 on Build GRCh38
rs895029237 Chr.6: 139174253-139174253 on Build GRCh38
rs774012183 Chr.6: 139174587-139174587 on Build GRCh38
rs779688798 Chr.6: 139174482-139174482 on Build GRCh38
rs1056781316 Chr.6: 139174220-139174220 on Build GRCh38
rs753878485 Chr.6: 139174462-139174462 on Build GRCh38
rs1004702428 Chr.6: 139174136-139174136 on Build GRCh38
rs760993349 Chr.6: 139174583-139174583 on Build GRCh38
rs768154483 Chr.6: 139174509-139174509 on Build GRCh38
rs200861964 Chr.6: 139174574-139174574 on Build GRCh38
rs372964075 Chr.6: 139174594-139174594 on Build GRCh38
rs767769914 Chr.6: 139174401-139174401 on Build GRCh38
rs944420417 Chr.6: 139174601-139174601 on Build GRCh38
rs954290704 Chr.6: 139174595-139174595 on Build GRCh38
rs760227951 Chr.6: 139174350-139174350 on Build GRCh38
rs1033637653 Chr.6: 139174223-139174223 on Build GRCh38
rs144238240 Chr.6: 139174521-139174521 on Build GRCh38
rs373854251 Chr.6: 139174407-139174407 on Build GRCh38
rs376497421 Chr.6: 139174365-139174365 on Build GRCh38
rs368734916 Chr.6: 139174510-139174510 on Build GRCh38
rs368949928 Chr.6: 139174364-139174364 on Build GRCh38
rs373447611 Chr.6: 139174567-139174567 on Build GRCh38
rs989701101 Chr.6: 139174347-139174347 on Build GRCh38
rs764175724 Chr.6: 139174584-139174584 on Build GRCh38
rs1049714059 Chr.6: 139174130-139174130 on Build GRCh38
rs149553267 Chr.6: 139174498-139174498 on Build GRCh38
rs946776608 Chr.6: 139174149-139174149 on Build GRCh38
rs6570313 Chr.6: 139174189-139174189 on Build GRCh38
rs761348110 Chr.6: 139174540-139174540 on Build GRCh38
rs112676908 Chr.6: 139174189-139174189 on Build GRCh38
rs759914878 Chr.6: 139174397-139174397 on Build GRCh38
rs766942880 Chr.6: 139174382-139174382 on Build GRCh38
rs768258192 Chr.6: 139174354-139174354 on Build GRCh38
rs762452941 Chr.6: 139174184-139174184 on Build GRCh38
rs959616064 Chr.6: 139174260-139174260 on Build GRCh38
rs369215561 Chr.6: 139174377-139174377 on Build GRCh38
rs752973824 Chr.6: 139174582-139174582 on Build GRCh38
rs757212008 Chr.6: 139174464-139174464 on Build GRCh38
rs771909205 Chr.6: 139174477-139174477 on Build GRCh38
rs767974521 Chr.6: 139174580-139174580 on Build GRCh38
rs1042226057 Chr.6: 139174169-139174169 on Build GRCh38
rs777952667 Chr.6: 139174413-139174413 on Build GRCh38
rs534558406 Chr.6: 139174131-139174131 on Build GRCh38
rs771567213 Chr.6: 139174564-139174564 on Build GRCh38
rs754958026 Chr.6: 139174238-139174238 on Build GRCh38
rs1012340449 Chr.6: 139174265-139174265 on Build GRCh38

View Full Product Details