Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 2363929-2364497 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF3B
Entrez Gene ID
8662
Gene Name
eukaryotic translation initiation factor 3 subunit B
Gene Aliases
EIF3-ETA, EIF3-P110, EIF3-P116, EIF3S9, PRT1
UniGene
Hs.371001
Gene Chromosome Location
Chr.7: 2354540-2380745 on Build GRCh38
Transcripts
NM_001037283.1 NM_003751.3 XM_011515599.1 XM_011515600.1 XM_011515601.1 XM_017012752.1 XM_017012753.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3928981 c.1021C>T p.R341C Chr.7: 2364393-2364393 on Build GRCh38
7328691 c.1064G>A p.R355K Chr.7: 2364436-2364436 on Build GRCh38
4611210 c.1079delG p.G362fs*28 Chr.7: 2364451-2364451 on Build GRCh38
7143488 c.1011G>C p.E337D Chr.7: 2364383-2364383 on Build GRCh38
4878272 c.1093T>A p.F365I Chr.7: 2364465-2364465 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776107723 Chr.7: 2364415-2364415 on Build GRCh38
rs199859606 Chr.7: 2363997-2363997 on Build GRCh38
rs539631913 Chr.7: 2364316-2364316 on Build GRCh38
rs764010445 Chr.7: 2364450-2364451 on Build GRCh38
rs750621099 Chr.7: 2364448-2364448 on Build GRCh38
rs528160466 Chr.7: 2364218-2364218 on Build GRCh38
rs1039347132 Chr.7: 2364313-2364313 on Build GRCh38
rs764542868 Chr.7: 2364433-2364433 on Build GRCh38
rs778958122 Chr.7: 2364372-2364372 on Build GRCh38
rs994303662 Chr.7: 2364260-2364263 on Build GRCh38
rs532172839 Chr.7: 2364179-2364179 on Build GRCh38
rs188647183 Chr.7: 2364219-2364219 on Build GRCh38
rs757102227 Chr.7: 2364476-2364476 on Build GRCh38
rs542277582 Chr.7: 2364455-2364455 on Build GRCh38
rs1051513803 Chr.7: 2364091-2364091 on Build GRCh38
rs116940910 Chr.7: 2364276-2364276 on Build GRCh38
rs888515202 Chr.7: 2364278-2364278 on Build GRCh38
rs568018284 Chr.7: 2364255-2364255 on Build GRCh38
rs374521648 Chr.7: 2364390-2364390 on Build GRCh38
rs879177552 Chr.7: 2364260-2364260 on Build GRCh38
rs760782462 Chr.7: 2364451-2364451 on Build GRCh38
rs992142362 Chr.7: 2364220-2364220 on Build GRCh38
rs929354057 Chr.7: 2364253-2364253 on Build GRCh38
rs1056258171 Chr.7: 2364369-2364369 on Build GRCh38
rs757424003 Chr.7: 2364348-2364348 on Build GRCh38
rs138890182 Chr.7: 2364367-2364367 on Build GRCh38
rs751079442 Chr.7: 2364386-2364386 on Build GRCh38
rs897323032 Chr.7: 2364103-2364103 on Build GRCh38
rs916964823 Chr.7: 2364405-2364405 on Build GRCh38
rs113671104 Chr.7: 2364222-2364222 on Build GRCh38
rs190456894 Chr.7: 2364071-2364071 on Build GRCh38
rs374173451 Chr.7: 2364434-2364434 on Build GRCh38
rs563146184 Chr.7: 2364148-2364148 on Build GRCh38
rs777624565 Chr.7: 2364458-2364458 on Build GRCh38
rs952831132 Chr.7: 2364129-2364129 on Build GRCh38
rs1047245551 Chr.7: 2364229-2364229 on Build GRCh38
rs1035875253 Chr.7: 2364028-2364028 on Build GRCh38
rs374756115 Chr.7: 2364015-2364015 on Build GRCh38
rs766329360 Chr.7: 2364452-2364452 on Build GRCh38
rs763428861 Chr.7: 2364341-2364341 on Build GRCh38
rs763650060 Chr.7: 2364374-2364374 on Build GRCh38
rs994292264 Chr.7: 2364123-2364123 on Build GRCh38
rs565617616 Chr.7: 2364203-2364203 on Build GRCh38
rs1011065905 Chr.7: 2363984-2363984 on Build GRCh38
rs60498742 Chr.7: 2364078-2364078 on Build GRCh38
rs963688812 Chr.7: 2364228-2364228 on Build GRCh38
rs754839997 Chr.7: 2364259-2364260 on Build GRCh38
rs913101193 Chr.7: 2364161-2364161 on Build GRCh38
rs977564474 Chr.7: 2364019-2364019 on Build GRCh38
rs371264143 Chr.7: 2364425-2364425 on Build GRCh38
rs764337104 Chr.7: 2364343-2364343 on Build GRCh38
rs199507021 Chr.7: 2364346-2364346 on Build GRCh38
rs982369062 Chr.7: 2364440-2364440 on Build GRCh38
rs991641840 Chr.7: 2364403-2364403 on Build GRCh38
rs943278449 Chr.7: 2364143-2364143 on Build GRCh38
rs1027032321 Chr.7: 2364124-2364124 on Build GRCh38
rs145719280 Chr.7: 2364404-2364404 on Build GRCh38
rs745425816 Chr.7: 2364411-2364411 on Build GRCh38
rs949698582 Chr.7: 2364238-2364238 on Build GRCh38
rs1018625279 Chr.7: 2364135-2364135 on Build GRCh38
rs919271829 Chr.7: 2364246-2364246 on Build GRCh38
rs991756213 Chr.7: 2364060-2364060 on Build GRCh38
rs902238083 Chr.7: 2364166-2364166 on Build GRCh38
rs1010596546 Chr.7: 2364154-2364154 on Build GRCh38
rs375551545 Chr.7: 2364385-2364385 on Build GRCh38
rs60775658 Chr.7: 2364058-2364058 on Build GRCh38
rs929409789 Chr.7: 2364089-2364089 on Build GRCh38
rs1020748124 Chr.7: 2363989-2363989 on Build GRCh38
rs369812494 Chr.7: 2364333-2364333 on Build GRCh38
rs925669756 Chr.7: 2364216-2364216 on Build GRCh38
rs368514580 Chr.7: 2364393-2364393 on Build GRCh38
rs758073580 Chr.7: 2364333-2364333 on Build GRCh38
rs60538594 Chr.7: 2364186-2364186 on Build GRCh38
rs769410248 Chr.7: 2364412-2364412 on Build GRCh38
rs530040308 Chr.7: 2364155-2364155 on Build GRCh38
rs890290916 Chr.7: 2364130-2364130 on Build GRCh38
rs770639032 Chr.7: 2364400-2364400 on Build GRCh38
rs752702368 Chr.7: 2364453-2364453 on Build GRCh38
rs915892313 Chr.7: 2364067-2364067 on Build GRCh38
rs200316869 Chr.7: 2364366-2364366 on Build GRCh38
rs973352849 Chr.7: 2364235-2364235 on Build GRCh38
rs549102167 Chr.7: 2364227-2364227 on Build GRCh38
rs749496599 Chr.7: 2364047-2364047 on Build GRCh38
rs537741976 Chr.7: 2364261-2364261 on Build GRCh38
rs973882605 Chr.7: 2364077-2364077 on Build GRCh38
rs967523273 Chr.7: 2363990-2363990 on Build GRCh38
rs911707038 Chr.7: 2364265-2364265 on Build GRCh38
rs184095082 Chr.7: 2364147-2364147 on Build GRCh38
rs751464783 Chr.7: 2364470-2364470 on Build GRCh38
rs762029255 Chr.7: 2364441-2364441 on Build GRCh38
rs1006865037 Chr.7: 2364131-2364131 on Build GRCh38
rs946641447 Chr.7: 2364338-2364338 on Build GRCh38
rs571093235 Chr.7: 2364284-2364284 on Build GRCh38
rs573226159 Chr.7: 2364031-2364031 on Build GRCh38
rs767795115 Chr.7: 2364447-2364447 on Build GRCh38
rs767600494 Chr.7: 2364349-2364349 on Build GRCh38

View Full Product Details