Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 94927281-94927859 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NDUFAF6
Entrez Gene ID
137682
Gene Name
NADH:ubiquinone oxidoreductase complex assembly factor 6
Gene Aliases
C8orf38
UniGene
Hs.729144
Gene Chromosome Location
Chr.8: 94895271-95118496 on Build GRCh38
Transcripts
XM_011516835.2 XM_017013028.1 XM_017013030.1 XM_017013031.1 XM_017013032.1 XM_017013035.1 XM_017013036.1 XM_017013039.1 NM_001135733.1 NM_033285.3 XM_011517386.2

Gene Symbol
TP53INP1
Entrez Gene ID
94241
Gene Name
tumor protein p53 inducible nuclear protein 1
Gene Aliases
SIP, TP53DINP1, TP53INP1A, TP53INP1B, Teap, p53DINP1
UniGene
Hs.492261
Gene Chromosome Location
Chr.8: 94925972-94949387 on Build GRCh38
Transcripts
XM_011516835.2 XM_017013028.1 XM_017013030.1 XM_017013031.1 XM_017013032.1 XM_017013035.1 XM_017013036.1 XM_017013039.1 NM_001135733.1 NM_033285.3 XM_011517386.2

SNP Detail

SNP ID Chromosome Location
rs878920317 Chr.8: 94927734-94927734 on Build GRCh38
rs980159562 Chr.8: 94927751-94927751 on Build GRCh38
rs532954455 Chr.8: 94927674-94927678 on Build GRCh38
rs555773421 Chr.8: 94927420-94927420 on Build GRCh38
rs778123605 Chr.8: 94927739-94927739 on Build GRCh38
rs968339079 Chr.8: 94927591-94927591 on Build GRCh38
rs528567880 Chr.8: 94927511-94927511 on Build GRCh38
rs906110930 Chr.8: 94927472-94927472 on Build GRCh38
rs578111438 Chr.8: 94927649-94927649 on Build GRCh38
rs1043309929 Chr.8: 94927647-94927647 on Build GRCh38
rs983843426 Chr.8: 94927392-94927392 on Build GRCh38
rs74913254 Chr.8: 94927718-94927718 on Build GRCh38
rs755060312 Chr.8: 94927507-94927507 on Build GRCh38
rs28362543 Chr.8: 94927643-94927643 on Build GRCh38
rs558388904 Chr.8: 94927634-94927634 on Build GRCh38
rs1021717727 Chr.8: 94927641-94927641 on Build GRCh38
rs1053327902 Chr.8: 94927601-94927601 on Build GRCh38
rs1015841173 Chr.8: 94927379-94927379 on Build GRCh38
rs547122004 Chr.8: 94927359-94927359 on Build GRCh38
rs57500750 Chr.8: 94927783-94927783 on Build GRCh38
rs2945554 Chr.8: 94927366-94927366 on Build GRCh38
rs377132246 Chr.8: 94927709-94927709 on Build GRCh38
rs533212435 Chr.8: 94927806-94927806 on Build GRCh38
rs928763274 Chr.8: 94927568-94927568 on Build GRCh38
rs989316426 Chr.8: 94927789-94927789 on Build GRCh38
rs574456580 Chr.8: 94927733-94927733 on Build GRCh38
rs143709181 Chr.8: 94927653-94927653 on Build GRCh38
rs1008396223 Chr.8: 94927722-94927722 on Build GRCh38
rs572588633 Chr.8: 94927725-94927725 on Build GRCh38
rs938957839 Chr.8: 94927479-94927479 on Build GRCh38
rs778762715 Chr.8: 94927771-94927771 on Build GRCh38
rs143979042 Chr.8: 94927737-94927737 on Build GRCh38
rs541453663 Chr.8: 94927729-94927729 on Build GRCh38
rs569203891 Chr.8: 94927490-94927490 on Build GRCh38
rs1010700790 Chr.8: 94927630-94927630 on Build GRCh38
rs969947797 Chr.8: 94927744-94927744 on Build GRCh38
rs891959226 Chr.8: 94927606-94927606 on Build GRCh38
rs891794873 Chr.8: 94927557-94927557 on Build GRCh38
rs955288884 Chr.8: 94927336-94927336 on Build GRCh38
rs919812148 Chr.8: 94927453-94927453 on Build GRCh38
rs1044565505 Chr.8: 94927431-94927431 on Build GRCh38
rs938755646 Chr.8: 94927571-94927571 on Build GRCh38
rs76448027 Chr.8: 94927705-94927705 on Build GRCh38
rs980209509 Chr.8: 94927778-94927778 on Build GRCh38
rs559514295 Chr.8: 94927653-94927653 on Build GRCh38
rs564361386 Chr.8: 94927792-94927792 on Build GRCh38
rs191848451 Chr.8: 94927396-94927396 on Build GRCh38
rs947530925 Chr.8: 94927610-94927610 on Build GRCh38
rs765186754 Chr.8: 94927708-94927709 on Build GRCh38
rs942199598 Chr.8: 94927410-94927410 on Build GRCh38
rs996498613 Chr.8: 94927670-94927670 on Build GRCh38
rs947582141 Chr.8: 94927831-94927831 on Build GRCh38

View Full Product Details