Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 62515796-62516299 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AHNAK
Entrez Gene ID
79026
Gene Name
AHNAK nucleoprotein
Gene Aliases
AHNAKRS
UniGene
Hs.502756
Gene Chromosome Location
Chr.11: 62433542-62546860 on Build GRCh38
Transcripts
NM_001620.2 NM_024060.3 XM_005274240.1 XM_005274241.2 XM_005274245.1 XM_017018270.1 XM_017018271.1 XM_017018272.1 XM_017018273.1

SNP Detail

SNP ID Chromosome Location
rs1022371804 Chr.11: 62516108-62516108 on Build GRCh38
rs900378782 Chr.11: 62516263-62516263 on Build GRCh38
rs3179870 Chr.11: 62515903-62515903 on Build GRCh38
rs1031189320 Chr.11: 62516079-62516079 on Build GRCh38
rs916798031 Chr.11: 62516020-62516020 on Build GRCh38
rs879903359 Chr.11: 62516235-62516236 on Build GRCh38
rs527457147 Chr.11: 62515995-62515995 on Build GRCh38
rs1019331090 Chr.11: 62516262-62516262 on Build GRCh38
rs529730334 Chr.11: 62516083-62516083 on Build GRCh38
rs753552583 Chr.11: 62516031-62516031 on Build GRCh38
rs907548199 Chr.11: 62516085-62516085 on Build GRCh38
rs545974567 Chr.11: 62515840-62515840 on Build GRCh38
rs765042277 Chr.11: 62516047-62516047 on Build GRCh38
rs948531929 Chr.11: 62515862-62515862 on Build GRCh38
rs771305437 Chr.11: 62516264-62516264 on Build GRCh38
rs1047411646 Chr.11: 62516069-62516069 on Build GRCh38
rs986097754 Chr.11: 62516183-62516183 on Build GRCh38
rs1022805201 Chr.11: 62515950-62515950 on Build GRCh38
rs969961542 Chr.11: 62516021-62516021 on Build GRCh38
rs1011019407 Chr.11: 62516105-62516105 on Build GRCh38
rs750158447 Chr.11: 62515894-62515894 on Build GRCh38
rs930398649 Chr.11: 62516048-62516048 on Build GRCh38
rs953461924 Chr.11: 62516148-62516148 on Build GRCh38
rs999962490 Chr.11: 62516008-62516008 on Build GRCh38
rs1050059 Chr.11: 62515978-62515978 on Build GRCh38
rs558472931 Chr.11: 62515914-62515914 on Build GRCh38
rs1047364447 Chr.11: 62515874-62515874 on Build GRCh38
rs892660809 Chr.11: 62516099-62516099 on Build GRCh38
rs888183916 Chr.11: 62515942-62515942 on Build GRCh38
rs929728355 Chr.11: 62515918-62515918 on Build GRCh38
rs767292615 Chr.11: 62515868-62515868 on Build GRCh38
rs569529663 Chr.11: 62516260-62516260 on Build GRCh38
rs1048196154 Chr.11: 62515932-62515932 on Build GRCh38
rs761163559 Chr.11: 62515867-62515867 on Build GRCh38
rs1040892765 Chr.11: 62515901-62515901 on Build GRCh38
rs761352072 Chr.11: 62516151-62516151 on Build GRCh38
rs992402552 Chr.11: 62516018-62516018 on Build GRCh38
rs930322779 Chr.11: 62515873-62515873 on Build GRCh38
rs181802575 Chr.11: 62515943-62515943 on Build GRCh38
rs915310240 Chr.11: 62515824-62515824 on Build GRCh38
rs754963768 Chr.11: 62516043-62516043 on Build GRCh38
rs943914103 Chr.11: 62515876-62515876 on Build GRCh38
rs995990936 Chr.11: 62515948-62515948 on Build GRCh38
rs1018775670 Chr.11: 62515906-62515906 on Build GRCh38
rs1008752706 Chr.11: 62515900-62515900 on Build GRCh38
rs533785059 Chr.11: 62515919-62515919 on Build GRCh38
rs561140351 Chr.11: 62516056-62516056 on Build GRCh38
rs968137723 Chr.11: 62515953-62515953 on Build GRCh38
rs1045121357 Chr.11: 62515973-62515973 on Build GRCh38
rs1030989565 Chr.11: 62516013-62516013 on Build GRCh38
rs1040276721 Chr.11: 62516257-62516257 on Build GRCh38
rs113488999 Chr.11: 62516017-62516017 on Build GRCh38
rs3819120 Chr.11: 62516200-62516200 on Build GRCh38
rs578179028 Chr.11: 62516012-62516012 on Build GRCh38

View Full Product Details