Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 986677-987187 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP2A2
Entrez Gene ID
161
Gene Name
adaptor related protein complex 2 alpha 2 subunit
Gene Aliases
ADTAB, CLAPA2, HIP-9, HIP9, HYPJ
UniGene
Hs.19121
Gene Chromosome Location
Chr.11: 925809-1012245 on Build GRCh38
Transcripts
NM_001242837.1 NM_012305.3 XM_011519929.1 XM_011519930.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7427496 c.1065G>T p.L355L Chr.11: 986887-986887 on Build GRCh38
4908785 c.1122C>T p.N374N Chr.11: 986944-986944 on Build GRCh38
1605421 c.1046T>C p.L349P Chr.11: 986868-986868 on Build GRCh38
430125 c.1021G>A p.E341K Chr.11: 986843-986843 on Build GRCh38
933704 c.1124C>T p.A375V Chr.11: 986946-986946 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs577659804 Chr.11: 986759-986759 on Build GRCh38
rs546431226 Chr.11: 987084-987084 on Build GRCh38
rs747694186 Chr.11: 986994-986994 on Build GRCh38
rs528316140 Chr.11: 986884-986884 on Build GRCh38
rs914977618 Chr.11: 986717-986717 on Build GRCh38
rs370252249 Chr.11: 986835-986835 on Build GRCh38
rs987654036 Chr.11: 987038-987038 on Build GRCh38
rs768484784 Chr.11: 986883-986883 on Build GRCh38
rs536232392 Chr.11: 987096-987096 on Build GRCh38
rs1004061564 Chr.11: 987146-987146 on Build GRCh38
rs757133478 Chr.11: 986777-986777 on Build GRCh38
rs1004826284 Chr.11: 986917-986917 on Build GRCh38
rs771424101 Chr.11: 986863-986863 on Build GRCh38
rs755570936 Chr.11: 986802-986802 on Build GRCh38
rs964432869 Chr.11: 987054-987054 on Build GRCh38
rs914884687 Chr.11: 987048-987048 on Build GRCh38
rs776000978 Chr.11: 986783-986786 on Build GRCh38
rs751290624 Chr.11: 986768-986768 on Build GRCh38
rs761530670 Chr.11: 986923-986923 on Build GRCh38
rs934980532 Chr.11: 986709-986709 on Build GRCh38
rs529259457 Chr.11: 986708-986708 on Build GRCh38
rs138971712 Chr.11: 986735-986736 on Build GRCh38
rs374206027 Chr.11: 986876-986876 on Build GRCh38
rs1036333993 Chr.11: 987147-987147 on Build GRCh38
rs778802195 Chr.11: 986707-986707 on Build GRCh38
rs769366099 Chr.11: 986999-986999 on Build GRCh38
rs777218143 Chr.11: 986866-986866 on Build GRCh38
rs192570020 Chr.11: 987156-987156 on Build GRCh38
rs928190284 Chr.11: 987023-987023 on Build GRCh38
rs759619840 Chr.11: 986757-986757 on Build GRCh38
rs369507312 Chr.11: 986764-986764 on Build GRCh38
rs1051019348 Chr.11: 986851-986851 on Build GRCh38
rs763730669 Chr.11: 986819-986819 on Build GRCh38
rs766351471 Chr.11: 986932-986932 on Build GRCh38
rs778682143 Chr.11: 986840-986840 on Build GRCh38
rs933832684 Chr.11: 987094-987094 on Build GRCh38
rs754578586 Chr.11: 986935-986935 on Build GRCh38
rs1054560656 Chr.11: 987149-987149 on Build GRCh38
rs753270396 Chr.11: 986934-986934 on Build GRCh38
rs866971326 Chr.11: 986976-986976 on Build GRCh38
rs756014639 Chr.11: 986930-986930 on Build GRCh38
rs376964234 Chr.11: 986944-986944 on Build GRCh38
rs377141195 Chr.11: 986805-986805 on Build GRCh38
rs182744152 Chr.11: 986800-986800 on Build GRCh38
rs188213063 Chr.11: 987070-987070 on Build GRCh38
rs1052213658 Chr.11: 986716-986716 on Build GRCh38
rs551162253 Chr.11: 986955-986955 on Build GRCh38
rs1046118265 Chr.11: 987131-987131 on Build GRCh38
rs767320784 Chr.11: 986929-986929 on Build GRCh38
rs943204219 Chr.11: 986705-986705 on Build GRCh38
rs571066159 Chr.11: 986968-986968 on Build GRCh38
rs887007963 Chr.11: 987099-987099 on Build GRCh38
rs750366387 Chr.11: 986801-986801 on Build GRCh38
rs946428357 Chr.11: 986747-986747 on Build GRCh38
rs780608284 Chr.11: 986855-986855 on Build GRCh38
rs550619475 Chr.11: 987028-987028 on Build GRCh38
rs747326791 Chr.11: 986842-986842 on Build GRCh38
rs202104092 Chr.11: 986977-986977 on Build GRCh38
rs764420273 Chr.11: 986766-986766 on Build GRCh38
rs987826556 Chr.11: 987029-987029 on Build GRCh38
rs1038857272 Chr.11: 986706-986706 on Build GRCh38
rs1047767862 Chr.11: 986758-986758 on Build GRCh38
rs775808418 Chr.11: 986763-986763 on Build GRCh38
rs746278805 Chr.11: 986961-986961 on Build GRCh38
rs770406083 Chr.11: 986965-986965 on Build GRCh38
rs376429863 Chr.11: 986704-986704 on Build GRCh38
rs757650056 Chr.11: 986960-986960 on Build GRCh38
rs906057011 Chr.11: 986809-986809 on Build GRCh38
rs946466523 Chr.11: 987049-987049 on Build GRCh38
rs899134764 Chr.11: 987154-987154 on Build GRCh38
rs774701302 Chr.11: 986922-986922 on Build GRCh38
rs982809717 Chr.11: 987020-987020 on Build GRCh38
rs750964447 Chr.11: 986713-986713 on Build GRCh38
rs974573591 Chr.11: 987055-987055 on Build GRCh38

View Full Product Details