Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 183635696-183636193 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARPC5
Entrez Gene ID
10092
Gene Name
actin related protein 2/3 complex subunit 5
Gene Aliases
ARC16, dJ127C7.3, p16-Arc
UniGene
Hs.518609
Gene Chromosome Location
Chr.1: 183626193-183635941 on Build GRCh38
Transcripts
NM_001270439.1 NM_005717.3 NM_001297669.1 NM_001297670.1 NM_015149.4 XM_011509339.2 XM_011509341.1 XM_011509342.2

Gene Symbol
RGL1
Entrez Gene ID
23179
Gene Name
ral guanine nucleotide dissociation stimulator like 1
Gene Aliases
RGL
UniGene
Hs.497148
Gene Chromosome Location
Chr.1: 183636047-183928551 on Build GRCh38
Transcripts
NM_001270439.1 NM_005717.3 NM_001297669.1 NM_001297670.1 NM_015149.4 XM_011509339.2 XM_011509341.1 XM_011509342.2

SNP Detail

SNP ID Chromosome Location
rs551378575 Chr.1: 183635942-183635942 on Build GRCh38
rs930580335 Chr.1: 183635816-183635816 on Build GRCh38
rs914857311 Chr.1: 183635823-183635823 on Build GRCh38
rs974476280 Chr.1: 183636029-183636029 on Build GRCh38
rs886287328 Chr.1: 183635871-183635871 on Build GRCh38
rs1013990695 Chr.1: 183635872-183635872 on Build GRCh38
rs948828557 Chr.1: 183636163-183636163 on Build GRCh38
rs977432584 Chr.1: 183635860-183635860 on Build GRCh38
rs543319391 Chr.1: 183635763-183635763 on Build GRCh38
rs2767305 Chr.1: 183635770-183635770 on Build GRCh38
rs954361531 Chr.1: 183635765-183635765 on Build GRCh38
rs899120552 Chr.1: 183635975-183635975 on Build GRCh38
rs541224986 Chr.1: 183635849-183635849 on Build GRCh38
rs934041650 Chr.1: 183635720-183635720 on Build GRCh38
rs942676872 Chr.1: 183635773-183635773 on Build GRCh38
rs930528928 Chr.1: 183636058-183636058 on Build GRCh38
rs112313734 Chr.1: 183635900-183635900 on Build GRCh38
rs576310166 Chr.1: 183635756-183635756 on Build GRCh38
rs907817648 Chr.1: 183636061-183636061 on Build GRCh38
rs900238131 Chr.1: 183636063-183636063 on Build GRCh38
rs905608951 Chr.1: 183636080-183636080 on Build GRCh38
rs1025980418 Chr.1: 183635759-183635759 on Build GRCh38
rs112256505 Chr.1: 183635884-183635884 on Build GRCh38
rs1031856596 Chr.1: 183636098-183636098 on Build GRCh38
rs1047562392 Chr.1: 183635865-183635865 on Build GRCh38
rs993196716 Chr.1: 183635730-183635730 on Build GRCh38
rs1037531978 Chr.1: 183635960-183635960 on Build GRCh38
rs1047695313 Chr.1: 183636059-183636059 on Build GRCh38
rs1001296592 Chr.1: 183636083-183636083 on Build GRCh38
rs973444028 Chr.1: 183636164-183636164 on Build GRCh38
rs957814637 Chr.1: 183635909-183635909 on Build GRCh38
rs968936869 Chr.1: 183635850-183635850 on Build GRCh38
rs745922078 Chr.1: 183636062-183636062 on Build GRCh38
rs141408319 Chr.1: 183636136-183636137 on Build GRCh38
rs1021748219 Chr.1: 183636086-183636086 on Build GRCh38
rs986953409 Chr.1: 183635945-183635945 on Build GRCh38
rs997007112 Chr.1: 183636085-183636085 on Build GRCh38
rs955554923 Chr.1: 183635939-183635939 on Build GRCh38
rs561485132 Chr.1: 183636059-183636059 on Build GRCh38
rs113084640 Chr.1: 183635915-183635915 on Build GRCh38
rs890129952 Chr.1: 183636043-183636043 on Build GRCh38
rs879365769 Chr.1: 183636065-183636066 on Build GRCh38
rs1049528681 Chr.1: 183636139-183636139 on Build GRCh38
rs114491252 Chr.1: 183636108-183636108 on Build GRCh38
rs948587374 Chr.1: 183635938-183635938 on Build GRCh38
rs915978468 Chr.1: 183635936-183635936 on Build GRCh38
rs975590888 Chr.1: 183635786-183635786 on Build GRCh38
rs369097904 Chr.1: 183635903-183635903 on Build GRCh38
rs1045630152 Chr.1: 183636079-183636079 on Build GRCh38
rs993414537 Chr.1: 183635981-183635981 on Build GRCh38
rs529054008 Chr.1: 183635776-183635776 on Build GRCh38
rs148518448 Chr.1: 183636091-183636091 on Build GRCh38
rs146485943 Chr.1: 183636059-183636060 on Build GRCh38
rs1024044164 Chr.1: 183635876-183635876 on Build GRCh38
rs932504728 Chr.1: 183636121-183636121 on Build GRCh38
rs1047643152 Chr.1: 183636001-183636001 on Build GRCh38
rs1017324122 Chr.1: 183635774-183635774 on Build GRCh38
rs992828749 Chr.1: 183636117-183636117 on Build GRCh38
rs955650465 Chr.1: 183636117-183636117 on Build GRCh38
rs556505467 Chr.1: 183636058-183636059 on Build GRCh38
rs781464535 Chr.1: 183636051-183636051 on Build GRCh38
rs780354885 Chr.1: 183636089-183636089 on Build GRCh38

View Full Product Details