Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 14649610-14650009 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BFAR
Entrez Gene ID
51283
Gene Name
bifunctional apoptosis regulator
Gene Aliases
BAR, RNF47
UniGene
Hs.435556
Gene Chromosome Location
Chr.16: 14632811-14669236 on Build GRCh38
Transcripts
NM_016561.2 XM_005255350.2 XM_011522520.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1376006 c.547A>G p.T183A Chr.16: 14649882-14649882 on Build GRCh38
4578691 c.474C>T p.V158V Chr.16: 14649809-14649809 on Build GRCh38
6555429 c.603C>T p.Y201Y Chr.16: 14649938-14649938 on Build GRCh38
6611390 c.481G>A p.V161I Chr.16: 14649816-14649816 on Build GRCh38
967254 c.513C>T p.H171H Chr.16: 14649848-14649848 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772863445 Chr.16: 14649806-14649806 on Build GRCh38
rs1044122003 Chr.16: 14649693-14649693 on Build GRCh38
rs562492635 Chr.16: 14649743-14649743 on Build GRCh38
rs371409176 Chr.16: 14649911-14649911 on Build GRCh38
rs963342807 Chr.16: 14649763-14649763 on Build GRCh38
rs906965943 Chr.16: 14649714-14649714 on Build GRCh38
rs918755368 Chr.16: 14649846-14649846 on Build GRCh38
rs752156727 Chr.16: 14649816-14649816 on Build GRCh38
rs760095221 Chr.16: 14649872-14649880 on Build GRCh38
rs887422501 Chr.16: 14649671-14649671 on Build GRCh38
rs765201331 Chr.16: 14649953-14649953 on Build GRCh38
rs756398043 Chr.16: 14649686-14649686 on Build GRCh38
rs747698531 Chr.16: 14649876-14649876 on Build GRCh38
rs770604558 Chr.16: 14649886-14649886 on Build GRCh38
rs776118829 Chr.16: 14649796-14649796 on Build GRCh38
rs754112517 Chr.16: 14649962-14649962 on Build GRCh38
rs377450266 Chr.16: 14649759-14649759 on Build GRCh38
rs947646734 Chr.16: 14649908-14649908 on Build GRCh38
rs376171227 Chr.16: 14649784-14649784 on Build GRCh38
rs151058471 Chr.16: 14649961-14649961 on Build GRCh38
rs769381667 Chr.16: 14649805-14649805 on Build GRCh38
rs745744717 Chr.16: 14649758-14649758 on Build GRCh38
rs761771984 Chr.16: 14649947-14649948 on Build GRCh38
rs368712217 Chr.16: 14649835-14649835 on Build GRCh38
rs905856711 Chr.16: 14649972-14649979 on Build GRCh38
rs769576141 Chr.16: 14649877-14649877 on Build GRCh38
rs542933792 Chr.16: 14649649-14649649 on Build GRCh38
rs753729384 Chr.16: 14649894-14649896 on Build GRCh38
rs142851831 Chr.16: 14649887-14649887 on Build GRCh38
rs767714613 Chr.16: 14649905-14649905 on Build GRCh38
rs561315174 Chr.16: 14649664-14649664 on Build GRCh38
rs757300035 Chr.16: 14649979-14649979 on Build GRCh38
rs368538874 Chr.16: 14649647-14649647 on Build GRCh38
rs200444771 Chr.16: 14649902-14649902 on Build GRCh38
rs148268157 Chr.16: 14649809-14649809 on Build GRCh38
rs1044435805 Chr.16: 14649645-14649645 on Build GRCh38
rs374646053 Chr.16: 14649820-14649820 on Build GRCh38
rs916847142 Chr.16: 14649888-14649888 on Build GRCh38
rs1011963960 Chr.16: 14649633-14649633 on Build GRCh38
rs906917557 Chr.16: 14649974-14649974 on Build GRCh38
rs1044700375 Chr.16: 14649937-14649937 on Build GRCh38
rs200969701 Chr.16: 14649843-14649843 on Build GRCh38
rs757389544 Chr.16: 14649848-14649848 on Build GRCh38
rs779948887 Chr.16: 14649861-14649861 on Build GRCh38
rs760108645 Chr.16: 14649899-14649899 on Build GRCh38
rs560745143 Chr.16: 14649646-14649646 on Build GRCh38
rs767953902 Chr.16: 14649823-14649823 on Build GRCh38
rs370798120 Chr.16: 14649785-14649785 on Build GRCh38
rs1048715212 Chr.16: 14649651-14649651 on Build GRCh38
rs752810412 Chr.16: 14649832-14649832 on Build GRCh38
rs543884457 Chr.16: 14649740-14649740 on Build GRCh38
rs930872721 Chr.16: 14649842-14649842 on Build GRCh38
rs1044457859 Chr.16: 14649921-14649921 on Build GRCh38
rs761091096 Chr.16: 14649960-14649960 on Build GRCh38
rs777691307 Chr.16: 14649747-14649747 on Build GRCh38
rs777760259 Chr.16: 14649841-14649841 on Build GRCh38
rs1003028730 Chr.16: 14649976-14649976 on Build GRCh38
rs866005889 Chr.16: 14649907-14649907 on Build GRCh38
rs747009706 Chr.16: 14649862-14649862 on Build GRCh38
rs528892340 Chr.16: 14649672-14649672 on Build GRCh38
rs201159611 Chr.16: 14649883-14649883 on Build GRCh38
rs752855354 Chr.16: 14649685-14649685 on Build GRCh38
rs375968701 Chr.16: 14649919-14649919 on Build GRCh38
rs763611323 Chr.16: 14649874-14649874 on Build GRCh38
rs754691771 Chr.16: 14649874-14649874 on Build GRCh38
rs765392425 Chr.16: 14649982-14649982 on Build GRCh38
rs781064997 Chr.16: 14649875-14649875 on Build GRCh38
rs931491798 Chr.16: 14649872-14649872 on Build GRCh38
rs948389997 Chr.16: 14649916-14649916 on Build GRCh38
rs186678539 Chr.16: 14649815-14649815 on Build GRCh38
rs1013025965 Chr.16: 14649687-14649687 on Build GRCh38
rs780028803 Chr.16: 14649773-14649773 on Build GRCh38
rs377525507 Chr.16: 14649797-14649797 on Build GRCh38
rs748784777 Chr.16: 14649884-14649884 on Build GRCh38

View Full Product Details