Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 39522120-39522666 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BMP8A
Entrez Gene ID
353500
Gene Name
bone morphogenetic protein 8a
Gene Aliases
-
UniGene
Hs.472497
Gene Chromosome Location
Chr.1: 39491626-39529869 on Build GRCh38
Transcripts
NM_181809.3 NR_003929.2

Gene Symbol
PPIEL
Entrez Gene ID
728448
Gene Name
peptidylprolyl isomerase E like pseudogene
Gene Aliases
PPIEP1
UniGene
Hs.472508
Gene Chromosome Location
Chr.1: 39522280-39559698 on Build GRCh38
Transcripts
NM_181809.3 NR_003929.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4007829 c.880G>A p.G294S Chr.1: 39522414-39522414 on Build GRCh38
5955216 c.869-1G>C p.? Chr.1: 39522402-39522402 on Build GRCh38
1599867 c.878G>A p.R293H Chr.1: 39522412-39522412 on Build GRCh38
2168597 c.888C>T p.H296H Chr.1: 39522422-39522422 on Build GRCh38
6126265 c.900C>G p.V300V Chr.1: 39522434-39522434 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1034056053 Chr.1: 39522533-39522533 on Build GRCh38
rs148831881 Chr.1: 39522368-39522368 on Build GRCh38
rs979065267 Chr.1: 39522620-39522620 on Build GRCh38
rs533822280 Chr.1: 39522164-39522164 on Build GRCh38
rs797001273 Chr.1: 39522153-39522153 on Build GRCh38
rs911259674 Chr.1: 39522426-39522426 on Build GRCh38
rs15733 Chr.1: 39522362-39522362 on Build GRCh38
rs1001175143 Chr.1: 39522186-39522187 on Build GRCh38
rs765816990 Chr.1: 39522460-39522460 on Build GRCh38
rs530277521 Chr.1: 39522333-39522333 on Build GRCh38
rs749744565 Chr.1: 39522388-39522388 on Build GRCh38
rs994309598 Chr.1: 39522154-39522154 on Build GRCh38
rs6524 Chr.1: 39522473-39522473 on Build GRCh38
rs772235356 Chr.1: 39522445-39522445 on Build GRCh38
rs150437571 Chr.1: 39522559-39522559 on Build GRCh38
rs752248421 Chr.1: 39522530-39522530 on Build GRCh38
rs772984973 Chr.1: 39522455-39522455 on Build GRCh38
rs769409750 Chr.1: 39522507-39522507 on Build GRCh38
rs747057590 Chr.1: 39522495-39522495 on Build GRCh38
rs1034003388 Chr.1: 39522279-39522279 on Build GRCh38
rs547249403 Chr.1: 39522640-39522640 on Build GRCh38
rs779262942 Chr.1: 39522442-39522442 on Build GRCh38
rs774310870 Chr.1: 39522522-39522522 on Build GRCh38
rs762414161 Chr.1: 39522431-39522431 on Build GRCh38
rs143938877 Chr.1: 39522454-39522454 on Build GRCh38
rs541325890 Chr.1: 39522408-39522408 on Build GRCh38
rs754548275 Chr.1: 39522463-39522463 on Build GRCh38
rs143803708 Chr.1: 39522372-39522372 on Build GRCh38
rs765056618 Chr.1: 39522414-39522414 on Build GRCh38
rs748435654 Chr.1: 39522501-39522501 on Build GRCh38
rs368550248 Chr.1: 39522492-39522492 on Build GRCh38
rs967295555 Chr.1: 39522616-39522616 on Build GRCh38
rs775802393 Chr.1: 39522407-39522407 on Build GRCh38
rs372855499 Chr.1: 39522519-39522519 on Build GRCh38
rs768832662 Chr.1: 39522497-39522497 on Build GRCh38
rs772325200 Chr.1: 39522403-39522403 on Build GRCh38
rs753765223 Chr.1: 39522359-39522359 on Build GRCh38
rs536893761 Chr.1: 39522566-39522567 on Build GRCh38
rs757950052 Chr.1: 39522369-39522369 on Build GRCh38
rs751054754 Chr.1: 39522462-39522462 on Build GRCh38
rs370506690 Chr.1: 39522439-39522439 on Build GRCh38
rs61779338 Chr.1: 39522250-39522250 on Build GRCh38
rs368961164 Chr.1: 39522527-39522527 on Build GRCh38
rs757512908 Chr.1: 39522474-39522474 on Build GRCh38
rs368279529 Chr.1: 39522476-39522476 on Build GRCh38
rs763928974 Chr.1: 39522411-39522411 on Build GRCh38
rs371597606 Chr.1: 39522528-39522528 on Build GRCh38
rs146607525 Chr.1: 39522427-39522427 on Build GRCh38
rs139522712 Chr.1: 39522423-39522423 on Build GRCh38
rs754638242 Chr.1: 39522433-39522433 on Build GRCh38
rs762883509 Chr.1: 39522456-39522456 on Build GRCh38
rs61732885 Chr.1: 39522498-39522498 on Build GRCh38
rs1041311037 Chr.1: 39522144-39522144 on Build GRCh38
rs6525 Chr.1: 39522412-39522412 on Build GRCh38
rs746694562 Chr.1: 39522371-39522371 on Build GRCh38
rs936791496 Chr.1: 39522393-39522393 on Build GRCh38
rs710910 Chr.1: 39522227-39522227 on Build GRCh38
rs201171133 Chr.1: 39522441-39522441 on Build GRCh38
rs757209447 Chr.1: 39522577-39522577 on Build GRCh38
rs187060155 Chr.1: 39522585-39522585 on Build GRCh38
rs767044328 Chr.1: 39522464-39522464 on Build GRCh38
rs897353412 Chr.1: 39522147-39522148 on Build GRCh38
rs776955132 Chr.1: 39522448-39522448 on Build GRCh38
rs569812100 Chr.1: 39522317-39522317 on Build GRCh38
rs571256339 Chr.1: 39522159-39522159 on Build GRCh38
rs759500571 Chr.1: 39522402-39522402 on Build GRCh38
rs768759717 Chr.1: 39522447-39522447 on Build GRCh38
rs375479258 Chr.1: 39522487-39522487 on Build GRCh38
rs139291071 Chr.1: 39522521-39522521 on Build GRCh38
rs778013169 Chr.1: 39522380-39522380 on Build GRCh38
rs1020157302 Chr.1: 39522348-39522348 on Build GRCh38
rs771328743 Chr.1: 39522392-39522392 on Build GRCh38
rs763325002 Chr.1: 39522547-39522547 on Build GRCh38
rs780954766 Chr.1: 39522438-39522438 on Build GRCh38
rs556191502 Chr.1: 39522251-39522251 on Build GRCh38
rs775712272 Chr.1: 39522446-39522446 on Build GRCh38
rs774987392 Chr.1: 39522396-39522396 on Build GRCh38
rs750426402 Chr.1: 39522422-39522422 on Build GRCh38
rs761492913 Chr.1: 39522449-39522449 on Build GRCh38
rs910005 Chr.1: 39522225-39522225 on Build GRCh38
rs545115295 Chr.1: 39522565-39522565 on Build GRCh38
rs1020632470 Chr.1: 39522596-39522596 on Build GRCh38
rs888175274 Chr.1: 39522185-39522185 on Build GRCh38
rs547690412 Chr.1: 39522189-39522189 on Build GRCh38
rs765880860 Chr.1: 39522494-39522495 on Build GRCh38
rs773004673 Chr.1: 39522512-39522512 on Build GRCh38
rs527298107 Chr.1: 39522634-39522634 on Build GRCh38
rs761737627 Chr.1: 39522413-39522413 on Build GRCh38

View Full Product Details