Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 91942357-91942917 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTAF1
Entrez Gene ID
9044
Gene Name
B-TFIID TATA-box binding protein associated factor 1
Gene Aliases
MOT1, TAF(II)170, TAF172, TAFII170
UniGene
Hs.500526
Gene Chromosome Location
Chr.10: 91923769-92030998 on Build GRCh38
Transcripts
NM_003972.2 XM_011540326.1 XM_011540329.2 XM_017016877.1 XM_017016878.1 XM_017016879.1 XM_017016880.1 XM_017016881.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4484770 c.284C>T p.S95L Chr.10: 91942452-91942452 on Build GRCh38
7448378 c.400G>A p.G134S Chr.10: 91942568-91942568 on Build GRCh38
1969437 c.304A>C p.N102H Chr.10: 91942472-91942472 on Build GRCh38
1321980 c.328A>G p.R110G Chr.10: 91942496-91942496 on Build GRCh38
1969438 c.386A>G p.Q129R Chr.10: 91942554-91942554 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776674153 Chr.10: 91942604-91942604 on Build GRCh38
rs769372195 Chr.10: 91942615-91942615 on Build GRCh38
rs984584811 Chr.10: 91942473-91942473 on Build GRCh38
rs200862050 Chr.10: 91942577-91942577 on Build GRCh38
rs757396923 Chr.10: 91942540-91942540 on Build GRCh38
rs866064880 Chr.10: 91942702-91942702 on Build GRCh38
rs377624551 Chr.10: 91942617-91942617 on Build GRCh38
rs981368777 Chr.10: 91942843-91942843 on Build GRCh38
rs186332739 Chr.10: 91942760-91942760 on Build GRCh38
rs535651689 Chr.10: 91942701-91942701 on Build GRCh38
rs746363989 Chr.10: 91942834-91942834 on Build GRCh38
rs754739658 Chr.10: 91942465-91942465 on Build GRCh38
rs570500734 Chr.10: 91942873-91942873 on Build GRCh38
rs928411323 Chr.10: 91942806-91942806 on Build GRCh38
rs947084539 Chr.10: 91942601-91942601 on Build GRCh38
rs765617555 Chr.10: 91942424-91942424 on Build GRCh38
rs199538823 Chr.10: 91942442-91942442 on Build GRCh38
rs749331695 Chr.10: 91942387-91942387 on Build GRCh38
rs952799964 Chr.10: 91942715-91942715 on Build GRCh38
rs1028751329 Chr.10: 91942653-91942653 on Build GRCh38
rs746121749 Chr.10: 91942550-91942550 on Build GRCh38
rs113636788 Chr.10: 91942836-91942836 on Build GRCh38
rs775373054 Chr.10: 91942436-91942436 on Build GRCh38
rs1020463322 Chr.10: 91942777-91942777 on Build GRCh38
rs950040547 Chr.10: 91942433-91942433 on Build GRCh38
rs544300213 Chr.10: 91942859-91942859 on Build GRCh38
rs374223681 Chr.10: 91942588-91942588 on Build GRCh38
rs572783294 Chr.10: 91942717-91942717 on Build GRCh38
rs150594399 Chr.10: 91942512-91942512 on Build GRCh38
rs368980046 Chr.10: 91942515-91942515 on Build GRCh38
rs3814639 Chr.10: 91942686-91942686 on Build GRCh38
rs938501233 Chr.10: 91942830-91942830 on Build GRCh38
rs1019782952 Chr.10: 91942654-91942654 on Build GRCh38
rs947030960 Chr.10: 91942841-91942841 on Build GRCh38
rs112238331 Chr.10: 91942759-91942759 on Build GRCh38
rs370471025 Chr.10: 91942868-91942868 on Build GRCh38
rs764051948 Chr.10: 91942450-91942450 on Build GRCh38
rs199549229 Chr.10: 91942611-91942611 on Build GRCh38
rs942416904 Chr.10: 91942867-91942867 on Build GRCh38
rs1024125165 Chr.10: 91942753-91942753 on Build GRCh38
rs957541039 Chr.10: 91942731-91942731 on Build GRCh38
rs142485255 Chr.10: 91942486-91942486 on Build GRCh38
rs781693790 Chr.10: 91942712-91942712 on Build GRCh38
rs868633654 Chr.10: 91942431-91942431 on Build GRCh38
rs749754196 Chr.10: 91942518-91942518 on Build GRCh38
rs17107230 Chr.10: 91942571-91942571 on Build GRCh38
rs779996417 Chr.10: 91942564-91942564 on Build GRCh38
rs746045054 Chr.10: 91942412-91942412 on Build GRCh38
rs148405175 Chr.10: 91942453-91942453 on Build GRCh38
rs1036269487 Chr.10: 91942634-91942634 on Build GRCh38
rs3814640 Chr.10: 91942648-91942648 on Build GRCh38
rs916689914 Chr.10: 91942474-91942474 on Build GRCh38
rs1025908665 Chr.10: 91942706-91942706 on Build GRCh38
rs764625023 Chr.10: 91942469-91942469 on Build GRCh38
rs752993328 Chr.10: 91942498-91942498 on Build GRCh38
rs770137837 Chr.10: 91942856-91942856 on Build GRCh38
rs901524608 Chr.10: 91942636-91942636 on Build GRCh38
rs574856024 Chr.10: 91942789-91942789 on Build GRCh38
rs750142443 Chr.10: 91942479-91942479 on Build GRCh38
rs372700430 Chr.10: 91942462-91942462 on Build GRCh38
rs774701108 Chr.10: 91942408-91942408 on Build GRCh38
rs374999557 Chr.10: 91942495-91942495 on Build GRCh38
rs927289884 Chr.10: 91942846-91942846 on Build GRCh38
rs913164592 Chr.10: 91942838-91942838 on Build GRCh38
rs752955781 Chr.10: 91942504-91942504 on Build GRCh38
rs756628102 Chr.10: 91942510-91942510 on Build GRCh38
rs772039529 Chr.10: 91942413-91942413 on Build GRCh38
rs927358272 Chr.10: 91942460-91942460 on Build GRCh38
rs1005233522 Chr.10: 91942589-91942589 on Build GRCh38
rs190874493 Chr.10: 91942773-91942773 on Build GRCh38
rs1053025176 Chr.10: 91942575-91942575 on Build GRCh38
rs374249746 Chr.10: 91942388-91942388 on Build GRCh38
rs376933322 Chr.10: 91942541-91942541 on Build GRCh38
rs953072287 Chr.10: 91942669-91942669 on Build GRCh38

View Full Product Details