Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 111293437-111293988 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COLCA1
Entrez Gene ID
399948
Gene Name
colorectal cancer associated 1
Gene Aliases
C11orf92, CASC12, LOH11CR1F
UniGene
Hs.729225
Gene Chromosome Location
Chr.11: 111293389-111305048 on Build GRCh38
Transcripts
NM_001302644.1 NM_001302645.1 NM_001302646.1 NM_001302647.1 NM_001302648.1 NM_207429.3

SNP Detail

SNP ID Chromosome Location
rs1049514533 Chr.11: 111293680-111293680 on Build GRCh38
rs953042261 Chr.11: 111293740-111293740 on Build GRCh38
rs909327277 Chr.11: 111293490-111293490 on Build GRCh38
rs551466182 Chr.11: 111293554-111293554 on Build GRCh38
rs911640531 Chr.11: 111293811-111293811 on Build GRCh38
rs931661172 Chr.11: 111293544-111293544 on Build GRCh38
rs772260275 Chr.11: 111293595-111293595 on Build GRCh38
rs983487247 Chr.11: 111293752-111293752 on Build GRCh38
rs1020156318 Chr.11: 111293700-111293700 on Build GRCh38
rs557672176 Chr.11: 111293901-111293901 on Build GRCh38
rs1049281690 Chr.11: 111293842-111293842 on Build GRCh38
rs184657278 Chr.11: 111293636-111293636 on Build GRCh38
rs539581975 Chr.11: 111293900-111293900 on Build GRCh38
rs978679532 Chr.11: 111293724-111293724 on Build GRCh38
rs992662038 Chr.11: 111293495-111293495 on Build GRCh38
rs1009675931 Chr.11: 111293913-111293913 on Build GRCh38
rs566601657 Chr.11: 111293863-111293863 on Build GRCh38
rs550827624 Chr.11: 111293817-111293817 on Build GRCh38
rs1000626762 Chr.11: 111293903-111293903 on Build GRCh38
rs930820054 Chr.11: 111293838-111293838 on Build GRCh38
rs1038247361 Chr.11: 111293520-111293520 on Build GRCh38
rs531099110 Chr.11: 111293633-111293633 on Build GRCh38
rs945286294 Chr.11: 111293865-111293865 on Build GRCh38
rs115778629 Chr.11: 111293733-111293733 on Build GRCh38
rs113059789 Chr.11: 111293699-111293699 on Build GRCh38
rs980561439 Chr.11: 111293808-111293808 on Build GRCh38
rs944839115 Chr.11: 111293599-111293599 on Build GRCh38
rs752479668 Chr.11: 111293689-111293689 on Build GRCh38
rs905445340 Chr.11: 111293864-111293864 on Build GRCh38
rs960819098 Chr.11: 111293698-111293698 on Build GRCh38
rs913148860 Chr.11: 111293504-111293504 on Build GRCh38
rs569075015 Chr.11: 111293855-111293855 on Build GRCh38
rs967680771 Chr.11: 111293709-111293709 on Build GRCh38
rs144051104 Chr.11: 111293519-111293519 on Build GRCh38
rs1036529823 Chr.11: 111293601-111293601 on Build GRCh38
rs752476184 Chr.11: 111293963-111293963 on Build GRCh38
rs200137348 Chr.11: 111293521-111293521 on Build GRCh38
rs566393298 Chr.11: 111293468-111293468 on Build GRCh38
rs959825771 Chr.11: 111293479-111293479 on Build GRCh38
rs942131114 Chr.11: 111293787-111293787 on Build GRCh38
rs904986164 Chr.11: 111293540-111293540 on Build GRCh38
rs763846883 Chr.11: 111293834-111293834 on Build GRCh38
rs925663781 Chr.11: 111293527-111293527 on Build GRCh38
rs759198902 Chr.11: 111293915-111293937 on Build GRCh38
rs547004154 Chr.11: 111293721-111293721 on Build GRCh38
rs535727408 Chr.11: 111293816-111293816 on Build GRCh38
rs967320687 Chr.11: 111293508-111293508 on Build GRCh38
rs976342620 Chr.11: 111293711-111293711 on Build GRCh38
rs938023063 Chr.11: 111293887-111293887 on Build GRCh38
rs764947694 Chr.11: 111293684-111293684 on Build GRCh38
rs1054768 Chr.11: 111293695-111293695 on Build GRCh38
rs1056282269 Chr.11: 111293550-111293550 on Build GRCh38
rs900684849 Chr.11: 111293941-111293941 on Build GRCh38
rs11213821 Chr.11: 111293926-111293926 on Build GRCh38
rs986205010 Chr.11: 111293753-111293753 on Build GRCh38
rs762028578 Chr.11: 111293563-111293563 on Build GRCh38
rs902990072 Chr.11: 111293927-111293927 on Build GRCh38

View Full Product Details