Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 157148378-157148901 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCNL1
Entrez Gene ID
57018
Gene Name
cyclin L1
Gene Aliases
ANIA6A, BM-001, PRO1073, ania-6a
UniGene
Hs.4859
Gene Chromosome Location
Chr.3: 157146502-157160760 on Build GRCh38
Transcripts
NM_001308185.1 NM_020307.3 XM_005247648.3 XM_006713710.3 XM_011513011.2 XM_017006891.1 XM_017006892.1 XM_017006893.1 XM_017006894.1 XM_017006895.1 XM_017006896.1 XM_017006897.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3589331 c.1435C>T p.R479W Chr.3: 157148387-157148387 on Build GRCh38
5964814 c.1243A>G p.R415G Chr.3: 157148579-157148579 on Build GRCh38
4404219 c.1302T>G p.S434R Chr.3: 157148520-157148520 on Build GRCh38
3774673 c.1325A>T p.N442I Chr.3: 157148497-157148497 on Build GRCh38
1040399 c.1411C>T p.R471C Chr.3: 157148411-157148411 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781270124 Chr.3: 157148503-157148503 on Build GRCh38
rs1043751666 Chr.3: 157148740-157148740 on Build GRCh38
rs1047984114 Chr.3: 157148664-157148664 on Build GRCh38
rs972799170 Chr.3: 157148876-157148876 on Build GRCh38
rs747331518 Chr.3: 157148530-157148530 on Build GRCh38
rs551931843 Chr.3: 157148647-157148647 on Build GRCh38
rs778485731 Chr.3: 157148438-157148438 on Build GRCh38
rs1007142524 Chr.3: 157148877-157148877 on Build GRCh38
rs1051650634 Chr.3: 157148467-157148467 on Build GRCh38
rs761277889 Chr.3: 157148587-157148587 on Build GRCh38
rs767422683 Chr.3: 157148410-157148410 on Build GRCh38
rs771814832 Chr.3: 157148569-157148569 on Build GRCh38
rs546060937 Chr.3: 157148404-157148404 on Build GRCh38
rs746220067 Chr.3: 157148464-157148464 on Build GRCh38
rs764909288 Chr.3: 157148598-157148598 on Build GRCh38
rs62274131 Chr.3: 157148707-157148707 on Build GRCh38
rs774195037 Chr.3: 157148403-157148403 on Build GRCh38
rs769303845 Chr.3: 157148495-157148495 on Build GRCh38
rs537235801 Chr.3: 157148823-157148823 on Build GRCh38
rs548396334 Chr.3: 157148658-157148658 on Build GRCh38
rs755542466 Chr.3: 157148638-157148638 on Build GRCh38
rs1029844869 Chr.3: 157148492-157148492 on Build GRCh38
rs552398751 Chr.3: 157148833-157148833 on Build GRCh38
rs773357422 Chr.3: 157148531-157148531 on Build GRCh38
rs970471661 Chr.3: 157148795-157148795 on Build GRCh38
rs774540849 Chr.3: 157148488-157148488 on Build GRCh38
rs762545896 Chr.3: 157148879-157148888 on Build GRCh38
rs142030218 Chr.3: 157148500-157148500 on Build GRCh38
rs748246366 Chr.3: 157148509-157148509 on Build GRCh38
rs756077496 Chr.3: 157148429-157148429 on Build GRCh38
rs747466174 Chr.3: 157148494-157148494 on Build GRCh38
rs939268186 Chr.3: 157148702-157148702 on Build GRCh38
rs984172003 Chr.3: 157148805-157148805 on Build GRCh38
rs528424179 Chr.3: 157148479-157148479 on Build GRCh38
rs753373067 Chr.3: 157148583-157148583 on Build GRCh38
rs940109542 Chr.3: 157148825-157148825 on Build GRCh38
rs1030205771 Chr.3: 157148807-157148807 on Build GRCh38
rs780594103 Chr.3: 157148480-157148480 on Build GRCh38
rs376153963 Chr.3: 157148628-157148628 on Build GRCh38
rs955565562 Chr.3: 157148864-157148864 on Build GRCh38
rs112810419 Chr.3: 157148412-157148412 on Build GRCh38
rs1023178447 Chr.3: 157148760-157148760 on Build GRCh38
rs372986934 Chr.3: 157148608-157148608 on Build GRCh38
rs764005118 Chr.3: 157148581-157148581 on Build GRCh38
rs181135757 Chr.3: 157148550-157148550 on Build GRCh38
rs753292523 Chr.3: 157148432-157148432 on Build GRCh38
rs750053960 Chr.3: 157148599-157148599 on Build GRCh38
rs185779824 Chr.3: 157148646-157148646 on Build GRCh38
rs902567435 Chr.3: 157148752-157148752 on Build GRCh38
rs763612660 Chr.3: 157148430-157148430 on Build GRCh38
rs1018569140 Chr.3: 157148879-157148879 on Build GRCh38
rs112429746 Chr.3: 157148683-157148683 on Build GRCh38
rs760539080 Chr.3: 157148575-157148575 on Build GRCh38
rs999625412 Chr.3: 157148802-157148802 on Build GRCh38
rs761958662 Chr.3: 157148715-157148715 on Build GRCh38
rs964410288 Chr.3: 157148604-157148604 on Build GRCh38
rs751981790 Chr.3: 157148635-157148635 on Build GRCh38
rs112473291 Chr.3: 157148731-157148731 on Build GRCh38
rs756930379 Chr.3: 157148434-157148434 on Build GRCh38

View Full Product Details