Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 44522599-44523142 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEACAM20
Entrez Gene ID
125931
Gene Name
carcinoembryonic antigen related cell adhesion molecule 20
Gene Aliases
UNQ9366
UniGene
Hs.689632
Gene Chromosome Location
Chr.19: 44503838-44530100 on Build GRCh38
Transcripts
NM_001102597.2 NM_001102598.2 NM_001102599.2 NM_001102600.2 XM_011526429.2 XM_011526430.2 XM_011526431.2 XM_011526432.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3535625 c.727G>A p.G243S Chr.19: 44522658-44522658 on Build GRCh38
1159103 c.566C>T p.A189V Chr.19: 44522819-44522819 on Build GRCh38
5856097 c.511G>T p.V171F Chr.19: 44522874-44522874 on Build GRCh38
3535627 c.485C>T p.P162L Chr.19: 44522900-44522900 on Build GRCh38
3535626 c.617C>T p.S206F Chr.19: 44522768-44522768 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369349636 Chr.19: 44522797-44522797 on Build GRCh38
rs761671938 Chr.19: 44522936-44522936 on Build GRCh38
rs549840052 Chr.19: 44523016-44523016 on Build GRCh38
rs190082074 Chr.19: 44522959-44522959 on Build GRCh38
rs764457765 Chr.19: 44522956-44522957 on Build GRCh38
rs145824457 Chr.19: 44523108-44523108 on Build GRCh38
rs559911609 Chr.19: 44522925-44522925 on Build GRCh38
rs750486847 Chr.19: 44522705-44522705 on Build GRCh38
rs937902776 Chr.19: 44523035-44523035 on Build GRCh38
rs757992589 Chr.19: 44522857-44522857 on Build GRCh38
rs374188048 Chr.19: 44522933-44522933 on Build GRCh38
rs368941407 Chr.19: 44522643-44522643 on Build GRCh38
rs140913297 Chr.19: 44522924-44522924 on Build GRCh38
rs372889795 Chr.19: 44522818-44522818 on Build GRCh38
rs771423208 Chr.19: 44522750-44522750 on Build GRCh38
rs771785095 Chr.19: 44522662-44522662 on Build GRCh38
rs777758011 Chr.19: 44523043-44523043 on Build GRCh38
rs779525407 Chr.19: 44522862-44522862 on Build GRCh38
rs781151078 Chr.19: 44522982-44522982 on Build GRCh38
rs751103480 Chr.19: 44522960-44522960 on Build GRCh38
rs774956877 Chr.19: 44522664-44522664 on Build GRCh38
rs1019878370 Chr.19: 44522890-44522890 on Build GRCh38
rs776227125 Chr.19: 44522880-44522880 on Build GRCh38
rs148998031 Chr.19: 44523109-44523109 on Build GRCh38
rs759148089 Chr.19: 44522661-44522661 on Build GRCh38
rs754408705 Chr.19: 44522670-44522670 on Build GRCh38
rs202136602 Chr.19: 44522886-44522886 on Build GRCh38
rs763601783 Chr.19: 44522951-44522952 on Build GRCh38
rs756950141 Chr.19: 44522843-44522843 on Build GRCh38
rs905785519 Chr.19: 44522999-44522999 on Build GRCh38
rs765281538 Chr.19: 44522687-44522687 on Build GRCh38
rs745723023 Chr.19: 44522848-44522848 on Build GRCh38
rs762295727 Chr.19: 44522681-44522681 on Build GRCh38
rs575078262 Chr.19: 44522814-44522814 on Build GRCh38
rs530000725 Chr.19: 44523006-44523006 on Build GRCh38
rs571679459 Chr.19: 44522734-44522734 on Build GRCh38
rs769668085 Chr.19: 44522627-44522627 on Build GRCh38
rs569810369 Chr.19: 44523040-44523040 on Build GRCh38
rs199934659 Chr.19: 44522793-44522793 on Build GRCh38
rs766649226 Chr.19: 44522709-44522709 on Build GRCh38
rs749173684 Chr.19: 44522642-44522642 on Build GRCh38
rs761041865 Chr.19: 44522666-44522666 on Build GRCh38
rs778921574 Chr.19: 44522847-44522847 on Build GRCh38
rs538467753 Chr.19: 44523047-44523047 on Build GRCh38
rs745404885 Chr.19: 44522748-44522748 on Build GRCh38
rs778673424 Chr.19: 44522740-44522740 on Build GRCh38
rs35542660 Chr.19: 44522644-44522644 on Build GRCh38
rs764937261 Chr.19: 44522949-44522949 on Build GRCh38
rs913769717 Chr.19: 44522973-44522973 on Build GRCh38
rs777868464 Chr.19: 44522633-44522633 on Build GRCh38
rs1046290798 Chr.19: 44522685-44522685 on Build GRCh38
rs920238317 Chr.19: 44523055-44523055 on Build GRCh38
rs767871619 Chr.19: 44522932-44522932 on Build GRCh38
rs1010324785 Chr.19: 44523076-44523076 on Build GRCh38
rs1038061639 Chr.19: 44522640-44522640 on Build GRCh38
rs1020406329 Chr.19: 44523091-44523091 on Build GRCh38
rs968903381 Chr.19: 44523094-44523094 on Build GRCh38
rs574733969 Chr.19: 44522706-44522706 on Build GRCh38
rs773887584 Chr.19: 44522646-44522646 on Build GRCh38
rs751778179 Chr.19: 44522719-44522719 on Build GRCh38
rs746659853 Chr.19: 44522759-44522759 on Build GRCh38
rs377750015 Chr.19: 44522922-44522922 on Build GRCh38
rs887263323 Chr.19: 44522855-44522855 on Build GRCh38
rs1001517912 Chr.19: 44523033-44523033 on Build GRCh38
rs752793453 Chr.19: 44522810-44522811 on Build GRCh38
rs528463074 Chr.19: 44522950-44522950 on Build GRCh38
rs764016955 Chr.19: 44522827-44522827 on Build GRCh38
rs374407949 Chr.19: 44522910-44522910 on Build GRCh38
rs929102299 Chr.19: 44523098-44523098 on Build GRCh38
rs775212192 Chr.19: 44522753-44522753 on Build GRCh38
rs753715606 Chr.19: 44522840-44522840 on Build GRCh38
rs757184711 Chr.19: 44522738-44522738 on Build GRCh38
rs867739257 Chr.19: 44522906-44522906 on Build GRCh38
rs1037611306 Chr.19: 44522983-44522983 on Build GRCh38
rs766306939 Chr.19: 44522815-44522815 on Build GRCh38
rs540641400 Chr.19: 44522791-44522791 on Build GRCh38
rs746713334 Chr.19: 44522864-44522864 on Build GRCh38
rs567402945 Chr.19: 44522965-44522965 on Build GRCh38
rs753987014 Chr.19: 44522669-44522669 on Build GRCh38
rs558195493 Chr.19: 44523068-44523068 on Build GRCh38
rs374925599 Chr.19: 44522819-44522819 on Build GRCh38
rs765805694 Chr.19: 44522795-44522795 on Build GRCh38
rs769067425 Chr.19: 44522789-44522789 on Build GRCh38
rs769997966 Chr.19: 44522891-44522891 on Build GRCh38
rs964648289 Chr.19: 44523119-44523119 on Build GRCh38
rs968916812 Chr.19: 44522801-44522801 on Build GRCh38
rs1035747406 Chr.19: 44522703-44522703 on Build GRCh38
rs758247414 Chr.19: 44522954-44522954 on Build GRCh38
rs563895998 Chr.19: 44522868-44522868 on Build GRCh38
rs755981556 Chr.19: 44522729-44522729 on Build GRCh38
rs200839672 Chr.19: 44522813-44522813 on Build GRCh38

View Full Product Details