Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 42733368-42733882 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLDN19
Entrez Gene ID
149461
Gene Name
claudin 19
Gene Aliases
HOMG5
UniGene
Hs.496270
Gene Chromosome Location
Chr.1: 42733093-42740254 on Build GRCh38
Transcripts
NM_001123395.1 NM_001185117.1 NM_148960.2

SNP Detail

SNP ID Chromosome Location
rs753515106 Chr.1: 42733844-42733844 on Build GRCh38
rs976373319 Chr.1: 42733802-42733802 on Build GRCh38
rs958906433 Chr.1: 42733446-42733446 on Build GRCh38
rs766100803 Chr.1: 42733800-42733800 on Build GRCh38
rs962046453 Chr.1: 42733761-42733761 on Build GRCh38
rs149256970 Chr.1: 42733658-42733658 on Build GRCh38
rs933476755 Chr.1: 42733579-42733579 on Build GRCh38
rs796901250 Chr.1: 42733625-42733634 on Build GRCh38
rs200527665 Chr.1: 42733729-42733729 on Build GRCh38
rs866481412 Chr.1: 42733417-42733417 on Build GRCh38
rs373188579 Chr.1: 42733676-42733676 on Build GRCh38
rs985131155 Chr.1: 42733748-42733748 on Build GRCh38
rs539642986 Chr.1: 42733412-42733412 on Build GRCh38
rs546730630 Chr.1: 42733797-42733797 on Build GRCh38
rs1040226305 Chr.1: 42733554-42733554 on Build GRCh38
rs553394106 Chr.1: 42733425-42733425 on Build GRCh38
rs1054107067 Chr.1: 42733646-42733646 on Build GRCh38
rs1057515568 Chr.1: 42733774-42733774 on Build GRCh38
rs56345167 Chr.1: 42733444-42733444 on Build GRCh38
rs889365281 Chr.1: 42733630-42733630 on Build GRCh38
rs1014880889 Chr.1: 42733683-42733683 on Build GRCh38
rs958630245 Chr.1: 42733745-42733745 on Build GRCh38
rs562296433 Chr.1: 42733625-42733631 on Build GRCh38
rs933422896 Chr.1: 42733833-42733833 on Build GRCh38
rs981285231 Chr.1: 42733428-42733428 on Build GRCh38
rs1051069098 Chr.1: 42733603-42733603 on Build GRCh38
rs529449539 Chr.1: 42733634-42733636 on Build GRCh38
rs567823099 Chr.1: 42733709-42733709 on Build GRCh38
rs972101560 Chr.1: 42733774-42733774 on Build GRCh38
rs76855155 Chr.1: 42733759-42733759 on Build GRCh38
rs556016559 Chr.1: 42733537-42733537 on Build GRCh38
rs969836365 Chr.1: 42733443-42733443 on Build GRCh38
rs923346842 Chr.1: 42733777-42733777 on Build GRCh38
rs931824276 Chr.1: 42733595-42733595 on Build GRCh38
rs71654253 Chr.1: 42733557-42733557 on Build GRCh38
rs745884385 Chr.1: 42733733-42733733 on Build GRCh38
rs113021721 Chr.1: 42733492-42733492 on Build GRCh38
rs544361263 Chr.1: 42733685-42733685 on Build GRCh38
rs1057515454 Chr.1: 42733465-42733465 on Build GRCh38
rs770989443 Chr.1: 42733568-42733568 on Build GRCh38
rs926038941 Chr.1: 42733857-42733857 on Build GRCh38
rs763565629 Chr.1: 42733490-42733490 on Build GRCh38
rs1024484460 Chr.1: 42733707-42733707 on Build GRCh38
rs765727450 Chr.1: 42733388-42733388 on Build GRCh38
rs944594774 Chr.1: 42733553-42733553 on Build GRCh38
rs371301647 Chr.1: 42733495-42733495 on Build GRCh38
rs965993611 Chr.1: 42733796-42733796 on Build GRCh38
rs1009910087 Chr.1: 42733663-42733663 on Build GRCh38
rs1014521584 Chr.1: 42733776-42733776 on Build GRCh38
rs951754166 Chr.1: 42733482-42733482 on Build GRCh38
rs1001707873 Chr.1: 42733723-42733724 on Build GRCh38
rs1049876055 Chr.1: 42733627-42733627 on Build GRCh38
rs972155294 Chr.1: 42733517-42733517 on Build GRCh38
rs1057515567 Chr.1: 42733603-42733603 on Build GRCh38
rs923397849 Chr.1: 42733566-42733566 on Build GRCh38
rs760068656 Chr.1: 42733763-42733763 on Build GRCh38
rs904588472 Chr.1: 42733642-42733642 on Build GRCh38
rs984795189 Chr.1: 42733480-42733480 on Build GRCh38
rs796175275 Chr.1: 42733636-42733636 on Build GRCh38
rs573446568 Chr.1: 42733437-42733437 on Build GRCh38

View Full Product Details