Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 93076809-93077343 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP26A1
Entrez Gene ID
1592
Gene Name
cytochrome P450 family 26 subfamily A member 1
Gene Aliases
CP26, CYP26, P450RAI, P450RAI1
UniGene
Hs.150595
Gene Chromosome Location
Chr.10: 93073475-93077884 on Build GRCh38
Transcripts
NM_000783.3 NM_057157.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7411468 c.965G>C p.G322A Chr.10: 93076982-93076982 on Build GRCh38
4568807 c.1119T>C p.C373C Chr.10: 93077136-93077136 on Build GRCh38
5529495 c.1227C>T p.P409P Chr.10: 93077244-93077244 on Build GRCh38
4016680 c.1378G>A p.A460T Chr.10: 93077188-93077188 on Build GRCh38
3441582 c.1278G>A p.G426G Chr.10: 93077295-93077295 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1051702361 Chr.10: 93076851-93076851 on Build GRCh38
rs146552397 Chr.10: 93077185-93077185 on Build GRCh38
rs200820945 Chr.10: 93077300-93077300 on Build GRCh38
rs867165563 Chr.10: 93077131-93077131 on Build GRCh38
rs776389104 Chr.10: 93076921-93076921 on Build GRCh38
rs748305490 Chr.10: 93077148-93077148 on Build GRCh38
rs140117559 Chr.10: 93077132-93077132 on Build GRCh38
rs202120226 Chr.10: 93077046-93077046 on Build GRCh38
rs547906104 Chr.10: 93076982-93076982 on Build GRCh38
rs757863072 Chr.10: 93076950-93076950 on Build GRCh38
rs759138719 Chr.10: 93077216-93077216 on Build GRCh38
rs1010072383 Chr.10: 93076899-93076899 on Build GRCh38
rs267602619 Chr.10: 93077205-93077205 on Build GRCh38
rs757601865 Chr.10: 93076994-93076994 on Build GRCh38
rs756334642 Chr.10: 93077278-93077278 on Build GRCh38
rs762843948 Chr.10: 93077179-93077179 on Build GRCh38
rs73319400 Chr.10: 93077001-93077001 on Build GRCh38
rs766895381 Chr.10: 93077254-93077254 on Build GRCh38
rs984252420 Chr.10: 93077062-93077062 on Build GRCh38
rs1021469654 Chr.10: 93077255-93077255 on Build GRCh38
rs762945730 Chr.10: 93077068-93077068 on Build GRCh38
rs893631201 Chr.10: 93077241-93077241 on Build GRCh38
rs763936477 Chr.10: 93076919-93076919 on Build GRCh38
rs752009503 Chr.10: 93077261-93077261 on Build GRCh38
rs888924713 Chr.10: 93076858-93076858 on Build GRCh38
rs958139700 Chr.10: 93076890-93076890 on Build GRCh38
rs375225582 Chr.10: 93077253-93077253 on Build GRCh38
rs186901298 Chr.10: 93077118-93077118 on Build GRCh38
rs1037964029 Chr.10: 93076900-93076901 on Build GRCh38
rs750957354 Chr.10: 93077122-93077122 on Build GRCh38
rs765122042 Chr.10: 93076937-93076937 on Build GRCh38
rs750330267 Chr.10: 93076945-93076945 on Build GRCh38
rs768162822 Chr.10: 93077316-93077317 on Build GRCh38
rs200904706 Chr.10: 93076993-93076993 on Build GRCh38
rs748379581 Chr.10: 93077277-93077277 on Build GRCh38
rs780171621 Chr.10: 93077044-93077044 on Build GRCh38
rs910088132 Chr.10: 93077078-93077078 on Build GRCh38
rs1032214397 Chr.10: 93076888-93076888 on Build GRCh38
rs759298995 Chr.10: 93077087-93077087 on Build GRCh38
rs757574991 Chr.10: 93077117-93077117 on Build GRCh38
rs776793858 Chr.10: 93077058-93077058 on Build GRCh38
rs983953239 Chr.10: 93077262-93077262 on Build GRCh38
rs773044740 Chr.10: 93077172-93077172 on Build GRCh38
rs752401058 Chr.10: 93077236-93077236 on Build GRCh38
rs747893905 Chr.10: 93077142-93077142 on Build GRCh38
rs1051458953 Chr.10: 93077151-93077151 on Build GRCh38
rs768836264 Chr.10: 93077055-93077055 on Build GRCh38
rs762717980 Chr.10: 93077175-93077176 on Build GRCh38
rs751088799 Chr.10: 93077085-93077085 on Build GRCh38
rs200246602 Chr.10: 93077018-93077018 on Build GRCh38
rs747123621 Chr.10: 93077128-93077128 on Build GRCh38
rs927404017 Chr.10: 93076928-93076928 on Build GRCh38
rs746326359 Chr.10: 93077002-93077002 on Build GRCh38
rs889031645 Chr.10: 93077152-93077152 on Build GRCh38
rs773031908 Chr.10: 93077067-93077067 on Build GRCh38
rs769546059 Chr.10: 93077166-93077166 on Build GRCh38
rs767077820 Chr.10: 93077230-93077230 on Build GRCh38
rs750770595 Chr.10: 93077248-93077248 on Build GRCh38
rs774387193 Chr.10: 93077198-93077198 on Build GRCh38
rs371966523 Chr.10: 93077140-93077140 on Build GRCh38
rs778327215 Chr.10: 93076991-93076991 on Build GRCh38
rs770796705 Chr.10: 93077312-93077312 on Build GRCh38
rs777466689 Chr.10: 93077296-93077296 on Build GRCh38
rs761622656 Chr.10: 93077060-93077060 on Build GRCh38
rs755058456 Chr.10: 93077268-93077268 on Build GRCh38
rs768677075 Chr.10: 93077136-93077136 on Build GRCh38
rs114797676 Chr.10: 93076883-93076883 on Build GRCh38
rs754661514 Chr.10: 93076985-93076985 on Build GRCh38
rs144194304 Chr.10: 93077139-93077139 on Build GRCh38
rs765850598 Chr.10: 93076968-93076968 on Build GRCh38
rs775129137 Chr.10: 93077143-93077143 on Build GRCh38
rs765862190 Chr.10: 93077073-93077073 on Build GRCh38
rs767305072 Chr.10: 93077112-93077112 on Build GRCh38
rs2229104 Chr.10: 93077124-93077124 on Build GRCh38
rs774442573 Chr.10: 93077316-93077316 on Build GRCh38
rs895553734 Chr.10: 93077000-93077000 on Build GRCh38
rs761779807 Chr.10: 93076923-93076923 on Build GRCh38
rs771590214 Chr.10: 93077063-93077063 on Build GRCh38
rs765572348 Chr.10: 93077247-93077247 on Build GRCh38
rs752348123 Chr.10: 93077116-93077116 on Build GRCh38

View Full Product Details