Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 140670643-140671159 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DND1
Entrez Gene ID
373863
Gene Name
DND microRNA-mediated repression inhibitor 1
Gene Aliases
RBMS4
UniGene
Hs.591262
Gene Chromosome Location
Chr.5: 140670796-140673586 on Build GRCh38
Transcripts
NM_194249.2 NM_017706.4 XM_005268469.2 XM_017009600.1

Gene Symbol
WDR55
Entrez Gene ID
54853
Gene Name
WD repeat domain 55
Gene Aliases
-
UniGene
Hs.286261
Gene Chromosome Location
Chr.5: 140664799-140674119 on Build GRCh38
Transcripts
NM_194249.2 NM_017706.4 XM_005268469.2 XM_017009600.1

SNP Detail

SNP ID Chromosome Location
rs73791745 Chr.5: 140671102-140671102 on Build GRCh38
rs751002250 Chr.5: 140670790-140670790 on Build GRCh38
rs961937431 Chr.5: 140670713-140670713 on Build GRCh38
rs1047079061 Chr.5: 140670959-140670959 on Build GRCh38
rs1021168673 Chr.5: 140671104-140671104 on Build GRCh38
rs928359466 Chr.5: 140670966-140670966 on Build GRCh38
rs765882882 Chr.5: 140670740-140670740 on Build GRCh38
rs866872867 Chr.5: 140670681-140670681 on Build GRCh38
rs111716569 Chr.5: 140670928-140670928 on Build GRCh38
rs1030364774 Chr.5: 140670849-140670850 on Build GRCh38
rs551438669 Chr.5: 140670708-140670708 on Build GRCh38
rs755542325 Chr.5: 140670810-140670810 on Build GRCh38
rs894246812 Chr.5: 140671007-140671007 on Build GRCh38
rs566898193 Chr.5: 140670675-140670675 on Build GRCh38
rs958082289 Chr.5: 140670918-140670918 on Build GRCh38
rs1052760 Chr.5: 140671051-140671051 on Build GRCh38
rs774566708 Chr.5: 140670938-140670940 on Build GRCh38
rs574068916 Chr.5: 140670885-140670885 on Build GRCh38
rs938329117 Chr.5: 140671000-140671000 on Build GRCh38
rs989666939 Chr.5: 140670927-140670928 on Build GRCh38
rs702397 Chr.5: 140670680-140670680 on Build GRCh38
rs1052767 Chr.5: 140670951-140670951 on Build GRCh38
rs1002661556 Chr.5: 140670672-140670672 on Build GRCh38
rs528686052 Chr.5: 140671138-140671138 on Build GRCh38
rs59018738 Chr.5: 140670852-140670852 on Build GRCh38
rs967278387 Chr.5: 140671128-140671128 on Build GRCh38
rs573252786 Chr.5: 140670780-140670780 on Build GRCh38
rs755886974 Chr.5: 140670860-140670860 on Build GRCh38
rs368759325 Chr.5: 140671057-140671057 on Build GRCh38
rs376301671 Chr.5: 140670999-140671001 on Build GRCh38
rs527406564 Chr.5: 140671046-140671046 on Build GRCh38
rs188995384 Chr.5: 140670678-140670678 on Build GRCh38
rs551515581 Chr.5: 140671020-140671020 on Build GRCh38
rs190826909 Chr.5: 140671053-140671053 on Build GRCh38
rs57919623 Chr.5: 140671082-140671082 on Build GRCh38
rs550863751 Chr.5: 140671130-140671130 on Build GRCh38
rs1017594933 Chr.5: 140671083-140671083 on Build GRCh38
rs910629344 Chr.5: 140670864-140670864 on Build GRCh38
rs575478532 Chr.5: 140670706-140670706 on Build GRCh38
rs557980145 Chr.5: 140670727-140670727 on Build GRCh38
rs1037504948 Chr.5: 140671024-140671024 on Build GRCh38
rs559983053 Chr.5: 140671003-140671003 on Build GRCh38
rs546131944 Chr.5: 140670731-140670731 on Build GRCh38
rs780037673 Chr.5: 140671016-140671016 on Build GRCh38
rs560010755 Chr.5: 140671042-140671042 on Build GRCh38
rs1052762 Chr.5: 140671037-140671037 on Build GRCh38
rs562770214 Chr.5: 140670999-140670999 on Build GRCh38

View Full Product Details